Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54597302-54597547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT11
Entrez Gene ID
26027
Gene Name
acyl-CoA thioesterase 11
Gene Aliases
BFIT, STARD14, THEA, THEM1
UniGene
Hs.745173
Gene Chromosome Location
Chr.1: 54548134-54634744 on Build GRCh38
Transcripts
NM_015547.3 NM_147161.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3865799 c.746T>G p.V249G Chr.1: 54597397-54597397 on Build GRCh38
1238116 c.729G>A p.M243I Chr.1: 54597380-54597380 on Build GRCh38
6246879 c.730G>T p.A244S Chr.1: 54597381-54597381 on Build GRCh38
4893982 c.710C>T p.T237I Chr.1: 54597361-54597361 on Build GRCh38
3490858 c.764+1G>A p.? Chr.1: 54597416-54597416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377530147 Chr.1: 54597447-54597447 on Build GRCh38
rs756819499 Chr.1: 54597336-54597336 on Build GRCh38
rs186785371 Chr.1: 54597520-54597520 on Build GRCh38
rs779269340 Chr.1: 54597464-54597464 on Build GRCh38
rs533236726 Chr.1: 54597458-54597458 on Build GRCh38
rs774057335 Chr.1: 54597446-54597446 on Build GRCh38
rs370958601 Chr.1: 54597456-54597456 on Build GRCh38
rs772259284 Chr.1: 54597378-54597378 on Build GRCh38
rs763723890 Chr.1: 54597441-54597441 on Build GRCh38
rs751016639 Chr.1: 54597331-54597331 on Build GRCh38
rs749922686 Chr.1: 54597454-54597454 on Build GRCh38
rs761198115 Chr.1: 54597416-54597416 on Build GRCh38
rs771375820 Chr.1: 54597418-54597418 on Build GRCh38
rs72903807 Chr.1: 54597359-54597359 on Build GRCh38
rs2304304 Chr.1: 54597457-54597457 on Build GRCh38
rs746597931 Chr.1: 54597398-54597398 on Build GRCh38
rs773558097 Chr.1: 54597411-54597411 on Build GRCh38
rs767105163 Chr.1: 54597449-54597449 on Build GRCh38
rs770280405 Chr.1: 54597444-54597444 on Build GRCh38
rs993686355 Chr.1: 54597427-54597427 on Build GRCh38
rs377457947 Chr.1: 54597466-54597466 on Build GRCh38
rs780987042 Chr.1: 54597337-54597337 on Build GRCh38
rs779287804 Chr.1: 54597370-54597370 on Build GRCh38
rs756037646 Chr.1: 54597455-54597455 on Build GRCh38
rs777871990 Chr.1: 54597467-54597467 on Build GRCh38
rs924644644 Chr.1: 54597517-54597517 on Build GRCh38
rs933159248 Chr.1: 54597525-54597525 on Build GRCh38
rs550272648 Chr.1: 54597518-54597518 on Build GRCh38
rs776918747 Chr.1: 54597420-54597420 on Build GRCh38
rs750416407 Chr.1: 54597341-54597341 on Build GRCh38
rs759911661 Chr.1: 54597423-54597423 on Build GRCh38
rs757705395 Chr.1: 54597454-54597456 on Build GRCh38
rs748418687 Chr.1: 54597465-54597465 on Build GRCh38
rs779847979 Chr.1: 54597344-54597344 on Build GRCh38
rs768931947 Chr.1: 54597369-54597369 on Build GRCh38
rs777268718 Chr.1: 54597377-54597377 on Build GRCh38

View Full Product Details