Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36490072-36490345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAF6
Entrez Gene ID
7189
Gene Name
TNF receptor associated factor 6
Gene Aliases
MGC:3310, RNF85
UniGene
Hs.444172
Gene Chromosome Location
Chr.11: 36483767-36510313 on Build GRCh38
Transcripts
NM_004620.3 NM_145803.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5014988 c.1204C>T p.R402C Chr.11: 36490203-36490203 on Build GRCh38
926723 c.1214A>G p.N405S Chr.11: 36490193-36490193 on Build GRCh38
926722 c.1303C>A p.L435I Chr.11: 36490104-36490104 on Build GRCh38
3447041 c.1126C>T p.H376Y Chr.11: 36490281-36490281 on Build GRCh38
3447040 c.1266C>T p.L422L Chr.11: 36490141-36490141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949300138 Chr.11: 36490156-36490156 on Build GRCh38
rs777821743 Chr.11: 36490252-36490252 on Build GRCh38
rs761368102 Chr.11: 36490149-36490149 on Build GRCh38
rs536779653 Chr.11: 36490294-36490294 on Build GRCh38
rs764878622 Chr.11: 36490150-36490150 on Build GRCh38
rs144270884 Chr.11: 36490261-36490261 on Build GRCh38
rs199830912 Chr.11: 36490202-36490202 on Build GRCh38
rs916447705 Chr.11: 36490148-36490148 on Build GRCh38
rs747509379 Chr.11: 36490321-36490321 on Build GRCh38
rs746111041 Chr.11: 36490302-36490302 on Build GRCh38
rs34165477 Chr.11: 36490221-36490222 on Build GRCh38
rs754966271 Chr.11: 36490231-36490231 on Build GRCh38
rs187625425 Chr.11: 36490258-36490258 on Build GRCh38
rs200661146 Chr.11: 36490107-36490107 on Build GRCh38
rs772536661 Chr.11: 36490303-36490303 on Build GRCh38
rs776263136 Chr.11: 36490119-36490119 on Build GRCh38
rs750059919 Chr.11: 36490183-36490183 on Build GRCh38
rs776049763 Chr.11: 36490316-36490316 on Build GRCh38
rs751475632 Chr.11: 36490213-36490213 on Build GRCh38
rs548543739 Chr.11: 36490203-36490203 on Build GRCh38

View Full Product Details