Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 33575912-33576165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS12
Entrez Gene ID
81792
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 12
Gene Aliases
PRO4389
UniGene
Hs.12680
Gene Chromosome Location
Chr.5: 33523535-33892180 on Build GRCh38
Transcripts
NM_001324512.1 NM_030955.3 XM_017009905.1 XM_017009906.1 XM_017009907.1 XM_017009908.1 XM_017009909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3947437 c.3960A>T p.G1320G Chr.5: 33576066-33576066 on Build GRCh38
3015011 c.3958G>A p.G1320R Chr.5: 33576068-33576068 on Build GRCh38
4347611 c.3936C>T p.H1312H Chr.5: 33576090-33576090 on Build GRCh38
6492118 c.3895T>G p.L1299V Chr.5: 33576131-33576131 on Build GRCh38
6325148 c.3886G>A p.E1296K Chr.5: 33576140-33576140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181202879 Chr.5: 33576020-33576020 on Build GRCh38
rs767143559 Chr.5: 33576007-33576007 on Build GRCh38
rs754659898 Chr.5: 33576005-33576005 on Build GRCh38
rs186724231 Chr.5: 33576072-33576072 on Build GRCh38
rs145641172 Chr.5: 33576008-33576008 on Build GRCh38
rs758939488 Chr.5: 33576088-33576088 on Build GRCh38
rs534661620 Chr.5: 33576011-33576011 on Build GRCh38
rs371530789 Chr.5: 33576041-33576041 on Build GRCh38
rs35581318 Chr.5: 33576134-33576134 on Build GRCh38
rs386687017 Chr.5: 33576040-33576041 on Build GRCh38
rs774040470 Chr.5: 33576129-33576129 on Build GRCh38
rs767233472 Chr.5: 33576071-33576071 on Build GRCh38
rs748174765 Chr.5: 33576050-33576050 on Build GRCh38
rs760305077 Chr.5: 33576073-33576073 on Build GRCh38
rs374664813 Chr.5: 33576120-33576120 on Build GRCh38
rs368589714 Chr.5: 33576040-33576040 on Build GRCh38
rs369209539 Chr.5: 33576030-33576030 on Build GRCh38
rs775018098 Chr.5: 33576056-33576056 on Build GRCh38
rs757864544 Chr.5: 33576009-33576009 on Build GRCh38
rs774187264 Chr.5: 33576059-33576059 on Build GRCh38
rs763639359 Chr.5: 33576075-33576075 on Build GRCh38
rs141717162 Chr.5: 33576096-33576096 on Build GRCh38
rs1012406509 Chr.5: 33576069-33576069 on Build GRCh38
rs371276803 Chr.5: 33576023-33576023 on Build GRCh38
rs929703135 Chr.5: 33575960-33575960 on Build GRCh38
rs777891199 Chr.5: 33576111-33576111 on Build GRCh38
rs769811124 Chr.5: 33576108-33576108 on Build GRCh38
rs774255323 Chr.5: 33575953-33575953 on Build GRCh38
rs780453858 Chr.5: 33576031-33576031 on Build GRCh38
rs377708478 Chr.5: 33576121-33576121 on Build GRCh38
rs568403595 Chr.5: 33576089-33576089 on Build GRCh38
rs373735144 Chr.5: 33575970-33575970 on Build GRCh38
rs772207271 Chr.5: 33576068-33576068 on Build GRCh38
rs373650661 Chr.5: 33576035-33576035 on Build GRCh38
rs200551057 Chr.5: 33576054-33576054 on Build GRCh38
rs138603217 Chr.5: 33576095-33576095 on Build GRCh38
rs375647714 Chr.5: 33576049-33576049 on Build GRCh38
rs868243658 Chr.5: 33576084-33576084 on Build GRCh38
rs750951515 Chr.5: 33576076-33576076 on Build GRCh38
rs143099189 Chr.5: 33576090-33576090 on Build GRCh38
rs371610056 Chr.5: 33576057-33576057 on Build GRCh38
rs575651328 Chr.5: 33576004-33576004 on Build GRCh38
rs769752715 Chr.5: 33576053-33576053 on Build GRCh38
rs201770938 Chr.5: 33576127-33576127 on Build GRCh38

View Full Product Details