Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134562151-134562404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JADE2
Entrez Gene ID
23338
Gene Name
jade family PHD finger 2
Gene Aliases
JADE-2, PHF15
UniGene
Hs.732155
Gene Chromosome Location
Chr.5: 134524154-134583230 on Build GRCh38
Transcripts
NM_001289984.1 NM_001289985.1 NM_001308143.1 NM_015288.5 XM_005271945.1 XM_005271946.1 XM_005271948.4 XM_011543291.2 XM_017009282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4761597 c.848C>T p.P283L Chr.5: 134562363-134562363 on Build GRCh38
4791991 c.838C>G p.L280V Chr.5: 134562353-134562353 on Build GRCh38
6448228 c.693C>T p.Y231Y Chr.5: 134562208-134562208 on Build GRCh38
3428909 c.826C>T p.R276* Chr.5: 134562293-134562293 on Build GRCh38
3826775 c.898G>C p.E300Q Chr.5: 134562365-134562365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755201957 Chr.5: 134562188-134562188 on Build GRCh38
rs750269434 Chr.5: 134562327-134562327 on Build GRCh38
rs376207647 Chr.5: 134562221-134562221 on Build GRCh38
rs1035301807 Chr.5: 134562224-134562224 on Build GRCh38
rs1026512686 Chr.5: 134562196-134562196 on Build GRCh38
rs569336433 Chr.5: 134562223-134562223 on Build GRCh38
rs554945499 Chr.5: 134562249-134562249 on Build GRCh38
rs950894823 Chr.5: 134562215-134562215 on Build GRCh38
rs759975546 Chr.5: 134562356-134562356 on Build GRCh38
rs775232480 Chr.5: 134562294-134562294 on Build GRCh38
rs371379573 Chr.5: 134562169-134562169 on Build GRCh38
rs527890032 Chr.5: 134562228-134562228 on Build GRCh38
rs758526161 Chr.5: 134562209-134562209 on Build GRCh38
rs200918265 Chr.5: 134562229-134562229 on Build GRCh38
rs115088442 Chr.5: 134562343-134562343 on Build GRCh38
rs1016855153 Chr.5: 134562219-134562219 on Build GRCh38
rs752677295 Chr.5: 134562208-134562208 on Build GRCh38
rs199572513 Chr.5: 134562355-134562355 on Build GRCh38
rs754046759 Chr.5: 134562181-134562181 on Build GRCh38
rs745504684 Chr.5: 134562264-134562264 on Build GRCh38
rs959580111 Chr.5: 134562298-134562298 on Build GRCh38
rs753525400 Chr.5: 134562185-134562185 on Build GRCh38
rs972622070 Chr.5: 134562378-134562378 on Build GRCh38
rs953444401 Chr.5: 134562191-134562191 on Build GRCh38
rs370554814 Chr.5: 134562352-134562352 on Build GRCh38
rs773706007 Chr.5: 134562351-134562351 on Build GRCh38
rs374430070 Chr.5: 134562193-134562193 on Build GRCh38
rs777790775 Chr.5: 134562211-134562211 on Build GRCh38
rs768159397 Chr.5: 134562349-134562349 on Build GRCh38
rs769295036 Chr.5: 134562283-134562283 on Build GRCh38
rs114733528 Chr.5: 134562250-134562250 on Build GRCh38
rs766795436 Chr.5: 134562176-134562176 on Build GRCh38

View Full Product Details