Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 112509-112770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPRL3
Entrez Gene ID
8131
Gene Name
NPR3 like, GATOR1 complex subunit
Gene Aliases
C16orf35, CGTHBA, HS-40, MARE, NPR3, RMD11
UniGene
Hs.19699
Gene Chromosome Location
Chr.16: 85801-138698 on Build GRCh38
Transcripts
NM_001039476.2 NM_001077350.2 NM_001243247.1 NM_001243248.1 NM_001243249.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5054678 c.440G>A p.R147H Chr.16: 112729-112729 on Build GRCh38
6212347 c.455T>A p.V152E Chr.16: 112714-112714 on Build GRCh38
2127456 c.516G>A p.A172A Chr.16: 112653-112653 on Build GRCh38
1375749 c.408G>A p.P136P Chr.16: 112761-112761 on Build GRCh38
1629795 c.399C>T p.N133N Chr.16: 112770-112770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367592989 Chr.16: 112722-112722 on Build GRCh38
rs978342878 Chr.16: 112600-112600 on Build GRCh38
rs762922931 Chr.16: 112699-112699 on Build GRCh38
rs370299128 Chr.16: 112693-112693 on Build GRCh38
rs774187145 Chr.16: 112704-112704 on Build GRCh38
rs775239262 Chr.16: 112697-112697 on Build GRCh38
rs775513193 Chr.16: 112605-112605 on Build GRCh38
rs959374963 Chr.16: 112741-112741 on Build GRCh38
rs558045556 Chr.16: 112716-112716 on Build GRCh38
rs140092767 Chr.16: 112635-112635 on Build GRCh38
rs201069648 Chr.16: 112727-112727 on Build GRCh38
rs758642036 Chr.16: 112653-112653 on Build GRCh38
rs539649449 Chr.16: 112715-112715 on Build GRCh38
rs937579594 Chr.16: 112563-112563 on Build GRCh38
rs766649807 Chr.16: 112721-112721 on Build GRCh38
rs535609281 Chr.16: 112612-112612 on Build GRCh38
rs776092921 Chr.16: 112581-112581 on Build GRCh38
rs770619319 Chr.16: 112576-112576 on Build GRCh38
rs764927474 Chr.16: 112742-112742 on Build GRCh38
rs373390759 Chr.16: 112606-112606 on Build GRCh38
rs753280871 Chr.16: 112652-112652 on Build GRCh38
rs370796954 Chr.16: 112613-112613 on Build GRCh38
rs374060440 Chr.16: 112703-112703 on Build GRCh38
rs569119130 Chr.16: 112670-112670 on Build GRCh38
rs568519242 Chr.16: 112599-112599 on Build GRCh38
rs945494998 Chr.16: 112591-112591 on Build GRCh38
rs769300436 Chr.16: 112694-112694 on Build GRCh38
rs770570210 Chr.16: 112675-112675 on Build GRCh38
rs778177582 Chr.16: 112654-112654 on Build GRCh38
rs546697735 Chr.16: 112595-112595 on Build GRCh38
rs371406653 Chr.16: 112737-112737 on Build GRCh38
rs376237512 Chr.16: 112680-112680 on Build GRCh38
rs528432093 Chr.16: 112562-112562 on Build GRCh38
rs755120736 Chr.16: 112634-112634 on Build GRCh38
rs766774400 Chr.16: 112628-112628 on Build GRCh38
rs753953832 Chr.16: 112632-112632 on Build GRCh38
rs202129021 Chr.16: 112730-112730 on Build GRCh38

View Full Product Details