Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 115126750-115127023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRCH2
Entrez Gene ID
57631
Gene Name
leucine rich repeats and calponin homology domain containing 2
Gene Aliases
dA204F4.4
UniGene
Hs.65366
Gene Chromosome Location
Chr.X: 115110611-115234082 on Build GRCh38
Transcripts
NM_001243963.1 NM_020871.3 XM_006724724.3

SNP Detail

SNP ID Chromosome Location
rs781874713 Chr.X: 115126923-115126923 on Build GRCh38
rs1001282674 Chr.X: 115126780-115126780 on Build GRCh38
rs781788066 Chr.X: 115126890-115126890 on Build GRCh38
rs957024548 Chr.X: 115126824-115126824 on Build GRCh38
rs782485146 Chr.X: 115126901-115126901 on Build GRCh38
rs918988187 Chr.X: 115126989-115126989 on Build GRCh38
rs181443053 Chr.X: 115126918-115126918 on Build GRCh38
rs369287642 Chr.X: 115126877-115126877 on Build GRCh38
rs941664233 Chr.X: 115126920-115126920 on Build GRCh38
rs980696682 Chr.X: 115126908-115126908 on Build GRCh38
rs781988224 Chr.X: 115126809-115126809 on Build GRCh38
rs148716926 Chr.X: 115126874-115126874 on Build GRCh38
rs1031046228 Chr.X: 115126803-115126803 on Build GRCh38
rs782010407 Chr.X: 115126833-115126833 on Build GRCh38

View Full Product Details