Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163456898-163457123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDCD2
Entrez Gene ID
134492
Gene Name
NudC domain containing 2
Gene Aliases
NudCL2
UniGene
Hs.140443
Gene Chromosome Location
Chr.5: 163453580-163460137 on Build GRCh38
Transcripts
NM_145266.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6418014 c.239A>T p.E80V Chr.5: 163457080-163457080 on Build GRCh38
1733524 c.239-3delT p.? Chr.5: 163457083-163457083 on Build GRCh38
5060901 c.330G>A p.A110A Chr.5: 163456989-163456989 on Build GRCh38
6170310 c.373G>C p.E125Q Chr.5: 163456946-163456946 on Build GRCh38
5060900 c.336T>A p.P112P Chr.5: 163456983-163456983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368381995 Chr.5: 163456993-163456993 on Build GRCh38
rs113515886 Chr.5: 163457093-163457094 on Build GRCh38
rs866182845 Chr.5: 163457033-163457033 on Build GRCh38
rs764231026 Chr.5: 163456989-163456989 on Build GRCh38
rs373523422 Chr.5: 163457040-163457043 on Build GRCh38
rs762060193 Chr.5: 163457097-163457097 on Build GRCh38
rs747274702 Chr.5: 163457039-163457042 on Build GRCh38
rs543711948 Chr.5: 163457063-163457063 on Build GRCh38
rs907570590 Chr.5: 163456996-163456996 on Build GRCh38
rs773237977 Chr.5: 163456957-163456957 on Build GRCh38
rs749002124 Chr.5: 163457091-163457092 on Build GRCh38
rs372168863 Chr.5: 163457070-163457070 on Build GRCh38
rs369487816 Chr.5: 163457010-163457010 on Build GRCh38
rs769395107 Chr.5: 163457083-163457084 on Build GRCh38
rs746568322 Chr.5: 163457032-163457032 on Build GRCh38
rs150464483 Chr.5: 163457031-163457031 on Build GRCh38
rs201931869 Chr.5: 163457093-163457093 on Build GRCh38
rs767606090 Chr.5: 163456968-163456968 on Build GRCh38
rs932595253 Chr.5: 163457004-163457004 on Build GRCh38
rs150010030 Chr.5: 163457067-163457067 on Build GRCh38
rs200190066 Chr.5: 163457091-163457091 on Build GRCh38
rs747457697 Chr.5: 163457083-163457083 on Build GRCh38
rs755112083 Chr.5: 163456946-163456947 on Build GRCh38
rs142159368 Chr.5: 163457007-163457007 on Build GRCh38
rs778317662 Chr.5: 163457082-163457083 on Build GRCh38
rs140642151 Chr.5: 163456998-163456998 on Build GRCh38
rs376036154 Chr.5: 163456984-163456984 on Build GRCh38
rs779401727 Chr.5: 163457092-163457093 on Build GRCh38
rs774345989 Chr.5: 163457087-163457087 on Build GRCh38
rs370580721 Chr.5: 163457049-163457049 on Build GRCh38
rs769674467 Chr.5: 163457058-163457058 on Build GRCh38
rs761634543 Chr.5: 163456950-163456950 on Build GRCh38
rs752773367 Chr.5: 163456976-163456976 on Build GRCh38
rs762831815 Chr.5: 163456967-163456967 on Build GRCh38
rs758910013 Chr.5: 163457041-163457042 on Build GRCh38
rs371791584 Chr.5: 163456944-163456944 on Build GRCh38
rs150776333 Chr.5: 163457009-163457009 on Build GRCh38
rs768250901 Chr.5: 163456938-163456938 on Build GRCh38
rs576383191 Chr.5: 163457035-163457036 on Build GRCh38
rs147218174 Chr.5: 163456956-163456956 on Build GRCh38
rs937134231 Chr.5: 163457060-163457060 on Build GRCh38
rs747827504 Chr.5: 163457050-163457050 on Build GRCh38
rs78759977 Chr.5: 163457092-163457092 on Build GRCh38
rs139292728 Chr.5: 163457042-163457042 on Build GRCh38

View Full Product Details