Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52881852-52882123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF320
Entrez Gene ID
162967
Gene Name
zinc finger protein 320
Gene Aliases
ZFPL
UniGene
Hs.369632
Gene Chromosome Location
Chr.19: 52863790-52897695 on Build GRCh38
Transcripts
NM_207333.2 XM_006723059.3 XM_017026393.1 XM_017026394.1 XM_017026395.1 XM_017026396.1 XM_017026397.1 XM_017026398.1 XM_017026399.1 XM_017026400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3990243 c.172T>C p.L58L Chr.19: 52881954-52881954 on Build GRCh38
440194 c.257G>C p.C86S Chr.19: 52881869-52881869 on Build GRCh38
364480 c.225G>A p.Q75Q Chr.19: 52881901-52881901 on Build GRCh38
3960408 c.241C>A p.H81N Chr.19: 52881885-52881885 on Build GRCh38
7238589 c.241C>T p.H81Y Chr.19: 52881885-52881885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576930162 Chr.19: 52882064-52882064 on Build GRCh38
rs923172775 Chr.19: 52881875-52881875 on Build GRCh38
rs138794752 Chr.19: 52881881-52881881 on Build GRCh38
rs146633824 Chr.19: 52882047-52882047 on Build GRCh38
rs775603708 Chr.19: 52881980-52881980 on Build GRCh38
rs996721800 Chr.19: 52881956-52881956 on Build GRCh38
rs762681807 Chr.19: 52882022-52882022 on Build GRCh38
rs965215696 Chr.19: 52881929-52881929 on Build GRCh38
rs766160221 Chr.19: 52882031-52882031 on Build GRCh38
rs375096963 Chr.19: 52881983-52881983 on Build GRCh38
rs750220999 Chr.19: 52882038-52882038 on Build GRCh38
rs776352204 Chr.19: 52882074-52882078 on Build GRCh38
rs967308528 Chr.19: 52882062-52882062 on Build GRCh38
rs754569604 Chr.19: 52881927-52881927 on Build GRCh38
rs779402897 Chr.19: 52881942-52881942 on Build GRCh38
rs761800624 Chr.19: 52881999-52881999 on Build GRCh38
rs187853286 Chr.19: 52882095-52882095 on Build GRCh38
rs907491491 Chr.19: 52882065-52882065 on Build GRCh38
rs773104391 Chr.19: 52882006-52882006 on Build GRCh38
rs371953748 Chr.19: 52881964-52881964 on Build GRCh38
rs751211351 Chr.19: 52881916-52881916 on Build GRCh38
rs140549030 Chr.19: 52881904-52881904 on Build GRCh38
rs201067087 Chr.19: 52881885-52881885 on Build GRCh38
rs184513423 Chr.19: 52882067-52882067 on Build GRCh38
rs761679799 Chr.19: 52881882-52881882 on Build GRCh38
rs746188412 Chr.19: 52881959-52881959 on Build GRCh38
rs953615919 Chr.19: 52881997-52881997 on Build GRCh38
rs1050131264 Chr.19: 52881898-52881898 on Build GRCh38
rs765134537 Chr.19: 52881883-52881883 on Build GRCh38
rs780786172 Chr.19: 52881928-52881928 on Build GRCh38
rs749830981 Chr.19: 52881932-52881932 on Build GRCh38
rs1047766772 Chr.19: 52882060-52882060 on Build GRCh38
rs765052162 Chr.19: 52882000-52882000 on Build GRCh38
rs369753383 Chr.19: 52882055-52882056 on Build GRCh38
rs766162690 Chr.19: 52881912-52881913 on Build GRCh38
rs187983315 Chr.19: 52881893-52881893 on Build GRCh38
rs757691626 Chr.19: 52881940-52881940 on Build GRCh38

View Full Product Details