Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32210004-32210267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POFUT1
Entrez Gene ID
23509
Gene Name
protein O-fucosyltransferase 1
Gene Aliases
DDD2, FUT12, O-FUT, O-Fuc-T, O-FucT-1, OFUCT1
UniGene
Hs.472409
Gene Chromosome Location
Chr.20: 32207822-32238664 on Build GRCh38
Transcripts
NM_015352.1 NM_172236.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6046894 c.203C>T p.P68L Chr.20: 32210149-32210149 on Build GRCh38
1153903 c.210G>T p.W70C Chr.20: 32210156-32210156 on Build GRCh38
1725924 c.160T>C p.S54P Chr.20: 32210106-32210106 on Build GRCh38
4848378 c.195G>C p.L65F Chr.20: 32210141-32210141 on Build GRCh38
1190235 c.212T>C p.I71T Chr.20: 32210158-32210158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762752243 Chr.20: 32210095-32210095 on Build GRCh38
rs370909689 Chr.20: 32210091-32210091 on Build GRCh38
rs753456199 Chr.20: 32210031-32210031 on Build GRCh38
rs572332685 Chr.20: 32210211-32210211 on Build GRCh38
rs748521046 Chr.20: 32210057-32210057 on Build GRCh38
rs770942717 Chr.20: 32210090-32210090 on Build GRCh38
rs775625035 Chr.20: 32210094-32210094 on Build GRCh38
rs777694790 Chr.20: 32210023-32210023 on Build GRCh38
rs74183482 Chr.20: 32210026-32210026 on Build GRCh38
rs759801588 Chr.20: 32210092-32210092 on Build GRCh38
rs745942782 Chr.20: 32210191-32210191 on Build GRCh38
rs780713516 Chr.20: 32210045-32210045 on Build GRCh38
rs767489103 Chr.20: 32210212-32210212 on Build GRCh38
rs113552438 Chr.20: 32210093-32210093 on Build GRCh38
rs771446102 Chr.20: 32210181-32210181 on Build GRCh38
rs758853318 Chr.20: 32210177-32210177 on Build GRCh38
rs142484235 Chr.20: 32210173-32210173 on Build GRCh38
rs756741372 Chr.20: 32210034-32210034 on Build GRCh38
rs779481012 Chr.20: 32210056-32210056 on Build GRCh38
rs776630840 Chr.20: 32210186-32210186 on Build GRCh38
rs371242837 Chr.20: 32210226-32210226 on Build GRCh38
rs757192494 Chr.20: 32210110-32210110 on Build GRCh38
rs769551690 Chr.20: 32210053-32210053 on Build GRCh38
rs754641501 Chr.20: 32210229-32210229 on Build GRCh38
rs774177053 Chr.20: 32210200-32210200 on Build GRCh38
rs766989038 Chr.20: 32210117-32210117 on Build GRCh38
rs367609340 Chr.20: 32210225-32210225 on Build GRCh38
rs772495633 Chr.20: 32210065-32210065 on Build GRCh38
rs553681367 Chr.20: 32210193-32210193 on Build GRCh38
rs73903578 Chr.20: 32210213-32210213 on Build GRCh38
rs751482931 Chr.20: 32210106-32210106 on Build GRCh38
rs763389557 Chr.20: 32210052-32210052 on Build GRCh38
rs763902006 Chr.20: 32210099-32210099 on Build GRCh38
rs773602865 Chr.20: 32210072-32210072 on Build GRCh38
rs148829108 Chr.20: 32210134-32210134 on Build GRCh38
rs761191707 Chr.20: 32210073-32210073 on Build GRCh38
rs779855384 Chr.20: 32210161-32210161 on Build GRCh38
rs538485488 Chr.20: 32210157-32210157 on Build GRCh38
rs755899831 Chr.20: 32210138-32210138 on Build GRCh38
rs778155129 Chr.20: 32210180-32210180 on Build GRCh38
rs763123143 Chr.20: 32210192-32210192 on Build GRCh38
rs753573556 Chr.20: 32210228-32210228 on Build GRCh38

View Full Product Details