Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 13850120-13850389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEL1L2
Entrez Gene ID
80343
Gene Name
SEL1L2 ERAD E3 ligase adaptor subunit
Gene Aliases
C20orf50
UniGene
Hs.590879
Gene Chromosome Location
Chr.20: 13849188-13995333 on Build GRCh38
Transcripts
NM_001271539.1 NR_073206.1 NR_073207.1 XM_006723646.2 XM_006723647.2 XM_006723648.2 XM_006723650.2 XM_006723651.2 XM_006723652.2 XM_006723654.3 XM_011529374.1 XM_011529375.1 XM_011529376.1 XM_011529377.1 XM_011529378.1 XM_011529379.1 XM_017028084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
222397 c.1845C>A p.Y615* Chr.20: 13850293-13850293 on Build GRCh38
5914313 c.1927C>T p.R643W Chr.20: 13850211-13850211 on Build GRCh38
5906449 c.1868C>T p.P623L Chr.20: 13850270-13850270 on Build GRCh38
6195873 c.1909G>A p.E637K Chr.20: 13850229-13850229 on Build GRCh38
375436 c.1899C>A p.V633V Chr.20: 13850239-13850239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201001973 Chr.20: 13850221-13850221 on Build GRCh38
rs746232756 Chr.20: 13850241-13850241 on Build GRCh38
rs950616189 Chr.20: 13850154-13850154 on Build GRCh38
rs779707948 Chr.20: 13850243-13850243 on Build GRCh38
rs749038886 Chr.20: 13850344-13850344 on Build GRCh38
rs757712501 Chr.20: 13850307-13850307 on Build GRCh38
rs778109667 Chr.20: 13850234-13850234 on Build GRCh38
rs140015347 Chr.20: 13850293-13850293 on Build GRCh38
rs780706643 Chr.20: 13850203-13850203 on Build GRCh38
rs755102295 Chr.20: 13850169-13850169 on Build GRCh38
rs1044147650 Chr.20: 13850225-13850225 on Build GRCh38
rs761870351 Chr.20: 13850272-13850272 on Build GRCh38
rs752836544 Chr.20: 13850177-13850177 on Build GRCh38
rs752173370 Chr.20: 13850235-13850235 on Build GRCh38
rs771917871 Chr.20: 13850275-13850275 on Build GRCh38
rs964734684 Chr.20: 13850346-13850346 on Build GRCh38
rs376034120 Chr.20: 13850215-13850215 on Build GRCh38
rs778367315 Chr.20: 13850186-13850186 on Build GRCh38
rs778430871 Chr.20: 13850354-13850354 on Build GRCh38
rs192865162 Chr.20: 13850276-13850276 on Build GRCh38
rs750733078 Chr.20: 13850337-13850337 on Build GRCh38
rs567838713 Chr.20: 13850268-13850268 on Build GRCh38
rs552223044 Chr.20: 13850263-13850263 on Build GRCh38
rs754436210 Chr.20: 13850350-13850350 on Build GRCh38
rs779652854 Chr.20: 13850339-13850339 on Build GRCh38
rs531215174 Chr.20: 13850210-13850210 on Build GRCh38
rs758908921 Chr.20: 13850185-13850185 on Build GRCh38
rs373069541 Chr.20: 13850211-13850211 on Build GRCh38
rs769453517 Chr.20: 13850209-13850209 on Build GRCh38
rs771368420 Chr.20: 13850192-13850192 on Build GRCh38
rs747502974 Chr.20: 13850242-13850242 on Build GRCh38
rs747314495 Chr.20: 13850191-13850191 on Build GRCh38
rs765534191 Chr.20: 13850287-13850287 on Build GRCh38
rs745581158 Chr.20: 13850205-13850205 on Build GRCh38
rs374088645 Chr.20: 13850355-13850355 on Build GRCh38
rs377287167 Chr.20: 13850332-13850334 on Build GRCh38
rs765561040 Chr.20: 13850228-13850228 on Build GRCh38
rs760990335 Chr.20: 13850328-13850330 on Build GRCh38
rs768038881 Chr.20: 13850316-13850316 on Build GRCh38
rs766485229 Chr.20: 13850148-13850148 on Build GRCh38
rs774154645 Chr.20: 13850269-13850269 on Build GRCh38
rs200801217 Chr.20: 13850237-13850237 on Build GRCh38
rs752785125 Chr.20: 13850229-13850229 on Build GRCh38
rs867244978 Chr.20: 13850362-13850362 on Build GRCh38
rs895470197 Chr.20: 13850219-13850219 on Build GRCh38
rs377665267 Chr.20: 13850286-13850286 on Build GRCh38
rs762935961 Chr.20: 13850292-13850292 on Build GRCh38
rs754101601 Chr.20: 13850222-13850222 on Build GRCh38
rs750604079 Chr.20: 13850325-13850327 on Build GRCh38
rs758553223 Chr.20: 13850230-13850230 on Build GRCh38
rs775894281 Chr.20: 13850290-13850290 on Build GRCh38
rs754150785 Chr.20: 13850149-13850149 on Build GRCh38
rs748777741 Chr.20: 13850250-13850250 on Build GRCh38
rs779189961 Chr.20: 13850176-13850176 on Build GRCh38

View Full Product Details