Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 103640050-103640321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAB2
Entrez Gene ID
55576
Gene Name
stabilin 2
Gene Aliases
FEEL2, FELE-2, FELL2, FEX2, HARE, SCARH1
UniGene
Hs.408249
Gene Chromosome Location
Chr.12: 103587250-103766724 on Build GRCh38
Transcripts
NM_017564.9 XM_011538537.2 XM_011538538.2 XM_017019585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5373995 c.912C>T p.H304H Chr.12: 103640128-103640128 on Build GRCh38
3967928 c.993G>A p.P331P Chr.12: 103640209-103640209 on Build GRCh38
4987248 c.972T>G p.D324E Chr.12: 103640188-103640188 on Build GRCh38
4759672 c.1019C>T p.T340I Chr.12: 103640235-103640235 on Build GRCh38
5556514 c.999T>C p.H333H Chr.12: 103640215-103640215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753172320 Chr.12: 103640229-103640229 on Build GRCh38
rs755546166 Chr.12: 103640136-103640136 on Build GRCh38
rs769417312 Chr.12: 103640088-103640088 on Build GRCh38
rs369059792 Chr.12: 103640173-103640173 on Build GRCh38
rs12319476 Chr.12: 103640132-103640132 on Build GRCh38
rs200956819 Chr.12: 103640209-103640209 on Build GRCh38
rs780562898 Chr.12: 103640254-103640257 on Build GRCh38
rs144975292 Chr.12: 103640158-103640158 on Build GRCh38
rs761913490 Chr.12: 103640188-103640188 on Build GRCh38
rs942325779 Chr.12: 103640141-103640141 on Build GRCh38
rs369288429 Chr.12: 103640255-103640255 on Build GRCh38
rs373857284 Chr.12: 103640261-103640261 on Build GRCh38
rs766890838 Chr.12: 103640113-103640113 on Build GRCh38
rs745609486 Chr.12: 103640240-103640240 on Build GRCh38
rs200010553 Chr.12: 103640131-103640131 on Build GRCh38
rs781551820 Chr.12: 103640149-103640149 on Build GRCh38
rs891784423 Chr.12: 103640094-103640094 on Build GRCh38
rs747916924 Chr.12: 103640201-103640201 on Build GRCh38
rs375893662 Chr.12: 103640218-103640218 on Build GRCh38
rs536590694 Chr.12: 103640108-103640108 on Build GRCh38
rs115840458 Chr.12: 103640270-103640270 on Build GRCh38
rs138889157 Chr.12: 103640272-103640272 on Build GRCh38
rs193098116 Chr.12: 103640236-103640236 on Build GRCh38
rs374001698 Chr.12: 103640159-103640159 on Build GRCh38
rs752007498 Chr.12: 103640294-103640294 on Build GRCh38
rs774358763 Chr.12: 103640165-103640165 on Build GRCh38
rs764080749 Chr.12: 103640119-103640119 on Build GRCh38
rs769902788 Chr.12: 103640252-103640252 on Build GRCh38
rs377527797 Chr.12: 103640237-103640237 on Build GRCh38
rs781686028 Chr.12: 103640227-103640227 on Build GRCh38
rs573879048 Chr.12: 103640292-103640292 on Build GRCh38
rs774411687 Chr.12: 103640111-103640111 on Build GRCh38
rs753893082 Chr.12: 103640122-103640122 on Build GRCh38
rs755307975 Chr.12: 103640213-103640213 on Build GRCh38
rs747500222 Chr.12: 103640134-103640134 on Build GRCh38
rs1023272794 Chr.12: 103640273-103640273 on Build GRCh38
rs777209439 Chr.12: 103640231-103640231 on Build GRCh38
rs145026397 Chr.12: 103640250-103640250 on Build GRCh38
rs747751345 Chr.12: 103640080-103640080 on Build GRCh38
rs149929067 Chr.12: 103640208-103640208 on Build GRCh38
rs769207192 Chr.12: 103640152-103640152 on Build GRCh38
rs759607794 Chr.12: 103640118-103640118 on Build GRCh38
rs369631239 Chr.12: 103640232-103640232 on Build GRCh38
rs750619431 Chr.12: 103640202-103640202 on Build GRCh38
rs372004718 Chr.12: 103640123-103640123 on Build GRCh38
rs765275209 Chr.12: 103640190-103640190 on Build GRCh38
rs190090440 Chr.12: 103640175-103640175 on Build GRCh38
rs772748750 Chr.12: 103640100-103640100 on Build GRCh38
rs758430862 Chr.12: 103640077-103640080 on Build GRCh38
rs184603034 Chr.12: 103640109-103640109 on Build GRCh38
rs368200388 Chr.12: 103640102-103640102 on Build GRCh38
rs747632593 Chr.12: 103640151-103640151 on Build GRCh38
rs543702395 Chr.12: 103640093-103640093 on Build GRCh38
rs765451081 Chr.12: 103640129-103640129 on Build GRCh38

View Full Product Details