Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29259897-29260150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHBDD3
Entrez Gene ID
25807
Gene Name
rhomboid domain containing 3
Gene Aliases
C22orf3, HS984G1A, PTAG
UniGene
Hs.106730
Gene Chromosome Location
Chr.22: 29259855-29268196 on Build GRCh38
Transcripts
NM_012265.1 XM_006724224.3 XM_011530107.2 XM_017028749.1 XM_017028750.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6421779 c.1142A>T p.E381V Chr.22: 29260079-29260079 on Build GRCh38
4436519 c.1141G>A p.E381K Chr.22: 29260080-29260080 on Build GRCh38
1033117 c.1079T>A p.V360D Chr.22: 29260142-29260142 on Build GRCh38
1415475 c.1072C>T p.L358L Chr.22: 29260149-29260149 on Build GRCh38
3553104 c.1117G>A p.G373R Chr.22: 29260104-29260104 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051653303 Chr.22: 29260039-29260039 on Build GRCh38
rs750288267 Chr.22: 29260106-29260106 on Build GRCh38
rs768193317 Chr.22: 29260080-29260080 on Build GRCh38
rs375171092 Chr.22: 29260081-29260081 on Build GRCh38
rs867183253 Chr.22: 29260127-29260127 on Build GRCh38
rs766327953 Chr.22: 29260124-29260124 on Build GRCh38
rs968266907 Chr.22: 29260033-29260033 on Build GRCh38
rs547853294 Chr.22: 29259993-29259993 on Build GRCh38
rs1020813053 Chr.22: 29259994-29259994 on Build GRCh38
rs760570839 Chr.22: 29260109-29260109 on Build GRCh38
rs112726426 Chr.22: 29260066-29260066 on Build GRCh38
rs761174134 Chr.22: 29260084-29260084 on Build GRCh38
rs767495403 Chr.22: 29260086-29260086 on Build GRCh38
rs529738381 Chr.22: 29259992-29259992 on Build GRCh38
rs1000680622 Chr.22: 29260059-29260059 on Build GRCh38

View Full Product Details