Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 56350731-56350970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKFN1
Entrez Gene ID
162282
Gene Name
ankyrin repeat and fibronectin type III domain containing 1
Gene Aliases
-
UniGene
Hs.673040
Gene Chromosome Location
Chr.17: 56110935-56517016 on Build GRCh38
Transcripts
NM_153228.2 XM_006721728.3 XM_011524428.2 XM_011524429.2 XM_011524430.2 XM_011524431.2 XM_017024263.1 XM_017024264.1 XM_017024265.1 XM_017024266.1 XM_017024267.1 XM_017024268.1 XM_017024269.1 XM_017024270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
309123 c.397G>T p.V133L Chr.17: 56350965-56350965 on Build GRCh38
1610505 c.218C>T p.T73M Chr.17: 56350786-56350786 on Build GRCh38
3194339 c.204T>A p.C68* Chr.17: 56350772-56350772 on Build GRCh38
3194340 c.206G>A p.R69H Chr.17: 56350774-56350774 on Build GRCh38
4269122 c.210G>A p.V70V Chr.17: 56350778-56350778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022755635 Chr.17: 56350895-56350895 on Build GRCh38
rs200633862 Chr.17: 56350910-56350910 on Build GRCh38
rs778609641 Chr.17: 56350902-56350902 on Build GRCh38
rs755628674 Chr.17: 56350879-56350879 on Build GRCh38
rs746044298 Chr.17: 56350821-56350821 on Build GRCh38
rs954800788 Chr.17: 56350905-56350905 on Build GRCh38
rs892200206 Chr.17: 56350809-56350809 on Build GRCh38
rs775638491 Chr.17: 56350831-56350831 on Build GRCh38
rs751215331 Chr.17: 56350774-56350774 on Build GRCh38
rs768740920 Chr.17: 56350923-56350923 on Build GRCh38
rs898975164 Chr.17: 56350869-56350869 on Build GRCh38
rs566437342 Chr.17: 56350795-56350795 on Build GRCh38
rs183282741 Chr.17: 56350911-56350911 on Build GRCh38
rs368152107 Chr.17: 56350860-56350860 on Build GRCh38
rs773925251 Chr.17: 56350935-56350935 on Build GRCh38
rs201436645 Chr.17: 56350850-56350850 on Build GRCh38
rs775181306 Chr.17: 56350950-56350950 on Build GRCh38
rs778713948 Chr.17: 56350819-56350819 on Build GRCh38
rs768663674 Chr.17: 56350834-56350834 on Build GRCh38
rs777160054 Chr.17: 56350836-56350836 on Build GRCh38
rs763556266 Chr.17: 56350882-56350882 on Build GRCh38
rs376262477 Chr.17: 56350828-56350828 on Build GRCh38
rs770430594 Chr.17: 56350931-56350931 on Build GRCh38
rs765737413 Chr.17: 56350773-56350773 on Build GRCh38
rs141896298 Chr.17: 56350786-56350786 on Build GRCh38
rs772150910 Chr.17: 56350898-56350910 on Build GRCh38
rs753771498 Chr.17: 56350887-56350887 on Build GRCh38
rs1026430282 Chr.17: 56350888-56350888 on Build GRCh38
rs200776343 Chr.17: 56350827-56350827 on Build GRCh38
rs749236128 Chr.17: 56350802-56350802 on Build GRCh38
rs755647296 Chr.17: 56350797-56350797 on Build GRCh38
rs904791065 Chr.17: 56350780-56350780 on Build GRCh38
rs765645893 Chr.17: 56350859-56350859 on Build GRCh38
rs770686590 Chr.17: 56350806-56350806 on Build GRCh38
rs150654625 Chr.17: 56350942-56350942 on Build GRCh38
rs745655072 Chr.17: 56350906-56350906 on Build GRCh38
rs747389410 Chr.17: 56350800-56350800 on Build GRCh38
rs993596357 Chr.17: 56350884-56350884 on Build GRCh38
rs555482403 Chr.17: 56350928-56350928 on Build GRCh38
rs757211915 Chr.17: 56350892-56350892 on Build GRCh38
rs767276313 Chr.17: 56350861-56350861 on Build GRCh38
rs370848906 Chr.17: 56350787-56350787 on Build GRCh38
rs748606806 Chr.17: 56350929-56350929 on Build GRCh38
rs747095155 Chr.17: 56350832-56350832 on Build GRCh38
rs111831672 Chr.17: 56350862-56350862 on Build GRCh38
rs538479637 Chr.17: 56350855-56350855 on Build GRCh38
rs763397126 Chr.17: 56350941-56350941 on Build GRCh38

View Full Product Details