Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34620324-34620551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN3
Entrez Gene ID
11258
Gene Name
dynactin subunit 3
Gene Aliases
DCTN-22, DCTN22
UniGene
Hs.511768
Gene Chromosome Location
Chr.9: 34613545-34620523 on Build GRCh38
Transcripts
NM_001281425.1 NM_001281426.1 NM_001281427.1 NM_007234.4 NM_024348.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7275852 c.13A>G p.T5A Chr.9: 34620452-34620452 on Build GRCh38
6517051 c.38G>T p.R13L Chr.9: 34620427-34620427 on Build GRCh38
6517048 c.38G>T p.R13L Chr.9: 34620427-34620427 on Build GRCh38
6517055 c.21G>T p.L7F Chr.9: 34620444-34620444 on Build GRCh38
6517052 c.21G>T p.L7F Chr.9: 34620444-34620444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754696482 Chr.9: 34620346-34620346 on Build GRCh38
rs532890244 Chr.9: 34620498-34620498 on Build GRCh38
rs796583645 Chr.9: 34620524-34620524 on Build GRCh38
rs375071063 Chr.9: 34620353-34620353 on Build GRCh38
rs961742356 Chr.9: 34620442-34620442 on Build GRCh38
rs755360360 Chr.9: 34620499-34620499 on Build GRCh38
rs765006029 Chr.9: 34620453-34620453 on Build GRCh38
rs114192026 Chr.9: 34620485-34620485 on Build GRCh38
rs1052973998 Chr.9: 34620481-34620481 on Build GRCh38
rs1027421552 Chr.9: 34620454-34620454 on Build GRCh38
rs752277002 Chr.9: 34620458-34620458 on Build GRCh38
rs10972171 Chr.9: 34620504-34620504 on Build GRCh38
rs1019103633 Chr.9: 34620480-34620480 on Build GRCh38
rs770810056 Chr.9: 34620447-34620447 on Build GRCh38
rs777181060 Chr.9: 34620471-34620471 on Build GRCh38
rs370090780 Chr.9: 34620474-34620474 on Build GRCh38
rs11546834 Chr.9: 34620443-34620443 on Build GRCh38
rs778682603 Chr.9: 34620347-34620347 on Build GRCh38
rs35734048 Chr.9: 34620447-34620447 on Build GRCh38
rs757840840 Chr.9: 34620377-34620377 on Build GRCh38
rs760722293 Chr.9: 34620441-34620441 on Build GRCh38
rs924582756 Chr.9: 34620526-34620526 on Build GRCh38
rs768906077 Chr.9: 34620405-34620405 on Build GRCh38
rs759374476 Chr.9: 34620452-34620452 on Build GRCh38
rs766366712 Chr.9: 34620445-34620445 on Build GRCh38
rs746372465 Chr.9: 34620388-34620388 on Build GRCh38
rs770247400 Chr.9: 34620399-34620399 on Build GRCh38
rs908947675 Chr.9: 34620438-34620438 on Build GRCh38
rs758008889 Chr.9: 34620469-34620469 on Build GRCh38
rs984533655 Chr.9: 34620435-34620435 on Build GRCh38
rs772060854 Chr.9: 34620428-34620428 on Build GRCh38
rs781751202 Chr.9: 34620384-34620384 on Build GRCh38
rs762013693 Chr.9: 34620419-34620419 on Build GRCh38
rs367782478 Chr.9: 34620503-34620503 on Build GRCh38
rs1043204647 Chr.9: 34620500-34620500 on Build GRCh38
rs543091855 Chr.9: 34620393-34620393 on Build GRCh38
rs866504249 Chr.9: 34620408-34620408 on Build GRCh38
rs780584674 Chr.9: 34620383-34620383 on Build GRCh38
rs776622875 Chr.9: 34620461-34620480 on Build GRCh38
rs866205299 Chr.9: 34620463-34620463 on Build GRCh38
rs774436674 Chr.9: 34620407-34620407 on Build GRCh38
rs749630657 Chr.9: 34620404-34620404 on Build GRCh38
rs780419837 Chr.9: 34620403-34620403 on Build GRCh38
rs773274648 Chr.9: 34620431-34620431 on Build GRCh38
rs372180175 Chr.9: 34620475-34620475 on Build GRCh38
rs1019993776 Chr.9: 34620531-34620531 on Build GRCh38
rs528829993 Chr.9: 34620507-34620507 on Build GRCh38

View Full Product Details