Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 987352-987577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
UniGene
Hs.653258
Gene Chromosome Location
Chr.7: 978485-1138291 on Build GRCh38
Transcripts
XM_011515580.2 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012719.1 XM_017012720.1 XM_017012721.1 NM_017781.2 XM_011515440.2 XM_011515441.2

Gene Symbol
CYP2W1
Entrez Gene ID
54905
Gene Name
cytochrome P450 family 2 subfamily W member 1
Gene Aliases
-
UniGene
Hs.272795
Gene Chromosome Location
Chr.7: 983157-989640 on Build GRCh38
Transcripts
XM_011515580.2 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012719.1 XM_017012720.1 XM_017012721.1 NM_017781.2 XM_011515440.2 XM_011515441.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6639269 c.1048G>A p.V350M Chr.7: 987436-987436 on Build GRCh38
6400112 c.1108G>A p.A370T Chr.7: 987496-987496 on Build GRCh38
6432068 c.1097G>A p.R366H Chr.7: 987485-987485 on Build GRCh38
3431016 c.1126G>A p.G376S Chr.7: 987514-987514 on Build GRCh38
1547498 c.1035C>A p.P345P Chr.7: 987423-987423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535368072 Chr.7: 987537-987537 on Build GRCh38
rs954940852 Chr.7: 987550-987550 on Build GRCh38
rs549964637 Chr.7: 987478-987478 on Build GRCh38
rs750149055 Chr.7: 987474-987474 on Build GRCh38
rs766645260 Chr.7: 987378-987378 on Build GRCh38
rs755761762 Chr.7: 987543-987543 on Build GRCh38
rs767515933 Chr.7: 987472-987472 on Build GRCh38
rs771271278 Chr.7: 987500-987500 on Build GRCh38
rs777045679 Chr.7: 987501-987501 on Build GRCh38
rs748741548 Chr.7: 987408-987408 on Build GRCh38
rs200372185 Chr.7: 987397-987397 on Build GRCh38
rs374069373 Chr.7: 987373-987373 on Build GRCh38
rs370228058 Chr.7: 987438-987438 on Build GRCh38
rs755645891 Chr.7: 987536-987537 on Build GRCh38
rs201612311 Chr.7: 987514-987514 on Build GRCh38
rs935432945 Chr.7: 987481-987481 on Build GRCh38
rs775384793 Chr.7: 987522-987522 on Build GRCh38
rs199680587 Chr.7: 987440-987440 on Build GRCh38
rs564802773 Chr.7: 987375-987375 on Build GRCh38
rs779796122 Chr.7: 987483-987483 on Build GRCh38
rs113803206 Chr.7: 987382-987382 on Build GRCh38
rs755050270 Chr.7: 987389-987389 on Build GRCh38
rs761132341 Chr.7: 987444-987444 on Build GRCh38
rs574975882 Chr.7: 987539-987539 on Build GRCh38
rs778347244 Chr.7: 987499-987499 on Build GRCh38
rs769250658 Chr.7: 987370-987370 on Build GRCh38
rs753528839 Chr.7: 987484-987484 on Build GRCh38
rs149240463 Chr.7: 987371-987371 on Build GRCh38
rs766521461 Chr.7: 987445-987445 on Build GRCh38
rs763125088 Chr.7: 987528-987528 on Build GRCh38
rs772417891 Chr.7: 987494-987494 on Build GRCh38
rs755676275 Chr.7: 987398-987398 on Build GRCh38
rs762212624 Chr.7: 987372-987372 on Build GRCh38
rs765293283 Chr.7: 987455-987455 on Build GRCh38
rs749923076 Chr.7: 987434-987435 on Build GRCh38
rs753874337 Chr.7: 987388-987388 on Build GRCh38
rs747713803 Chr.7: 987493-987493 on Build GRCh38
rs779734967 Chr.7: 987538-987539 on Build GRCh38
rs760784943 Chr.7: 987376-987376 on Build GRCh38
rs547544258 Chr.7: 987433-987433 on Build GRCh38
rs751481845 Chr.7: 987396-987396 on Build GRCh38
rs752143691 Chr.7: 987464-987464 on Build GRCh38
rs768296469 Chr.7: 987432-987432 on Build GRCh38
rs566362502 Chr.7: 987535-987535 on Build GRCh38
rs1031844025 Chr.7: 987510-987510 on Build GRCh38
rs763958659 Chr.7: 987390-987390 on Build GRCh38
rs759809358 Chr.7: 987454-987454 on Build GRCh38
rs530199388 Chr.7: 987465-987465 on Build GRCh38
rs748600320 Chr.7: 987435-987435 on Build GRCh38
rs776847452 Chr.7: 987450-987450 on Build GRCh38
rs555143980 Chr.7: 987538-987538 on Build GRCh38
rs755945940 Chr.7: 987477-987477 on Build GRCh38
rs1048949977 Chr.7: 987436-987436 on Build GRCh38
rs770052941 Chr.7: 987515-987515 on Build GRCh38
rs539008687 Chr.7: 987492-987492 on Build GRCh38
rs377096714 Chr.7: 987423-987423 on Build GRCh38
rs200641657 Chr.7: 987485-987485 on Build GRCh38
rs1035339319 Chr.7: 987508-987508 on Build GRCh38

View Full Product Details