Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6317765-6318266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHG6
Entrez Gene ID
55200
Gene Name
pleckstrin homology and RhoGEF domain containing G6
Gene Aliases
MyoGEF
UniGene
Hs.631660
Gene Chromosome Location
Chr.12: 6310436-6328506 on Build GRCh38
Transcripts
NM_001144856.1 NM_001144857.1 NM_018173.3 XM_005253704.4 XM_006718985.3 XM_011520967.2 XM_017019555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4520131 c.1043G>A p.R348Q Chr.12: 6317882-6317882 on Build GRCh38
4210228 c.1025C>T p.A342V Chr.12: 6317864-6317864 on Build GRCh38
5980236 c.1070G>T p.G357V Chr.12: 6317909-6317909 on Build GRCh38
50708 c.1105C>T p.R369C Chr.12: 6317944-6317944 on Build GRCh38
7317694 c.1141G>A p.D381N Chr.12: 6317980-6317980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563628413 Chr.12: 6318059-6318059 on Build GRCh38
rs71584814 Chr.12: 6317909-6317909 on Build GRCh38
rs766464638 Chr.12: 6317935-6317935 on Build GRCh38
rs767946453 Chr.12: 6317831-6317831 on Build GRCh38
rs148707090 Chr.12: 6317865-6317865 on Build GRCh38
rs890667592 Chr.12: 6318079-6318079 on Build GRCh38
rs559478252 Chr.12: 6318098-6318098 on Build GRCh38
rs975614587 Chr.12: 6317812-6317812 on Build GRCh38
rs370268581 Chr.12: 6317843-6317843 on Build GRCh38
rs747780024 Chr.12: 6317880-6317880 on Build GRCh38
rs534778087 Chr.12: 6317958-6317958 on Build GRCh38
rs936543748 Chr.12: 6317834-6317834 on Build GRCh38
rs747423077 Chr.12: 6318168-6318168 on Build GRCh38
rs374452757 Chr.12: 6317868-6317868 on Build GRCh38
rs539641667 Chr.12: 6317887-6317887 on Build GRCh38
rs868434829 Chr.12: 6317842-6317842 on Build GRCh38
rs762568419 Chr.12: 6318122-6318122 on Build GRCh38
rs183009791 Chr.12: 6318150-6318150 on Build GRCh38
rs572667773 Chr.12: 6317994-6317994 on Build GRCh38
rs186384085 Chr.12: 6317835-6317835 on Build GRCh38
rs957799325 Chr.12: 6318155-6318155 on Build GRCh38
rs899321605 Chr.12: 6317889-6317889 on Build GRCh38
rs949393220 Chr.12: 6317852-6317852 on Build GRCh38
rs760720992 Chr.12: 6317836-6317836 on Build GRCh38
rs892331009 Chr.12: 6318220-6318220 on Build GRCh38
rs532134782 Chr.12: 6318118-6318118 on Build GRCh38
rs778583531 Chr.12: 6317863-6317863 on Build GRCh38
rs74491659 Chr.12: 6318009-6318009 on Build GRCh38
rs111256024 Chr.12: 6318085-6318085 on Build GRCh38
rs760050925 Chr.12: 6317830-6317830 on Build GRCh38
rs780715801 Chr.12: 6317879-6317879 on Build GRCh38
rs912274195 Chr.12: 6317794-6317794 on Build GRCh38
rs145264028 Chr.12: 6317930-6317930 on Build GRCh38
rs982041019 Chr.12: 6318235-6318235 on Build GRCh38
rs1020747990 Chr.12: 6318178-6318178 on Build GRCh38
rs191957730 Chr.12: 6318080-6318080 on Build GRCh38
rs746570884 Chr.12: 6317875-6317875 on Build GRCh38
rs780444275 Chr.12: 6317987-6317988 on Build GRCh38
rs938043025 Chr.12: 6318021-6318021 on Build GRCh38
rs758249038 Chr.12: 6317931-6317931 on Build GRCh38
rs202225852 Chr.12: 6318073-6318073 on Build GRCh38
rs957847973 Chr.12: 6317952-6317952 on Build GRCh38
rs1032031216 Chr.12: 6318143-6318143 on Build GRCh38
rs771234741 Chr.12: 6317816-6317816 on Build GRCh38
rs750668231 Chr.12: 6318038-6318038 on Build GRCh38
rs543830608 Chr.12: 6318017-6318017 on Build GRCh38
rs747358297 Chr.12: 6318033-6318034 on Build GRCh38
rs755790295 Chr.12: 6317956-6317956 on Build GRCh38
rs772615324 Chr.12: 6317890-6317890 on Build GRCh38
rs962111214 Chr.12: 6318082-6318082 on Build GRCh38
rs867050451 Chr.12: 6317901-6317901 on Build GRCh38
rs368449918 Chr.12: 6317788-6317788 on Build GRCh38
rs929436832 Chr.12: 6318053-6318053 on Build GRCh38
rs905177228 Chr.12: 6318245-6318245 on Build GRCh38
rs899269240 Chr.12: 6318046-6318046 on Build GRCh38
rs1026549540 Chr.12: 6317903-6317903 on Build GRCh38
rs975182871 Chr.12: 6317949-6317949 on Build GRCh38
rs1008279462 Chr.12: 6317916-6317916 on Build GRCh38
rs2286715 Chr.12: 6318189-6318189 on Build GRCh38
rs550586055 Chr.12: 6317858-6317858 on Build GRCh38
rs775944078 Chr.12: 6317902-6317902 on Build GRCh38
rs768253554 Chr.12: 6317877-6317877 on Build GRCh38
rs765488380 Chr.12: 6317925-6317925 on Build GRCh38
rs774729777 Chr.12: 6317825-6317825 on Build GRCh38
rs747565138 Chr.12: 6318071-6318071 on Build GRCh38
rs368643831 Chr.12: 6317944-6317944 on Build GRCh38
rs758623341 Chr.12: 6318041-6318041 on Build GRCh38
rs764101649 Chr.12: 6317910-6317910 on Build GRCh38
rs761944236 Chr.12: 6317920-6317920 on Build GRCh38
rs561855927 Chr.12: 6318180-6318180 on Build GRCh38
rs71584815 Chr.12: 6318015-6318015 on Build GRCh38
rs754917335 Chr.12: 6317945-6317945 on Build GRCh38
rs913795047 Chr.12: 6317982-6317982 on Build GRCh38
rs573086260 Chr.12: 6317911-6317911 on Build GRCh38
rs985998182 Chr.12: 6318142-6318142 on Build GRCh38
rs550644322 Chr.12: 6318242-6318242 on Build GRCh38
rs141353120 Chr.12: 6317881-6317881 on Build GRCh38
rs758612963 Chr.12: 6317950-6317950 on Build GRCh38
rs747630236 Chr.12: 6317953-6317953 on Build GRCh38
rs976158997 Chr.12: 6317895-6317895 on Build GRCh38
rs754035942 Chr.12: 6317853-6317853 on Build GRCh38
rs952879241 Chr.12: 6317859-6317859 on Build GRCh38
rs780582316 Chr.12: 6317950-6317950 on Build GRCh38
rs1046498334 Chr.12: 6318236-6318236 on Build GRCh38
rs567928929 Chr.12: 6317808-6317808 on Build GRCh38
rs1025385696 Chr.12: 6318017-6318017 on Build GRCh38
rs772456454 Chr.12: 6317959-6317959 on Build GRCh38
rs1468603 Chr.12: 6317886-6317886 on Build GRCh38

View Full Product Details