Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 47236049-47236540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCED1B
Entrez Gene ID
91523
Gene Name
PC-esterase domain containing 1B
Gene Aliases
FAM113B
UniGene
Hs.560100
Gene Chromosome Location
Chr.12: 47079603-47236663 on Build GRCh38
Transcripts
NM_001281429.1 NM_138371.2 XM_005269224.4 XM_011538978.2 XM_017020207.1 XM_017020208.1 XM_017020209.1 XM_017020210.1 XM_017020211.1 XM_017020212.1 XM_017020213.1 XM_017020214.1 XM_017020215.1 XM_017020216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3461143 c.1249C>T p.R417* Chr.12: 47236312-47236312 on Build GRCh38
4523854 c.1238G>A p.G413E Chr.12: 47236301-47236301 on Build GRCh38
3359739 c.1212T>A p.R404R Chr.12: 47236275-47236275 on Build GRCh38
365651 c.1217G>T p.R406L Chr.12: 47236280-47236280 on Build GRCh38
431149 c.1229C>T p.T410M Chr.12: 47236292-47236292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751665541 Chr.12: 47236247-47236247 on Build GRCh38
rs753671267 Chr.12: 47236253-47236253 on Build GRCh38
rs368596518 Chr.12: 47236349-47236349 on Build GRCh38
rs145379534 Chr.12: 47236345-47236345 on Build GRCh38
rs988945975 Chr.12: 47236101-47236101 on Build GRCh38
rs147692732 Chr.12: 47236412-47236412 on Build GRCh38
rs749663670 Chr.12: 47236383-47236383 on Build GRCh38
rs771683925 Chr.12: 47236269-47236269 on Build GRCh38
rs375990358 Chr.12: 47236371-47236371 on Build GRCh38
rs202147059 Chr.12: 47236260-47236260 on Build GRCh38
rs530311372 Chr.12: 47236331-47236331 on Build GRCh38
rs148451539 Chr.12: 47236182-47236182 on Build GRCh38
rs375607179 Chr.12: 47236235-47236235 on Build GRCh38
rs974541328 Chr.12: 47236500-47236500 on Build GRCh38
rs374835427 Chr.12: 47236325-47236325 on Build GRCh38
rs2692368 Chr.12: 47236347-47236347 on Build GRCh38
rs748154819 Chr.12: 47236085-47236085 on Build GRCh38
rs761223135 Chr.12: 47236117-47236117 on Build GRCh38
rs780041489 Chr.12: 47236170-47236170 on Build GRCh38
rs372663196 Chr.12: 47236179-47236179 on Build GRCh38
rs756174691 Chr.12: 47236249-47236249 on Build GRCh38
rs371203659 Chr.12: 47236353-47236353 on Build GRCh38
rs368685456 Chr.12: 47236124-47236124 on Build GRCh38
rs772155227 Chr.12: 47236379-47236379 on Build GRCh38
rs144729661 Chr.12: 47236481-47236481 on Build GRCh38
rs769657966 Chr.12: 47236099-47236099 on Build GRCh38
rs746084235 Chr.12: 47236315-47236316 on Build GRCh38
rs142383431 Chr.12: 47236413-47236413 on Build GRCh38
rs956254543 Chr.12: 47236452-47236452 on Build GRCh38
rs2543737 Chr.12: 47236348-47236348 on Build GRCh38
rs143995933 Chr.12: 47236143-47236143 on Build GRCh38
rs1033286595 Chr.12: 47236434-47236434 on Build GRCh38
rs201970972 Chr.12: 47236183-47236183 on Build GRCh38
rs768418049 Chr.12: 47236178-47236178 on Build GRCh38
rs752514731 Chr.12: 47236327-47236327 on Build GRCh38
rs745664723 Chr.12: 47236095-47236095 on Build GRCh38
rs745754589 Chr.12: 47236167-47236167 on Build GRCh38
rs759818683 Chr.12: 47236210-47236210 on Build GRCh38
rs764393378 Chr.12: 47236302-47236302 on Build GRCh38
rs754832107 Chr.12: 47236316-47236316 on Build GRCh38
rs567946095 Chr.12: 47236486-47236486 on Build GRCh38
rs752994533 Chr.12: 47236364-47236364 on Build GRCh38
rs770009860 Chr.12: 47236292-47236292 on Build GRCh38
rs768072486 Chr.12: 47236122-47236122 on Build GRCh38
rs781294958 Chr.12: 47236157-47236157 on Build GRCh38
rs754671446 Chr.12: 47236254-47236254 on Build GRCh38
rs746529376 Chr.12: 47236342-47236342 on Build GRCh38
rs192013203 Chr.12: 47236497-47236497 on Build GRCh38
rs144499270 Chr.12: 47236280-47236280 on Build GRCh38
rs765365140 Chr.12: 47236145-47236145 on Build GRCh38
rs907570133 Chr.12: 47236445-47236445 on Build GRCh38
rs749007819 Chr.12: 47236112-47236112 on Build GRCh38
rs775815786 Chr.12: 47236293-47236293 on Build GRCh38
rs764307116 Chr.12: 47236236-47236236 on Build GRCh38
rs749058444 Chr.12: 47236085-47236086 on Build GRCh38
rs573318402 Chr.12: 47236148-47236148 on Build GRCh38
rs771520499 Chr.12: 47236196-47236196 on Build GRCh38
rs752806462 Chr.12: 47236231-47236231 on Build GRCh38
rs773738748 Chr.12: 47236197-47236197 on Build GRCh38
rs201223112 Chr.12: 47236256-47236256 on Build GRCh38
rs766691196 Chr.12: 47236129-47236129 on Build GRCh38
rs763311567 Chr.12: 47236300-47236300 on Build GRCh38
rs959988900 Chr.12: 47236146-47236146 on Build GRCh38
rs770913140 Chr.12: 47236133-47236134 on Build GRCh38
rs145266382 Chr.12: 47236388-47236388 on Build GRCh38
rs774578341 Chr.12: 47236209-47236211 on Build GRCh38
rs200331752 Chr.12: 47236223-47236223 on Build GRCh38
rs139056948 Chr.12: 47236081-47236081 on Build GRCh38
rs886325466 Chr.12: 47236284-47236284 on Build GRCh38
rs753668115 Chr.12: 47236313-47236313 on Build GRCh38
rs1036994877 Chr.12: 47236333-47236333 on Build GRCh38
rs535665336 Chr.12: 47236114-47236114 on Build GRCh38
rs565421898 Chr.12: 47236365-47236365 on Build GRCh38
rs754173521 Chr.12: 47236136-47236136 on Build GRCh38
rs866765655 Chr.12: 47236382-47236382 on Build GRCh38
rs773649009 Chr.12: 47236123-47236123 on Build GRCh38
rs531193288 Chr.12: 47236201-47236201 on Build GRCh38
rs751753564 Chr.12: 47236306-47236306 on Build GRCh38
rs187610959 Chr.12: 47236100-47236100 on Build GRCh38
rs777112473 Chr.12: 47236209-47236209 on Build GRCh38
rs781735977 Chr.12: 47236354-47236354 on Build GRCh38
rs771406055 Chr.12: 47236208-47236208 on Build GRCh38
rs112959993 Chr.12: 47236279-47236279 on Build GRCh38
rs765504758 Chr.12: 47236222-47236222 on Build GRCh38
rs757280610 Chr.12: 47236091-47236091 on Build GRCh38
rs992484764 Chr.12: 47236168-47236168 on Build GRCh38
rs866532176 Chr.12: 47236286-47236286 on Build GRCh38
rs763118814 Chr.12: 47236232-47236232 on Build GRCh38
rs868755947 Chr.12: 47236473-47236473 on Build GRCh38
rs138547957 Chr.12: 47236258-47236258 on Build GRCh38
rs267603480 Chr.12: 47236171-47236171 on Build GRCh38
rs763429044 Chr.12: 47236399-47236399 on Build GRCh38
rs759961201 Chr.12: 47236281-47236281 on Build GRCh38
rs773965399 Chr.12: 47236113-47236113 on Build GRCh38
rs988446943 Chr.12: 47236478-47236478 on Build GRCh38
rs141120293 Chr.12: 47236273-47236273 on Build GRCh38
rs534344598 Chr.12: 47236368-47236368 on Build GRCh38
rs889851965 Chr.12: 47236291-47236291 on Build GRCh38
rs758347631 Chr.12: 47236154-47236154 on Build GRCh38
rs749019266 Chr.12: 47236174-47236174 on Build GRCh38
rs536440912 Chr.12: 47236508-47236508 on Build GRCh38
rs774656946 Chr.12: 47236391-47236391 on Build GRCh38
rs77526916 Chr.12: 47236335-47236335 on Build GRCh38
rs201315028 Chr.12: 47236307-47236307 on Build GRCh38
rs781275096 Chr.12: 47236092-47236092 on Build GRCh38
rs766431910 Chr.12: 47236312-47236312 on Build GRCh38
rs139764765 Chr.12: 47236274-47236274 on Build GRCh38
rs758107227 Chr.12: 47236330-47236330 on Build GRCh38

View Full Product Details