Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 63802288-63802800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM5
Entrez Gene ID
10329
Gene Name
transmembrane protein 5
Gene Aliases
HP10481, MDDGA10
UniGene
Hs.216386
Gene Chromosome Location
Chr.12: 63779803-63809558 on Build GRCh38
Transcripts
NM_001278237.1 NM_014254.2 XM_005268562.3 XM_005268563.3 XM_017018686.1 XM_017018687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3667194 c.660C>T p.G220G Chr.12: 63802322-63802322 on Build GRCh38
4044100 c.696C>T p.P232P Chr.12: 63802358-63802358 on Build GRCh38
6998284 c.743+1G>A p.? Chr.12: 63802406-63802406 on Build GRCh38
1990851 c.709G>A p.V237M Chr.12: 63802371-63802371 on Build GRCh38
4450637 c.743+1G>C p.? Chr.12: 63802406-63802406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990058569 Chr.12: 63802722-63802722 on Build GRCh38
rs113670193 Chr.12: 63802642-63802642 on Build GRCh38
rs940158205 Chr.12: 63802482-63802482 on Build GRCh38
rs746657899 Chr.12: 63802414-63802414 on Build GRCh38
rs527706413 Chr.12: 63802622-63802622 on Build GRCh38
rs370288545 Chr.12: 63802326-63802326 on Build GRCh38
rs1011791980 Chr.12: 63802614-63802614 on Build GRCh38
rs776498772 Chr.12: 63802405-63802406 on Build GRCh38
rs781405614 Chr.12: 63802366-63802366 on Build GRCh38
rs1047541304 Chr.12: 63802421-63802421 on Build GRCh38
rs778174763 Chr.12: 63802406-63802406 on Build GRCh38
rs755292481 Chr.12: 63802356-63802356 on Build GRCh38
rs756576089 Chr.12: 63802371-63802371 on Build GRCh38
rs761799102 Chr.12: 63802317-63802317 on Build GRCh38
rs768357640 Chr.12: 63802415-63802415 on Build GRCh38
rs779286890 Chr.12: 63802411-63802411 on Build GRCh38
rs1005035187 Chr.12: 63802621-63802621 on Build GRCh38
rs902584864 Chr.12: 63802469-63802469 on Build GRCh38
rs562914250 Chr.12: 63802666-63802666 on Build GRCh38
rs762872834 Chr.12: 63802335-63802335 on Build GRCh38
rs953424909 Chr.12: 63802696-63802696 on Build GRCh38
rs891157368 Chr.12: 63802555-63802555 on Build GRCh38
rs769543798 Chr.12: 63802410-63802411 on Build GRCh38
rs536580809 Chr.12: 63802659-63802659 on Build GRCh38
rs543832468 Chr.12: 63802654-63802654 on Build GRCh38
rs898707569 Chr.12: 63802536-63802536 on Build GRCh38
rs1028618142 Chr.12: 63802690-63802690 on Build GRCh38
rs766660116 Chr.12: 63802343-63802343 on Build GRCh38
rs928272073 Chr.12: 63802709-63802712 on Build GRCh38
rs761237366 Chr.12: 63802453-63802453 on Build GRCh38
rs116323777 Chr.12: 63802613-63802613 on Build GRCh38
rs887252847 Chr.12: 63802439-63802439 on Build GRCh38
rs1056446098 Chr.12: 63802544-63802544 on Build GRCh38
rs907595130 Chr.12: 63802750-63802751 on Build GRCh38
rs887986204 Chr.12: 63802620-63802620 on Build GRCh38
rs941796379 Chr.12: 63802771-63802771 on Build GRCh38
rs567958431 Chr.12: 63802652-63802652 on Build GRCh38
rs749612243 Chr.12: 63802407-63802407 on Build GRCh38
rs550391556 Chr.12: 63802739-63802739 on Build GRCh38
rs1028736469 Chr.12: 63802553-63802553 on Build GRCh38
rs989944091 Chr.12: 63802325-63802325 on Build GRCh38
rs868255015 Chr.12: 63802347-63802347 on Build GRCh38
rs751810776 Chr.12: 63802344-63802344 on Build GRCh38
rs143312527 Chr.12: 63802370-63802370 on Build GRCh38
rs761767587 Chr.12: 63802406-63802407 on Build GRCh38
rs914473311 Chr.12: 63802770-63802770 on Build GRCh38
rs748119742 Chr.12: 63802416-63802416 on Build GRCh38
rs377078737 Chr.12: 63802556-63802556 on Build GRCh38
rs113706160 Chr.12: 63802740-63802740 on Build GRCh38
rs760201973 Chr.12: 63802763-63802763 on Build GRCh38
rs959715399 Chr.12: 63802719-63802719 on Build GRCh38
rs776969523 Chr.12: 63802400-63802400 on Build GRCh38
rs776152671 Chr.12: 63802429-63802429 on Build GRCh38
rs1012472681 Chr.12: 63802579-63802579 on Build GRCh38
rs372706919 Chr.12: 63802504-63802504 on Build GRCh38
rs1031693239 Chr.12: 63802600-63802600 on Build GRCh38

View Full Product Details