Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 70535918-70536429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRB
Entrez Gene ID
5787
Gene Name
protein tyrosine phosphatase, receptor type B
Gene Aliases
HPTP-BETA, HPTPB, PTPB, R-PTP-BETA, VEPTP
UniGene
Hs.434375
Gene Chromosome Location
Chr.12: 70516850-70637440 on Build GRCh38
Transcripts
NM_001109754.2 NM_001206971.1 NM_001206972.1 NM_002837.4 XM_006719528.1 XM_006719529.3 XM_011538614.1 XM_017019723.1 XM_017019724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6574462 c.5987C>G p.P1996R Chr.12: 70536119-70536119 on Build GRCh38
7420458 c.5294G>A p.G1765D Chr.12: 70536158-70536158 on Build GRCh38
263176 c.6039T>G p.V2013V Chr.12: 70536067-70536067 on Build GRCh38
6574464 c.5333C>G p.P1778R Chr.12: 70536119-70536119 on Build GRCh38
1364008 c.5427+1G>C p.? Chr.12: 70536024-70536024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780239596 Chr.12: 70536057-70536057 on Build GRCh38
rs896337477 Chr.12: 70536165-70536165 on Build GRCh38
rs951495312 Chr.12: 70536227-70536227 on Build GRCh38
rs374328210 Chr.12: 70535989-70535989 on Build GRCh38
rs760259977 Chr.12: 70536008-70536008 on Build GRCh38
rs921415811 Chr.12: 70535967-70535967 on Build GRCh38
rs1017929257 Chr.12: 70536088-70536088 on Build GRCh38
rs899595228 Chr.12: 70536143-70536143 on Build GRCh38
rs748711640 Chr.12: 70536100-70536100 on Build GRCh38
rs187096273 Chr.12: 70536223-70536223 on Build GRCh38
rs775967586 Chr.12: 70536168-70536168 on Build GRCh38
rs761611289 Chr.12: 70536027-70536027 on Build GRCh38
rs771918914 Chr.12: 70536135-70536135 on Build GRCh38
rs372650901 Chr.12: 70536392-70536392 on Build GRCh38
rs750272288 Chr.12: 70536183-70536183 on Build GRCh38
rs761819090 Chr.12: 70536177-70536177 on Build GRCh38
rs529751141 Chr.12: 70536061-70536061 on Build GRCh38
rs547277356 Chr.12: 70536399-70536400 on Build GRCh38
rs570710312 Chr.12: 70536298-70536298 on Build GRCh38
rs762729918 Chr.12: 70536191-70536191 on Build GRCh38
rs1043390922 Chr.12: 70536070-70536070 on Build GRCh38
rs945073249 Chr.12: 70536062-70536062 on Build GRCh38
rs764170403 Chr.12: 70536018-70536018 on Build GRCh38
rs975673315 Chr.12: 70536337-70536337 on Build GRCh38
rs370392872 Chr.12: 70536179-70536179 on Build GRCh38
rs758931118 Chr.12: 70536181-70536182 on Build GRCh38
rs1013339402 Chr.12: 70536206-70536206 on Build GRCh38
rs1041659129 Chr.12: 70536343-70536343 on Build GRCh38
rs751756281 Chr.12: 70536202-70536202 on Build GRCh38
rs781597034 Chr.12: 70536089-70536089 on Build GRCh38
rs921480982 Chr.12: 70536378-70536378 on Build GRCh38
rs912486778 Chr.12: 70536359-70536359 on Build GRCh38
rs962987602 Chr.12: 70535941-70535941 on Build GRCh38
rs2278346 Chr.12: 70536281-70536281 on Build GRCh38
rs778237914 Chr.12: 70536282-70536282 on Build GRCh38
rs759268200 Chr.12: 70535979-70535979 on Build GRCh38
rs770159838 Chr.12: 70536117-70536117 on Build GRCh38
rs750114491 Chr.12: 70536044-70536044 on Build GRCh38
rs562645349 Chr.12: 70535950-70535950 on Build GRCh38
rs1004658471 Chr.12: 70536263-70536263 on Build GRCh38
rs533572654 Chr.12: 70536337-70536338 on Build GRCh38
rs755019217 Chr.12: 70536069-70536069 on Build GRCh38
rs746451547 Chr.12: 70536153-70536153 on Build GRCh38
rs1027825484 Chr.12: 70536148-70536148 on Build GRCh38
rs199503832 Chr.12: 70536152-70536152 on Build GRCh38
rs201163672 Chr.12: 70536026-70536026 on Build GRCh38
rs976017097 Chr.12: 70535964-70535964 on Build GRCh38
rs1042018388 Chr.12: 70535987-70535987 on Build GRCh38
rs995220166 Chr.12: 70536146-70536146 on Build GRCh38
rs890825053 Chr.12: 70536058-70536058 on Build GRCh38
rs778214231 Chr.12: 70536118-70536118 on Build GRCh38
rs749657518 Chr.12: 70536129-70536129 on Build GRCh38
rs927263781 Chr.12: 70536282-70536285 on Build GRCh38
rs779714075 Chr.12: 70536004-70536004 on Build GRCh38
rs771793754 Chr.12: 70535981-70535981 on Build GRCh38
rs766625654 Chr.12: 70536195-70536195 on Build GRCh38
rs934117443 Chr.12: 70535973-70535973 on Build GRCh38
rs765116092 Chr.12: 70536034-70536034 on Build GRCh38
rs758628215 Chr.12: 70536052-70536052 on Build GRCh38
rs749826663 Chr.12: 70536323-70536323 on Build GRCh38
rs566169232 Chr.12: 70536260-70536260 on Build GRCh38
rs946024457 Chr.12: 70535958-70535958 on Build GRCh38
rs144389405 Chr.12: 70536158-70536158 on Build GRCh38

View Full Product Details