Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108784977-108785486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSH1
Entrez Gene ID
54434
Gene Name
slingshot protein phosphatase 1
Gene Aliases
SSH1L
UniGene
Hs.199763
Gene Chromosome Location
Chr.12: 108782690-108857606 on Build GRCh38
Transcripts
NM_018984.3 XM_005268984.4 XM_005268985.2 XM_011538497.1 XM_011538499.1 XM_011538500.2 XM_011538501.2 XM_017019491.1 XM_017019492.1 XM_017019493.1 XM_017019494.1

SNP Detail

SNP ID Chromosome Location
rs193251433 Chr.12: 108785373-108785373 on Build GRCh38
rs896047520 Chr.12: 108785371-108785371 on Build GRCh38
rs116555862 Chr.12: 108785082-108785082 on Build GRCh38
rs916052280 Chr.12: 108785440-108785440 on Build GRCh38
rs955465389 Chr.12: 108785166-108785166 on Build GRCh38
rs759342367 Chr.12: 108785130-108785130 on Build GRCh38
rs1014541343 Chr.12: 108785063-108785063 on Build GRCh38
rs957179930 Chr.12: 108785147-108785147 on Build GRCh38
rs1024388521 Chr.12: 108785077-108785077 on Build GRCh38
rs991595896 Chr.12: 108785421-108785421 on Build GRCh38
rs766326130 Chr.12: 108785056-108785056 on Build GRCh38
rs548966434 Chr.12: 108785294-108785294 on Build GRCh38
rs894682360 Chr.12: 108785264-108785264 on Build GRCh38
rs772436741 Chr.12: 108785418-108785418 on Build GRCh38
rs1057398760 Chr.12: 108785323-108785323 on Build GRCh38
rs950260376 Chr.12: 108785451-108785451 on Build GRCh38
rs202098833 Chr.12: 108785431-108785432 on Build GRCh38
rs1036476353 Chr.12: 108785004-108785004 on Build GRCh38
rs950502437 Chr.12: 108785158-108785158 on Build GRCh38
rs760416670 Chr.12: 108785365-108785365 on Build GRCh38
rs964439582 Chr.12: 108785190-108785190 on Build GRCh38
rs934810651 Chr.12: 108785277-108785277 on Build GRCh38
rs977779669 Chr.12: 108785253-108785253 on Build GRCh38
rs930765266 Chr.12: 108785243-108785243 on Build GRCh38
rs930816140 Chr.12: 108785029-108785029 on Build GRCh38
rs1044793689 Chr.12: 108785379-108785379 on Build GRCh38
rs899130542 Chr.12: 108785385-108785385 on Build GRCh38
rs940713224 Chr.12: 108785184-108785184 on Build GRCh38
rs764986994 Chr.12: 108785139-108785139 on Build GRCh38
rs527307864 Chr.12: 108785270-108785270 on Build GRCh38
rs892897767 Chr.12: 108785319-108785319 on Build GRCh38
rs2003242 Chr.12: 108785197-108785197 on Build GRCh38
rs576624023 Chr.12: 108785049-108785049 on Build GRCh38
rs550046146 Chr.12: 108785397-108785397 on Build GRCh38
rs1046010733 Chr.12: 108785284-108785284 on Build GRCh38
rs367683190 Chr.12: 108785255-108785255 on Build GRCh38
rs920679714 Chr.12: 108785241-108785241 on Build GRCh38
rs374984812 Chr.12: 108785330-108785330 on Build GRCh38
rs150446267 Chr.12: 108785173-108785173 on Build GRCh38
rs758074444 Chr.12: 108785249-108785249 on Build GRCh38
rs756786656 Chr.12: 108785310-108785310 on Build GRCh38
rs544814127 Chr.12: 108785181-108785181 on Build GRCh38
rs1028092644 Chr.12: 108785364-108785364 on Build GRCh38
rs779452359 Chr.12: 108785402-108785402 on Build GRCh38
rs759671771 Chr.12: 108785448-108785448 on Build GRCh38
rs1018899120 Chr.12: 108785414-108785414 on Build GRCh38
rs986756709 Chr.12: 108785171-108785171 on Build GRCh38
rs1023082176 Chr.12: 108785135-108785135 on Build GRCh38
rs538810679 Chr.12: 108785435-108785435 on Build GRCh38
rs1026160860 Chr.12: 108785150-108785150 on Build GRCh38
rs185188232 Chr.12: 108785374-108785374 on Build GRCh38
rs996588494 Chr.12: 108785362-108785362 on Build GRCh38
rs1032685369 Chr.12: 108785316-108785316 on Build GRCh38
rs565286125 Chr.12: 108785113-108785113 on Build GRCh38
rs975220837 Chr.12: 108785155-108785155 on Build GRCh38
rs879090980 Chr.12: 108785123-108785123 on Build GRCh38
rs542588466 Chr.12: 108785017-108785017 on Build GRCh38
rs920628986 Chr.12: 108785413-108785413 on Build GRCh38
rs866646577 Chr.12: 108785081-108785081 on Build GRCh38
rs780582502 Chr.12: 108785332-108785332 on Build GRCh38
rs3741779 Chr.12: 108785032-108785032 on Build GRCh38
rs572142409 Chr.12: 108785016-108785016 on Build GRCh38
rs773430654 Chr.12: 108785021-108785021 on Build GRCh38
rs756802895 Chr.12: 108785331-108785332 on Build GRCh38
rs149473770 Chr.12: 108785411-108785411 on Build GRCh38
rs1028164762 Chr.12: 108785156-108785156 on Build GRCh38
rs994882492 Chr.12: 108785143-108785143 on Build GRCh38
rs1026378371 Chr.12: 108785403-108785403 on Build GRCh38
rs570956128 Chr.12: 108785311-108785311 on Build GRCh38
rs771896800 Chr.12: 108785267-108785267 on Build GRCh38
rs895839117 Chr.12: 108785122-108785123 on Build GRCh38

View Full Product Details