Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 117155587-117156080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO21
Entrez Gene ID
23014
Gene Name
F-box protein 21
Gene Aliases
FBX21
UniGene
Hs.740376
Gene Chromosome Location
Chr.12: 117143473-117190500 on Build GRCh38
Transcripts
NM_015002.2 NM_033624.2 XM_017019037.1 XM_017019038.1 XM_017019039.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6585072 c.1584G>A p.M528I Chr.12: 117155903-117155903 on Build GRCh38
3456650 c.1696G>C p.E566Q Chr.12: 117155791-117155791 on Build GRCh38
393084 c.1559T>C p.I520T Chr.12: 117155928-117155928 on Build GRCh38
1359102 c.1672G>A p.G558S Chr.12: 117155815-117155815 on Build GRCh38
4039116 c.1633G>A p.G545S Chr.12: 117155854-117155854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767028740 Chr.12: 117155651-117155651 on Build GRCh38
rs116529616 Chr.12: 117155741-117155741 on Build GRCh38
rs757101669 Chr.12: 117155855-117155855 on Build GRCh38
rs768212114 Chr.12: 117155784-117155784 on Build GRCh38
rs963494000 Chr.12: 117155628-117155628 on Build GRCh38
rs181286097 Chr.12: 117155984-117155984 on Build GRCh38
rs761051803 Chr.12: 117155907-117155918 on Build GRCh38
rs113669572 Chr.12: 117155774-117155774 on Build GRCh38
rs775990591 Chr.12: 117155784-117155785 on Build GRCh38
rs374267913 Chr.12: 117155959-117155959 on Build GRCh38
rs536937735 Chr.12: 117155813-117155813 on Build GRCh38
rs373307555 Chr.12: 117155962-117155962 on Build GRCh38
rs60423812 Chr.12: 117155858-117155858 on Build GRCh38
rs902568931 Chr.12: 117155773-117155773 on Build GRCh38
rs955047916 Chr.12: 117155678-117155678 on Build GRCh38
rs369432098 Chr.12: 117155934-117155934 on Build GRCh38
rs750162657 Chr.12: 117155867-117155867 on Build GRCh38
rs749105381 Chr.12: 117155756-117155756 on Build GRCh38
rs551167910 Chr.12: 117155704-117155704 on Build GRCh38
rs867748350 Chr.12: 117155823-117155823 on Build GRCh38
rs747792995 Chr.12: 117155958-117155958 on Build GRCh38
rs758169985 Chr.12: 117155996-117155996 on Build GRCh38
rs981151149 Chr.12: 117155645-117155645 on Build GRCh38
rs746467390 Chr.12: 117155781-117155781 on Build GRCh38
rs763358184 Chr.12: 117155630-117155630 on Build GRCh38
rs756296567 Chr.12: 117155754-117155754 on Build GRCh38
rs147009451 Chr.12: 117155924-117155924 on Build GRCh38
rs764143383 Chr.12: 117155979-117155979 on Build GRCh38
rs933491286 Chr.12: 117155987-117155987 on Build GRCh38
rs111980415 Chr.12: 117155977-117155977 on Build GRCh38
rs765381092 Chr.12: 117155986-117155986 on Build GRCh38
rs1017826740 Chr.12: 117155724-117155724 on Build GRCh38
rs774756911 Chr.12: 117155756-117155756 on Build GRCh38
rs373003107 Chr.12: 117155992-117155992 on Build GRCh38
rs759956434 Chr.12: 117155804-117155804 on Build GRCh38
rs950494928 Chr.12: 117155967-117155967 on Build GRCh38
rs1016254088 Chr.12: 117155616-117155616 on Build GRCh38
rs928972893 Chr.12: 117155680-117155680 on Build GRCh38
rs988296839 Chr.12: 117155758-117155758 on Build GRCh38
rs370369543 Chr.12: 117155991-117155991 on Build GRCh38
rs776386359 Chr.12: 117155785-117155785 on Build GRCh38
rs753031646 Chr.12: 117155751-117155751 on Build GRCh38
rs766832709 Chr.12: 117155801-117155801 on Build GRCh38
rs758277895 Chr.12: 117155914-117155914 on Build GRCh38
rs764339365 Chr.12: 117155830-117155830 on Build GRCh38
rs191261291 Chr.12: 117155976-117155976 on Build GRCh38
rs756171229 Chr.12: 117155816-117155816 on Build GRCh38
rs993980573 Chr.12: 117155629-117155629 on Build GRCh38
rs377322357 Chr.12: 117155775-117155775 on Build GRCh38
rs760063511 Chr.12: 117155779-117155779 on Build GRCh38
rs757013768 Chr.12: 117155757-117155757 on Build GRCh38
rs768001042 Chr.12: 117155811-117155811 on Build GRCh38
rs537612840 Chr.12: 117155691-117155691 on Build GRCh38
rs754124854 Chr.12: 117155980-117155980 on Build GRCh38
rs767827025 Chr.12: 117155780-117155781 on Build GRCh38
rs779990762 Chr.12: 117155916-117155916 on Build GRCh38
rs779899510 Chr.12: 117155999-117155999 on Build GRCh38
rs942219573 Chr.12: 117155738-117155738 on Build GRCh38
rs755137452 Chr.12: 117155750-117155750 on Build GRCh38
rs754034961 Chr.12: 117155854-117155854 on Build GRCh38
rs1026364717 Chr.12: 117155674-117155674 on Build GRCh38
rs61999294 Chr.12: 117155942-117155942 on Build GRCh38
rs888053913 Chr.12: 117155738-117155739 on Build GRCh38
rs549033309 Chr.12: 117155609-117155609 on Build GRCh38
rs553351940 Chr.12: 117155769-117155769 on Build GRCh38
rs763453150 Chr.12: 117155800-117155800 on Build GRCh38
rs377288420 Chr.12: 117155782-117155782 on Build GRCh38
rs964920096 Chr.12: 117155740-117155740 on Build GRCh38
rs200476709 Chr.12: 117155755-117155755 on Build GRCh38
rs772480753 Chr.12: 117155973-117155973 on Build GRCh38
rs186935483 Chr.12: 117155617-117155617 on Build GRCh38
rs1052342293 Chr.12: 117156011-117156011 on Build GRCh38

View Full Product Details