Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 33835947-33836464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFC3
Entrez Gene ID
5983
Gene Name
replication factor C subunit 3
Gene Aliases
RFC38
UniGene
Hs.115474
Gene Chromosome Location
Chr.13: 33818049-33973946 on Build GRCh38
Transcripts
NM_002915.3 NM_181558.2 XM_011535172.2 XM_011535173.2 XM_011535174.2 XM_011535175.1 XM_017020680.1 XM_017020681.1 XM_017020682.1 XM_017020683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6347415 c.969G>T p.Q323H Chr.13: 33836193-33836193 on Build GRCh38
4047167 c.1003T>C p.F335L Chr.13: 33836227-33836227 on Build GRCh38
4514087 c.961C>T p.R321C Chr.13: 33836185-33836185 on Build GRCh38
4450818 c.880-1G>A p.? Chr.13: 33836103-33836103 on Build GRCh38
4752578 c.934G>A p.A312T Chr.13: 33836158-33836158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755713921 Chr.13: 33836179-33836179 on Build GRCh38
rs564620451 Chr.13: 33836281-33836281 on Build GRCh38
rs768034481 Chr.13: 33836251-33836251 on Build GRCh38
rs759768187 Chr.13: 33836140-33836140 on Build GRCh38
rs768438949 Chr.13: 33836096-33836096 on Build GRCh38
rs763342958 Chr.13: 33836149-33836149 on Build GRCh38
rs771123471 Chr.13: 33836110-33836110 on Build GRCh38
rs541836078 Chr.13: 33836174-33836174 on Build GRCh38
rs995548019 Chr.13: 33836175-33836175 on Build GRCh38
rs375369387 Chr.13: 33836176-33836176 on Build GRCh38
rs752091239 Chr.13: 33836169-33836169 on Build GRCh38
rs778878736 Chr.13: 33836102-33836102 on Build GRCh38
rs749527002 Chr.13: 33836225-33836225 on Build GRCh38
rs780736330 Chr.13: 33836061-33836061 on Build GRCh38
rs9570340 Chr.13: 33836360-33836360 on Build GRCh38
rs755766837 Chr.13: 33836073-33836073 on Build GRCh38
rs780062296 Chr.13: 33836415-33836415 on Build GRCh38
rs775951678 Chr.13: 33836142-33836142 on Build GRCh38
rs148210921 Chr.13: 33836233-33836233 on Build GRCh38
rs878903532 Chr.13: 33836404-33836404 on Build GRCh38
rs770103853 Chr.13: 33836216-33836216 on Build GRCh38
rs752450764 Chr.13: 33836302-33836302 on Build GRCh38
rs912750333 Chr.13: 33836196-33836196 on Build GRCh38
rs757707789 Chr.13: 33836325-33836325 on Build GRCh38
rs373451224 Chr.13: 33836259-33836259 on Build GRCh38
rs772668318 Chr.13: 33836252-33836252 on Build GRCh38
rs766692309 Chr.13: 33836328-33836329 on Build GRCh38
rs377594733 Chr.13: 33836060-33836060 on Build GRCh38
rs921441893 Chr.13: 33836325-33836330 on Build GRCh38
rs979704966 Chr.13: 33836296-33836296 on Build GRCh38
rs199840307 Chr.13: 33836023-33836023 on Build GRCh38
rs370732006 Chr.13: 33836101-33836101 on Build GRCh38
rs1048693237 Chr.13: 33836362-33836362 on Build GRCh38
rs546322524 Chr.13: 33836242-33836242 on Build GRCh38
rs914857903 Chr.13: 33836241-33836241 on Build GRCh38
rs143372384 Chr.13: 33836185-33836185 on Build GRCh38
rs3135633 Chr.13: 33836190-33836190 on Build GRCh38
rs554603730 Chr.13: 33836095-33836095 on Build GRCh38
rs775756105 Chr.13: 33836218-33836218 on Build GRCh38
rs3135635 Chr.13: 33836394-33836394 on Build GRCh38
rs904985508 Chr.13: 33835974-33835974 on Build GRCh38
rs926335009 Chr.13: 33836371-33836371 on Build GRCh38
rs41546912 Chr.13: 33836000-33836000 on Build GRCh38
rs781366321 Chr.13: 33836201-33836201 on Build GRCh38
rs779651146 Chr.13: 33836330-33836330 on Build GRCh38
rs779766997 Chr.13: 33836080-33836080 on Build GRCh38
rs745971681 Chr.13: 33836209-33836209 on Build GRCh38
rs765923481 Chr.13: 33836268-33836268 on Build GRCh38
rs749946842 Chr.13: 33836177-33836177 on Build GRCh38
rs763056755 Chr.13: 33836254-33836256 on Build GRCh38
rs753474549 Chr.13: 33836184-33836184 on Build GRCh38
rs765018348 Chr.13: 33836284-33836284 on Build GRCh38
rs749129753 Chr.13: 33836086-33836086 on Build GRCh38
rs1037099008 Chr.13: 33836423-33836423 on Build GRCh38
rs750068057 Chr.13: 33836072-33836072 on Build GRCh38
rs748087235 Chr.13: 33836105-33836105 on Build GRCh38
rs754655089 Chr.13: 33836282-33836282 on Build GRCh38
rs746988113 Chr.13: 33836315-33836315 on Build GRCh38
rs749457194 Chr.13: 33836342-33836342 on Build GRCh38
rs767019612 Chr.13: 33836338-33836338 on Build GRCh38
rs777892589 Chr.13: 33836309-33836309 on Build GRCh38
rs749229788 Chr.13: 33836428-33836428 on Build GRCh38
rs556832657 Chr.13: 33836412-33836412 on Build GRCh38
rs3135632 Chr.13: 33836049-33836049 on Build GRCh38
rs181789277 Chr.13: 33836403-33836403 on Build GRCh38
rs779711845 Chr.13: 33836183-33836183 on Build GRCh38
rs372638132 Chr.13: 33836323-33836323 on Build GRCh38
rs764410746 Chr.13: 33836151-33836151 on Build GRCh38
rs752043626 Chr.13: 33836057-33836057 on Build GRCh38
rs758365672 Chr.13: 33836304-33836304 on Build GRCh38
rs954115115 Chr.13: 33836186-33836186 on Build GRCh38
rs1016105222 Chr.13: 33836146-33836146 on Build GRCh38
rs190504365 Chr.13: 33836021-33836021 on Build GRCh38
rs770158720 Chr.13: 33836141-33836141 on Build GRCh38
rs776921741 Chr.13: 33836112-33836112 on Build GRCh38
rs3135634 Chr.13: 33836226-33836226 on Build GRCh38
rs557666108 Chr.13: 33835991-33835991 on Build GRCh38
rs1053960 Chr.13: 33836402-33836402 on Build GRCh38

View Full Product Details