Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 67645561-67646058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARG2
Entrez Gene ID
384
Gene Name
arginase 2
Gene Aliases
-
UniGene
Hs.226007
Gene Chromosome Location
Chr.14: 67619862-67651720 on Build GRCh38
Transcripts
NM_001172.3 XM_017020925.1

Gene Symbol
GPHN
Entrez Gene ID
10243
Gene Name
gephyrin
Gene Aliases
GEPH, GPH, GPHRYN, HKPX1, MOCODC
UniGene
Hs.208765
Gene Chromosome Location
Chr.14: 66507407-67735831 on Build GRCh38
Transcripts
NM_001172.3 XM_017020925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1607872 c.444C>T p.I148I Chr.14: 67645724-67645724 on Build GRCh38
957203 c.432C>A p.A144A Chr.14: 67645712-67645712 on Build GRCh38
7105823 c.477C>T p.L159L Chr.14: 67645757-67645757 on Build GRCh38
6484237 c.364C>A p.L122M Chr.14: 67645644-67645644 on Build GRCh38
4051956 c.431C>T p.A144V Chr.14: 67645711-67645711 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1054627290 Chr.14: 67645657-67645657 on Build GRCh38
rs762909277 Chr.14: 67645771-67645771 on Build GRCh38
rs977519106 Chr.14: 67645941-67645941 on Build GRCh38
rs892906641 Chr.14: 67645610-67645610 on Build GRCh38
rs1008622917 Chr.14: 67645726-67645727 on Build GRCh38
rs750814917 Chr.14: 67645898-67645898 on Build GRCh38
rs11622821 Chr.14: 67645717-67645717 on Build GRCh38
rs867034454 Chr.14: 67645782-67645782 on Build GRCh38
rs781465749 Chr.14: 67645689-67645689 on Build GRCh38
rs761101186 Chr.14: 67646012-67646012 on Build GRCh38
rs374202986 Chr.14: 67645600-67645600 on Build GRCh38
rs144227690 Chr.14: 67645653-67645653 on Build GRCh38
rs766219021 Chr.14: 67645629-67645629 on Build GRCh38
rs201462969 Chr.14: 67645818-67645818 on Build GRCh38
rs368346070 Chr.14: 67645622-67645622 on Build GRCh38
rs1012702121 Chr.14: 67645615-67645615 on Build GRCh38
rs781590447 Chr.14: 67645854-67645854 on Build GRCh38
rs762927490 Chr.14: 67645598-67645598 on Build GRCh38
rs768910144 Chr.14: 67645603-67645603 on Build GRCh38
rs759505590 Chr.14: 67645811-67645811 on Build GRCh38
rs761978033 Chr.14: 67645616-67645616 on Build GRCh38
rs151039935 Chr.14: 67645675-67645675 on Build GRCh38
rs759014720 Chr.14: 67645672-67645672 on Build GRCh38
rs752095886 Chr.14: 67645842-67645842 on Build GRCh38
rs971662482 Chr.14: 67645910-67645910 on Build GRCh38
rs932874475 Chr.14: 67646006-67646008 on Build GRCh38
rs930792175 Chr.14: 67645800-67645800 on Build GRCh38
rs773405238 Chr.14: 67645767-67645767 on Build GRCh38
rs532382482 Chr.14: 67645862-67645862 on Build GRCh38
rs990070074 Chr.14: 67645962-67645962 on Build GRCh38
rs770949105 Chr.14: 67645799-67645799 on Build GRCh38
rs1051302958 Chr.14: 67646015-67646017 on Build GRCh38
rs891299105 Chr.14: 67645706-67645706 on Build GRCh38
rs767425450 Chr.14: 67645814-67645814 on Build GRCh38
rs371739738 Chr.14: 67645691-67645691 on Build GRCh38
rs370914377 Chr.14: 67645639-67645639 on Build GRCh38
rs753432021 Chr.14: 67645821-67645821 on Build GRCh38
rs758634122 Chr.14: 67645840-67645841 on Build GRCh38
rs934568601 Chr.14: 67645911-67645911 on Build GRCh38
rs747425817 Chr.14: 67645680-67645680 on Build GRCh38
rs141081622 Chr.14: 67645720-67645720 on Build GRCh38
rs746012448 Chr.14: 67645839-67645840 on Build GRCh38
rs751440215 Chr.14: 67645634-67645634 on Build GRCh38
rs1015118039 Chr.14: 67645791-67645791 on Build GRCh38
rs752482896 Chr.14: 67645816-67645816 on Build GRCh38
rs377297634 Chr.14: 67646009-67646009 on Build GRCh38
rs146079018 Chr.14: 67645896-67645896 on Build GRCh38
rs370547651 Chr.14: 67645592-67645592 on Build GRCh38
rs755416046 Chr.14: 67645846-67645846 on Build GRCh38
rs560870461 Chr.14: 67645637-67645637 on Build GRCh38
rs892999040 Chr.14: 67646004-67646004 on Build GRCh38
rs530366275 Chr.14: 67645964-67645964 on Build GRCh38
rs769892843 Chr.14: 67645693-67645693 on Build GRCh38
rs1051170396 Chr.14: 67645700-67645700 on Build GRCh38
rs779368145 Chr.14: 67645821-67645824 on Build GRCh38
rs763651099 Chr.14: 67645819-67645819 on Build GRCh38
rs767370288 Chr.14: 67645644-67645644 on Build GRCh38
rs569364869 Chr.14: 67645920-67645920 on Build GRCh38
rs772401599 Chr.14: 67645594-67645594 on Build GRCh38
rs769919060 Chr.14: 67645618-67645618 on Build GRCh38
rs1054363030 Chr.14: 67645994-67645994 on Build GRCh38
rs764682396 Chr.14: 67645840-67645840 on Build GRCh38
rs774452622 Chr.14: 67645808-67645808 on Build GRCh38
rs570183577 Chr.14: 67645591-67645591 on Build GRCh38
rs763000966 Chr.14: 67645624-67645624 on Build GRCh38
rs775616638 Chr.14: 67645792-67645795 on Build GRCh38
rs936013638 Chr.14: 67645961-67645961 on Build GRCh38
rs142854063 Chr.14: 67645590-67645590 on Build GRCh38
rs776814084 Chr.14: 67645613-67645613 on Build GRCh38
rs1022755748 Chr.14: 67646020-67646020 on Build GRCh38
rs542408224 Chr.14: 67645599-67645599 on Build GRCh38
rs912711214 Chr.14: 67646016-67646016 on Build GRCh38
rs372626945 Chr.14: 67645966-67645966 on Build GRCh38
rs924723286 Chr.14: 67645943-67645943 on Build GRCh38
rs754211155 Chr.14: 67645727-67645732 on Build GRCh38
rs755657562 Chr.14: 67645647-67645647 on Build GRCh38
rs368839100 Chr.14: 67645688-67645688 on Build GRCh38
rs757517296 Chr.14: 67645779-67645781 on Build GRCh38
rs756702536 Chr.14: 67645829-67645829 on Build GRCh38
rs531150856 Chr.14: 67645850-67645850 on Build GRCh38
rs548805126 Chr.14: 67646028-67646028 on Build GRCh38
rs551838214 Chr.14: 67645586-67645586 on Build GRCh38
rs374518024 Chr.14: 67645652-67645652 on Build GRCh38
rs944709587 Chr.14: 67645843-67645843 on Build GRCh38

View Full Product Details