Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 39617643-39618144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSIP1
Entrez Gene ID
161835
Gene Name
fibrous sheath interacting protein 1
Gene Aliases
HSD10
UniGene
Hs.129598
Gene Chromosome Location
Chr.15: 39598643-39782838 on Build GRCh38
Transcripts
NM_001324338.1 NM_152597.4 XM_011521305.2 XM_011521306.1 XM_011521311.2 XM_017021972.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
79364 c.1363G>T p.E455* Chr.15: 39618071-39618071 on Build GRCh38
4925346 c.1699G>A p.D567N Chr.15: 39617735-39617735 on Build GRCh38
147876 c.1583G>C p.G528A Chr.15: 39617851-39617851 on Build GRCh38
3886886 c.1324C>T p.P442S Chr.15: 39618110-39618110 on Build GRCh38
961052 c.1479G>A p.M493I Chr.15: 39617955-39617955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773495377 Chr.15: 39617825-39617827 on Build GRCh38
rs200011665 Chr.15: 39617695-39617695 on Build GRCh38
rs768825908 Chr.15: 39618111-39618111 on Build GRCh38
rs762074052 Chr.15: 39618119-39618119 on Build GRCh38
rs563517813 Chr.15: 39617700-39617700 on Build GRCh38
rs369221759 Chr.15: 39618118-39618118 on Build GRCh38
rs780407277 Chr.15: 39617734-39617734 on Build GRCh38
rs764313047 Chr.15: 39617991-39617991 on Build GRCh38
rs773256450 Chr.15: 39617744-39617744 on Build GRCh38
rs534974332 Chr.15: 39617871-39617871 on Build GRCh38
rs950595397 Chr.15: 39617945-39617945 on Build GRCh38
rs144527032 Chr.15: 39617862-39617862 on Build GRCh38
rs761910838 Chr.15: 39618009-39618009 on Build GRCh38
rs775207502 Chr.15: 39618106-39618106 on Build GRCh38
rs140697593 Chr.15: 39617993-39617993 on Build GRCh38
rs375568122 Chr.15: 39617827-39617827 on Build GRCh38
rs202143597 Chr.15: 39617895-39617895 on Build GRCh38
rs764149658 Chr.15: 39617891-39617891 on Build GRCh38
rs929238289 Chr.15: 39617825-39617825 on Build GRCh38
rs866086807 Chr.15: 39617957-39617957 on Build GRCh38
rs778539429 Chr.15: 39618092-39618092 on Build GRCh38
rs777355700 Chr.15: 39617923-39617923 on Build GRCh38
rs749833668 Chr.15: 39617711-39617711 on Build GRCh38
rs759002765 Chr.15: 39618041-39618041 on Build GRCh38
rs16969386 Chr.15: 39617851-39617851 on Build GRCh38
rs372276786 Chr.15: 39617728-39617728 on Build GRCh38
rs752780154 Chr.15: 39617795-39617795 on Build GRCh38
rs754329190 Chr.15: 39617809-39617809 on Build GRCh38
rs771000827 Chr.15: 39617856-39617856 on Build GRCh38
rs746231115 Chr.15: 39617814-39617814 on Build GRCh38
rs767383065 Chr.15: 39617867-39617867 on Build GRCh38
rs779307434 Chr.15: 39617723-39617723 on Build GRCh38
rs777878961 Chr.15: 39617808-39617808 on Build GRCh38
rs778481230 Chr.15: 39617713-39617714 on Build GRCh38
rs781361486 Chr.15: 39618091-39618091 on Build GRCh38
rs140585650 Chr.15: 39618081-39618081 on Build GRCh38
rs750874989 Chr.15: 39618026-39618026 on Build GRCh38
rs770437289 Chr.15: 39618047-39618047 on Build GRCh38
rs758206272 Chr.15: 39617897-39617897 on Build GRCh38
rs755100987 Chr.15: 39618083-39618083 on Build GRCh38
rs138630370 Chr.15: 39617813-39617813 on Build GRCh38
rs769816620 Chr.15: 39617741-39617741 on Build GRCh38
rs748823652 Chr.15: 39617936-39617936 on Build GRCh38
rs751608898 Chr.15: 39618049-39618054 on Build GRCh38
rs755772848 Chr.15: 39618062-39618062 on Build GRCh38
rs756588220 Chr.15: 39617691-39617691 on Build GRCh38
rs759738449 Chr.15: 39617789-39617789 on Build GRCh38
rs929237617 Chr.15: 39617687-39617687 on Build GRCh38
rs34939408 Chr.15: 39618098-39618099 on Build GRCh38
rs547583532 Chr.15: 39617883-39617883 on Build GRCh38
rs932730458 Chr.15: 39618047-39618047 on Build GRCh38
rs765683503 Chr.15: 39617896-39617896 on Build GRCh38
rs771747853 Chr.15: 39617970-39617970 on Build GRCh38
rs369862285 Chr.15: 39617835-39617835 on Build GRCh38
rs774122223 Chr.15: 39617955-39617955 on Build GRCh38
rs148535405 Chr.15: 39617898-39617898 on Build GRCh38
rs144954858 Chr.15: 39618002-39618002 on Build GRCh38
rs747790598 Chr.15: 39617902-39617902 on Build GRCh38
rs866552400 Chr.15: 39617750-39617750 on Build GRCh38
rs753985104 Chr.15: 39617727-39617727 on Build GRCh38
rs112328201 Chr.15: 39617954-39617954 on Build GRCh38
rs151331752 Chr.15: 39618024-39618024 on Build GRCh38
rs71472405 Chr.15: 39617752-39617752 on Build GRCh38
rs747876924 Chr.15: 39617830-39617830 on Build GRCh38
rs748770210 Chr.15: 39618077-39618077 on Build GRCh38
rs373020273 Chr.15: 39617980-39617980 on Build GRCh38
rs992284730 Chr.15: 39618090-39618090 on Build GRCh38
rs955641149 Chr.15: 39617697-39617697 on Build GRCh38
rs766512158 Chr.15: 39618005-39618006 on Build GRCh38
rs751378418 Chr.15: 39617771-39617771 on Build GRCh38
rs766264299 Chr.15: 39617753-39617753 on Build GRCh38
rs565166709 Chr.15: 39617848-39617848 on Build GRCh38
rs113534700 Chr.15: 39617683-39617683 on Build GRCh38
rs751981845 Chr.15: 39618057-39618057 on Build GRCh38
rs145447896 Chr.15: 39617914-39617914 on Build GRCh38
rs761593473 Chr.15: 39617816-39617816 on Build GRCh38
rs760941846 Chr.15: 39617887-39617887 on Build GRCh38
rs950744731 Chr.15: 39618094-39618094 on Build GRCh38
rs992148366 Chr.15: 39617812-39617812 on Build GRCh38
rs770945752 Chr.15: 39617947-39617947 on Build GRCh38
rs763812568 Chr.15: 39617688-39617688 on Build GRCh38
rs142422775 Chr.15: 39617722-39617722 on Build GRCh38
rs763036515 Chr.15: 39617943-39617943 on Build GRCh38
rs761249507 Chr.15: 39617739-39617739 on Build GRCh38
rs545023586 Chr.15: 39617810-39617810 on Build GRCh38
rs745864260 Chr.15: 39617962-39617962 on Build GRCh38
rs759273718 Chr.15: 39617863-39617863 on Build GRCh38
rs376261887 Chr.15: 39618069-39618069 on Build GRCh38
rs112954305 Chr.15: 39617803-39617803 on Build GRCh38
rs147256576 Chr.15: 39617973-39617973 on Build GRCh38
rs772046954 Chr.15: 39618097-39618097 on Build GRCh38
rs201832093 Chr.15: 39618107-39618107 on Build GRCh38

View Full Product Details