Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 55359976-55360508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCPG1
Entrez Gene ID
9236
Gene Name
cell cycle progression 1
Gene Aliases
CPR8
UniGene
Hs.285051
Gene Chromosome Location
Chr.15: 55355223-55408510 on Build GRCh38
Transcripts
NM_001204450.1 NM_001204451.1 NM_004748.4 NM_020739.3 NR_037923.1

Gene Symbol
DYX1C1-CCPG1
Entrez Gene ID
100533483
Gene Name
DYX1C1-CCPG1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.285051
Gene Chromosome Location
Chr.15: 55355223-55498584 on Build GRCh38
Transcripts
NM_001204450.1 NM_001204451.1 NM_004748.4 NM_020739.3 NR_037923.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4670270 c.1779G>A p.R593R Chr.15: 55359994-55359994 on Build GRCh38
3732249 c.1623G>T p.K541N Chr.15: 55360150-55360150 on Build GRCh38
6050890 c.1447G>A p.E483K Chr.15: 55360326-55360326 on Build GRCh38
4670271 c.1330C>T p.R444C Chr.15: 55360443-55360443 on Build GRCh38
700920 c.1317C>A p.T439T Chr.15: 55360456-55360456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200835536 Chr.15: 55360016-55360016 on Build GRCh38
rs182176640 Chr.15: 55360477-55360477 on Build GRCh38
rs752694937 Chr.15: 55360169-55360169 on Build GRCh38
rs748705203 Chr.15: 55360313-55360313 on Build GRCh38
rs34400683 Chr.15: 55360275-55360276 on Build GRCh38
rs17857026 Chr.15: 55360466-55360466 on Build GRCh38
rs370029108 Chr.15: 55360083-55360083 on Build GRCh38
rs201840284 Chr.15: 55360046-55360046 on Build GRCh38
rs751877374 Chr.15: 55360453-55360453 on Build GRCh38
rs772165467 Chr.15: 55360088-55360088 on Build GRCh38
rs1063565 Chr.15: 55360115-55360115 on Build GRCh38
rs1017699940 Chr.15: 55360125-55360125 on Build GRCh38
rs779100115 Chr.15: 55360075-55360075 on Build GRCh38
rs755867915 Chr.15: 55360174-55360174 on Build GRCh38
rs761139198 Chr.15: 55360008-55360008 on Build GRCh38
rs375002471 Chr.15: 55360480-55360480 on Build GRCh38
rs777588971 Chr.15: 55360181-55360181 on Build GRCh38
rs770261032 Chr.15: 55360322-55360322 on Build GRCh38
rs34699771 Chr.15: 55360273-55360273 on Build GRCh38
rs764926827 Chr.15: 55360030-55360030 on Build GRCh38
rs375733197 Chr.15: 55360154-55360154 on Build GRCh38
rs375939104 Chr.15: 55360256-55360256 on Build GRCh38
rs1006858719 Chr.15: 55360246-55360246 on Build GRCh38
rs190287946 Chr.15: 55360205-55360205 on Build GRCh38
rs781763735 Chr.15: 55360280-55360280 on Build GRCh38
rs773671879 Chr.15: 55360329-55360329 on Build GRCh38
rs750965809 Chr.15: 55360139-55360139 on Build GRCh38
rs757092396 Chr.15: 55360443-55360443 on Build GRCh38
rs755957778 Chr.15: 55360058-55360058 on Build GRCh38
rs747877827 Chr.15: 55360057-55360057 on Build GRCh38
rs761001149 Chr.15: 55360483-55360500 on Build GRCh38
rs752605059 Chr.15: 55360362-55360362 on Build GRCh38
rs753513647 Chr.15: 55360433-55360433 on Build GRCh38
rs762463706 Chr.15: 55360121-55360121 on Build GRCh38
rs777099590 Chr.15: 55360013-55360013 on Build GRCh38
rs764687244 Chr.15: 55360010-55360010 on Build GRCh38
rs753659669 Chr.15: 55360183-55360183 on Build GRCh38
rs373056381 Chr.15: 55360356-55360356 on Build GRCh38
rs780129307 Chr.15: 55360449-55360449 on Build GRCh38
rs570992702 Chr.15: 55360109-55360109 on Build GRCh38
rs1063563 Chr.15: 55360223-55360223 on Build GRCh38
rs780331180 Chr.15: 55360221-55360221 on Build GRCh38
rs1002626663 Chr.15: 55360375-55360375 on Build GRCh38
rs889787225 Chr.15: 55360231-55360231 on Build GRCh38
rs934849328 Chr.15: 55360031-55360031 on Build GRCh38
rs747005790 Chr.15: 55360237-55360237 on Build GRCh38
rs1063564 Chr.15: 55360116-55360116 on Build GRCh38
rs376261837 Chr.15: 55360378-55360378 on Build GRCh38
rs757978993 Chr.15: 55360044-55360044 on Build GRCh38
rs893386585 Chr.15: 55360317-55360317 on Build GRCh38
rs202145045 Chr.15: 55360472-55360472 on Build GRCh38
rs1063562 Chr.15: 55360343-55360343 on Build GRCh38
rs1006051935 Chr.15: 55360387-55360387 on Build GRCh38
rs376868399 Chr.15: 55360467-55360467 on Build GRCh38
rs1036181250 Chr.15: 55360130-55360130 on Build GRCh38
rs763379914 Chr.15: 55360332-55360332 on Build GRCh38
rs750160176 Chr.15: 55360043-55360043 on Build GRCh38
rs201276560 Chr.15: 55360165-55360165 on Build GRCh38
rs758962352 Chr.15: 55360353-55360353 on Build GRCh38
rs766086509 Chr.15: 55360128-55360128 on Build GRCh38
rs200030097 Chr.15: 55360098-55360098 on Build GRCh38
rs762276076 Chr.15: 55360025-55360025 on Build GRCh38
rs1051107484 Chr.15: 55360206-55360206 on Build GRCh38
rs746037942 Chr.15: 55360054-55360054 on Build GRCh38
rs1020476831 Chr.15: 55360350-55360350 on Build GRCh38
rs751102136 Chr.15: 55360052-55360052 on Build GRCh38
rs146958976 Chr.15: 55360407-55360407 on Build GRCh38
rs369329768 Chr.15: 55360352-55360352 on Build GRCh38
rs765764434 Chr.15: 55360118-55360118 on Build GRCh38
rs776030197 Chr.15: 55360000-55360000 on Build GRCh38
rs772151810 Chr.15: 55360103-55360103 on Build GRCh38
rs768980548 Chr.15: 55360107-55360107 on Build GRCh38
rs370202963 Chr.15: 55360474-55360474 on Build GRCh38
rs374633748 Chr.15: 55360416-55360416 on Build GRCh38
rs369002461 Chr.15: 55360162-55360162 on Build GRCh38
rs1063566 Chr.15: 55360003-55360003 on Build GRCh38
rs770431585 Chr.15: 55360347-55360347 on Build GRCh38
rs1010848232 Chr.15: 55360323-55360323 on Build GRCh38

View Full Product Details