Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 70882298-70882826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC49
Entrez Gene ID
54839
Gene Name
leucine rich repeat containing 49
Gene Aliases
PGs4
UniGene
Hs.12692
Gene Chromosome Location
Chr.15: 70852671-71050097 on Build GRCh38
Transcripts
NM_001284357.1 XM_017022357.1 XM_017022359.1 NM_020147.3

Gene Symbol
THAP10
Entrez Gene ID
56906
Gene Name
THAP domain containing 10
Gene Aliases
-
UniGene
Hs.591123
Gene Chromosome Location
Chr.15: 70881342-70892433 on Build GRCh38
Transcripts
NM_001284357.1 XM_017022357.1 XM_017022359.1 NM_020147.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
82411 c.654A>G p.R218R Chr.15: 70882574-70882574 on Build GRCh38
6249523 c.681C>A p.S227S Chr.15: 70882547-70882547 on Build GRCh38
3503561 c.653G>A p.R218K Chr.15: 70882575-70882575 on Build GRCh38
5383757 c.759A>G p.K253K Chr.15: 70882469-70882469 on Build GRCh38
7299245 c.540A>G p.T180T Chr.15: 70882798-70882798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768837499 Chr.15: 70882596-70882596 on Build GRCh38
rs375706115 Chr.15: 70882712-70882712 on Build GRCh38
rs780804830 Chr.15: 70882687-70882687 on Build GRCh38
rs751696400 Chr.15: 70882721-70882725 on Build GRCh38
rs947780542 Chr.15: 70882532-70882532 on Build GRCh38
rs531067507 Chr.15: 70882344-70882344 on Build GRCh38
rs770371874 Chr.15: 70882700-70882700 on Build GRCh38
rs1037390702 Chr.15: 70882399-70882399 on Build GRCh38
rs775801004 Chr.15: 70882442-70882442 on Build GRCh38
rs143098607 Chr.15: 70882628-70882628 on Build GRCh38
rs969445995 Chr.15: 70882589-70882590 on Build GRCh38
rs754847885 Chr.15: 70882681-70882681 on Build GRCh38
rs200741885 Chr.15: 70882799-70882799 on Build GRCh38
rs757388725 Chr.15: 70882475-70882475 on Build GRCh38
rs763113581 Chr.15: 70882550-70882551 on Build GRCh38
rs766428245 Chr.15: 70882669-70882669 on Build GRCh38
rs375372043 Chr.15: 70882604-70882604 on Build GRCh38
rs563940504 Chr.15: 70882436-70882436 on Build GRCh38
rs764288615 Chr.15: 70882446-70882446 on Build GRCh38
rs918490157 Chr.15: 70882383-70882383 on Build GRCh38
rs754759822 Chr.15: 70882509-70882509 on Build GRCh38
rs138467583 Chr.15: 70882560-70882560 on Build GRCh38
rs1011105873 Chr.15: 70882780-70882780 on Build GRCh38
rs138908806 Chr.15: 70882591-70882591 on Build GRCh38
rs373711314 Chr.15: 70882801-70882801 on Build GRCh38
rs759989555 Chr.15: 70882431-70882431 on Build GRCh38
rs999743278 Chr.15: 70882550-70882550 on Build GRCh38
rs917770502 Chr.15: 70882500-70882500 on Build GRCh38
rs372078971 Chr.15: 70882444-70882444 on Build GRCh38
rs752468296 Chr.15: 70882742-70882742 on Build GRCh38
rs150292756 Chr.15: 70882667-70882667 on Build GRCh38
rs375612486 Chr.15: 70882603-70882603 on Build GRCh38
rs935594754 Chr.15: 70882749-70882749 on Build GRCh38
rs890742613 Chr.15: 70882486-70882486 on Build GRCh38
rs746416963 Chr.15: 70882564-70882564 on Build GRCh38
rs776682503 Chr.15: 70882448-70882449 on Build GRCh38
rs924935418 Chr.15: 70882569-70882569 on Build GRCh38
rs148723779 Chr.15: 70882776-70882776 on Build GRCh38
rs145846479 Chr.15: 70882460-70882460 on Build GRCh38
rs774956626 Chr.15: 70882429-70882429 on Build GRCh38
rs772578888 Chr.15: 70882562-70882562 on Build GRCh38
rs747705301 Chr.15: 70882423-70882423 on Build GRCh38
rs748783865 Chr.15: 70882699-70882699 on Build GRCh38
rs900243836 Chr.15: 70882443-70882443 on Build GRCh38
rs759373457 Chr.15: 70882770-70882770 on Build GRCh38
rs79102621 Chr.15: 70882702-70882702 on Build GRCh38
rs200649526 Chr.15: 70882740-70882740 on Build GRCh38
rs144217156 Chr.15: 70882769-70882769 on Build GRCh38
rs780057860 Chr.15: 70882496-70882496 on Build GRCh38
rs550573394 Chr.15: 70882427-70882427 on Build GRCh38
rs765398362 Chr.15: 70882634-70882634 on Build GRCh38
rs758378668 Chr.15: 70882487-70882487 on Build GRCh38
rs755696347 Chr.15: 70882688-70882688 on Build GRCh38
rs766635598 Chr.15: 70882595-70882596 on Build GRCh38
rs960240233 Chr.15: 70882629-70882629 on Build GRCh38
rs774668883 Chr.15: 70882714-70882714 on Build GRCh38
rs755854309 Chr.15: 70882779-70882779 on Build GRCh38
rs941789415 Chr.15: 70882380-70882380 on Build GRCh38
rs542177705 Chr.15: 70882641-70882641 on Build GRCh38
rs761043928 Chr.15: 70882586-70882586 on Build GRCh38
rs1031593930 Chr.15: 70882554-70882554 on Build GRCh38
rs767922286 Chr.15: 70882434-70882434 on Build GRCh38
rs546502746 Chr.15: 70882718-70882718 on Build GRCh38
rs536371046 Chr.15: 70882482-70882482 on Build GRCh38
rs777413231 Chr.15: 70882692-70882692 on Build GRCh38
rs759128348 Chr.15: 70882724-70882724 on Build GRCh38
rs773506483 Chr.15: 70882746-70882746 on Build GRCh38
rs746783487 Chr.15: 70882507-70882507 on Build GRCh38
rs544136510 Chr.15: 70882775-70882775 on Build GRCh38
rs776059168 Chr.15: 70882406-70882406 on Build GRCh38
rs559810721 Chr.15: 70882381-70882381 on Build GRCh38
rs776932721 Chr.15: 70882600-70882600 on Build GRCh38
rs751473165 Chr.15: 70882671-70882671 on Build GRCh38
rs558623535 Chr.15: 70882729-70882729 on Build GRCh38
rs772564155 Chr.15: 70882567-70882567 on Build GRCh38
rs775890804 Chr.15: 70882580-70882580 on Build GRCh38
rs567289905 Chr.15: 70882476-70882476 on Build GRCh38
rs553026947 Chr.15: 70882594-70882594 on Build GRCh38
rs780899374 Chr.15: 70882556-70882556 on Build GRCh38
rs200034991 Chr.15: 70882558-70882558 on Build GRCh38
rs763126144 Chr.15: 70882755-70882755 on Build GRCh38

View Full Product Details