Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 70882625-70883129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC49
Entrez Gene ID
54839
Gene Name
leucine rich repeat containing 49
Gene Aliases
PGs4
UniGene
Hs.12692
Gene Chromosome Location
Chr.15: 70852671-71050097 on Build GRCh38
Transcripts
NM_001284357.1 XM_017022357.1 XM_017022359.1 NM_020147.3

Gene Symbol
THAP10
Entrez Gene ID
56906
Gene Name
THAP domain containing 10
Gene Aliases
-
UniGene
Hs.591123
Gene Chromosome Location
Chr.15: 70881342-70892433 on Build GRCh38
Transcripts
NM_001284357.1 XM_017022357.1 XM_017022359.1 NM_020147.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6998548 c.586G>A p.A196T Chr.15: 70882642-70882642 on Build GRCh38
5968574 c.465C>A p.P155P Chr.15: 70882873-70882873 on Build GRCh38
4733481 c.435G>A p.T145T Chr.15: 70882903-70882903 on Build GRCh38
7299245 c.540A>G p.T180T Chr.15: 70882798-70882798 on Build GRCh38
5515568 c.577+3A>G p.? Chr.15: 70882758-70882758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528727130 Chr.15: 70882956-70882956 on Build GRCh38
rs79239232 Chr.15: 70883063-70883063 on Build GRCh38
rs946245862 Chr.15: 70882851-70882851 on Build GRCh38
rs375706115 Chr.15: 70882712-70882712 on Build GRCh38
rs560972369 Chr.15: 70883104-70883104 on Build GRCh38
rs780804830 Chr.15: 70882687-70882687 on Build GRCh38
rs951391094 Chr.15: 70882942-70882942 on Build GRCh38
rs749470395 Chr.15: 70882866-70882866 on Build GRCh38
rs935035354 Chr.15: 70883105-70883105 on Build GRCh38
rs751696400 Chr.15: 70882721-70882725 on Build GRCh38
rs891422133 Chr.15: 70882983-70882983 on Build GRCh38
rs770371874 Chr.15: 70882700-70882700 on Build GRCh38
rs1039899499 Chr.15: 70883015-70883015 on Build GRCh38
rs747553182 Chr.15: 70882827-70882827 on Build GRCh38
rs754847885 Chr.15: 70882681-70882681 on Build GRCh38
rs770125720 Chr.15: 70882849-70882849 on Build GRCh38
rs200741885 Chr.15: 70882799-70882799 on Build GRCh38
rs137948946 Chr.15: 70883087-70883087 on Build GRCh38
rs372754529 Chr.15: 70882904-70882904 on Build GRCh38
rs766428245 Chr.15: 70882669-70882669 on Build GRCh38
rs574456869 Chr.15: 70882840-70882840 on Build GRCh38
rs756870764 Chr.15: 70882940-70882940 on Build GRCh38
rs771186877 Chr.15: 70882875-70882875 on Build GRCh38
rs760393490 Chr.15: 70882908-70882908 on Build GRCh38
rs1011105873 Chr.15: 70882780-70882780 on Build GRCh38
rs951393516 Chr.15: 70883089-70883089 on Build GRCh38
rs1004604265 Chr.15: 70882948-70882948 on Build GRCh38
rs753423117 Chr.15: 70882811-70882811 on Build GRCh38
rs373711314 Chr.15: 70882801-70882801 on Build GRCh38
rs755198125 Chr.15: 70882935-70882939 on Build GRCh38
rs749887897 Chr.15: 70882950-70882950 on Build GRCh38
rs12909568 Chr.15: 70882828-70882828 on Build GRCh38
rs773596354 Chr.15: 70882865-70882865 on Build GRCh38
rs560280498 Chr.15: 70882943-70882943 on Build GRCh38
rs757863518 Chr.15: 70882955-70882955 on Build GRCh38
rs752468296 Chr.15: 70882742-70882742 on Build GRCh38
rs150292756 Chr.15: 70882667-70882667 on Build GRCh38
rs935594754 Chr.15: 70882749-70882749 on Build GRCh38
rs148723779 Chr.15: 70882776-70882776 on Build GRCh38
rs748783865 Chr.15: 70882699-70882699 on Build GRCh38
rs756793161 Chr.15: 70882815-70882815 on Build GRCh38
rs763838386 Chr.15: 70882922-70882922 on Build GRCh38
rs759373457 Chr.15: 70882770-70882770 on Build GRCh38
rs79102621 Chr.15: 70882702-70882702 on Build GRCh38
rs781663155 Chr.15: 70882833-70882833 on Build GRCh38
rs776938799 Chr.15: 70882807-70882807 on Build GRCh38
rs200649526 Chr.15: 70882740-70882740 on Build GRCh38
rs144217156 Chr.15: 70882769-70882769 on Build GRCh38
rs973249691 Chr.15: 70882990-70882990 on Build GRCh38
rs1007054118 Chr.15: 70882992-70882992 on Build GRCh38
rs774544675 Chr.15: 70882882-70882882 on Build GRCh38
rs913588779 Chr.15: 70882841-70882841 on Build GRCh38
rs936464660 Chr.15: 70882812-70882812 on Build GRCh38
rs755696347 Chr.15: 70882688-70882688 on Build GRCh38
rs1528828 Chr.15: 70882957-70882957 on Build GRCh38
rs774668883 Chr.15: 70882714-70882714 on Build GRCh38
rs755854309 Chr.15: 70882779-70882779 on Build GRCh38
rs750102606 Chr.15: 70883036-70883036 on Build GRCh38
rs150853216 Chr.15: 70882903-70882903 on Build GRCh38
rs546502746 Chr.15: 70882718-70882718 on Build GRCh38
rs759128348 Chr.15: 70882724-70882724 on Build GRCh38
rs777413231 Chr.15: 70882692-70882692 on Build GRCh38
rs773506483 Chr.15: 70882746-70882746 on Build GRCh38
rs778370603 Chr.15: 70882823-70882823 on Build GRCh38
rs544136510 Chr.15: 70882775-70882775 on Build GRCh38
rs565298484 Chr.15: 70883012-70883012 on Build GRCh38
rs751473165 Chr.15: 70882671-70882671 on Build GRCh38
rs558623535 Chr.15: 70882729-70882729 on Build GRCh38
rs753535650 Chr.15: 70882926-70882926 on Build GRCh38
rs963258733 Chr.15: 70882958-70882958 on Build GRCh38
rs995620268 Chr.15: 70883066-70883066 on Build GRCh38
rs1003597602 Chr.15: 70882857-70882857 on Build GRCh38
rs763126144 Chr.15: 70882755-70882755 on Build GRCh38

View Full Product Details