Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2177574-2178082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASKIN1
Entrez Gene ID
57524
Gene Name
CASK interacting protein 1
Gene Aliases
ANKS5A
UniGene
Hs.643537
Gene Chromosome Location
Chr.16: 2177183-2196464 on Build GRCh38
Transcripts
NM_020764.3 NM_032271.2 XM_005255627.4 XM_011522700.1

Gene Symbol
TRAF7
Entrez Gene ID
84231
Gene Name
TNF receptor associated factor 7
Gene Aliases
RFWD1, RNF119
UniGene
Hs.713632
Gene Chromosome Location
Chr.16: 2155752-2178129 on Build GRCh38
Transcripts
NM_020764.3 NM_032271.2 XM_005255627.4 XM_011522700.1

SNP Detail

SNP ID Chromosome Location
rs1007432704 Chr.16: 2177853-2177853 on Build GRCh38
rs1028398353 Chr.16: 2177780-2177780 on Build GRCh38
rs1005545092 Chr.16: 2177694-2177694 on Build GRCh38
rs1043186666 Chr.16: 2177846-2177857 on Build GRCh38
rs954293535 Chr.16: 2177785-2177785 on Build GRCh38
rs1036993616 Chr.16: 2177699-2177699 on Build GRCh38
rs917042380 Chr.16: 2177716-2177716 on Build GRCh38
rs949927513 Chr.16: 2177721-2177721 on Build GRCh38
rs758817740 Chr.16: 2177972-2177972 on Build GRCh38
rs909781831 Chr.16: 2177700-2177700 on Build GRCh38
rs989138669 Chr.16: 2177619-2177619 on Build GRCh38
rs574068108 Chr.16: 2177635-2177635 on Build GRCh38
rs902255134 Chr.16: 2178025-2178025 on Build GRCh38
rs943244183 Chr.16: 2178004-2178004 on Build GRCh38
rs115022524 Chr.16: 2177871-2177871 on Build GRCh38
rs768539963 Chr.16: 2178016-2178016 on Build GRCh38
rs957719621 Chr.16: 2177602-2177602 on Build GRCh38
rs945569915 Chr.16: 2177636-2177636 on Build GRCh38
rs1010629519 Chr.16: 2177860-2177860 on Build GRCh38
rs542338717 Chr.16: 2177908-2177908 on Build GRCh38
rs1007791138 Chr.16: 2177726-2177726 on Build GRCh38
rs868582376 Chr.16: 2177742-2177742 on Build GRCh38
rs928189919 Chr.16: 2177788-2177788 on Build GRCh38
rs555579831 Chr.16: 2177669-2177669 on Build GRCh38
rs138680218 Chr.16: 2177988-2177988 on Build GRCh38
rs982361026 Chr.16: 2177762-2177762 on Build GRCh38
rs182199125 Chr.16: 2177865-2177865 on Build GRCh38
rs957771225 Chr.16: 2177743-2177743 on Build GRCh38
rs993395625 Chr.16: 2177712-2177712 on Build GRCh38
rs777987406 Chr.16: 2177633-2177633 on Build GRCh38
rs1055241257 Chr.16: 2177668-2177668 on Build GRCh38
rs938223208 Chr.16: 2177800-2177800 on Build GRCh38
rs551665155 Chr.16: 2177831-2177831 on Build GRCh38
rs891471879 Chr.16: 2177751-2177751 on Build GRCh38
rs145672192 Chr.16: 2177984-2177984 on Build GRCh38
rs1020648704 Chr.16: 2177864-2177864 on Build GRCh38
rs913013600 Chr.16: 2178027-2178027 on Build GRCh38
rs1020746752 Chr.16: 2178020-2178020 on Build GRCh38
rs954591235 Chr.16: 2177725-2177725 on Build GRCh38
rs888519651 Chr.16: 2177676-2177676 on Build GRCh38
rs890360626 Chr.16: 2177836-2177836 on Build GRCh38
rs552006424 Chr.16: 2178010-2178010 on Build GRCh38
rs564269694 Chr.16: 2177744-2177744 on Build GRCh38
rs967376579 Chr.16: 2177760-2177760 on Build GRCh38
rs1020596149 Chr.16: 2177755-2177755 on Build GRCh38
rs774292421 Chr.16: 2178031-2178031 on Build GRCh38
rs1042654597 Chr.16: 2177901-2177901 on Build GRCh38
rs912848897 Chr.16: 2177875-2177875 on Build GRCh38
rs893566279 Chr.16: 2177858-2177858 on Build GRCh38
rs979931341 Chr.16: 2177837-2177837 on Build GRCh38
rs575544982 Chr.16: 2177707-2177707 on Build GRCh38
rs965541666 Chr.16: 2177656-2177656 on Build GRCh38
rs1006010923 Chr.16: 2177983-2177983 on Build GRCh38
rs747263513 Chr.16: 2177651-2177651 on Build GRCh38
rs945679818 Chr.16: 2178041-2178044 on Build GRCh38
rs982174161 Chr.16: 2177866-2177866 on Build GRCh38
rs959657259 Chr.16: 2178045-2178045 on Build GRCh38
rs998387852 Chr.16: 2177779-2177779 on Build GRCh38
rs865875492 Chr.16: 2177790-2177790 on Build GRCh38
rs1035314049 Chr.16: 2177867-2177867 on Build GRCh38
rs565958781 Chr.16: 2177782-2177782 on Build GRCh38
rs1054508216 Chr.16: 2177745-2177745 on Build GRCh38
rs890380760 Chr.16: 2177989-2177989 on Build GRCh38
rs151281479 Chr.16: 2177637-2177637 on Build GRCh38
rs961353199 Chr.16: 2177986-2177986 on Build GRCh38
rs897134980 Chr.16: 2177829-2177829 on Build GRCh38
rs1017409648 Chr.16: 2177854-2177854 on Build GRCh38
rs539856895 Chr.16: 2177818-2177818 on Build GRCh38
rs544507949 Chr.16: 2177735-2177735 on Build GRCh38
rs910011124 Chr.16: 2177812-2177812 on Build GRCh38
rs1050326044 Chr.16: 2177964-2177964 on Build GRCh38
rs1051714732 Chr.16: 2177833-2177833 on Build GRCh38
rs941462705 Chr.16: 2177939-2177939 on Build GRCh38
rs910019515 Chr.16: 2177921-2177921 on Build GRCh38
rs1042663106 Chr.16: 2177763-2177763 on Build GRCh38
rs998335582 Chr.16: 2177948-2177948 on Build GRCh38
rs1038895788 Chr.16: 2178005-2178005 on Build GRCh38
rs945732094 Chr.16: 2177880-2177880 on Build GRCh38
rs758707042 Chr.16: 2177625-2177625 on Build GRCh38
rs749026241 Chr.16: 2177920-2177920 on Build GRCh38
rs1030593388 Chr.16: 2177719-2177719 on Build GRCh38
rs989146956 Chr.16: 2177747-2177747 on Build GRCh38
rs551483651 Chr.16: 2177820-2177821 on Build GRCh38
rs954062631 Chr.16: 2177680-2177680 on Build GRCh38
rs972768691 Chr.16: 2177943-2177943 on Build GRCh38
rs990942750 Chr.16: 2177810-2177810 on Build GRCh38
rs558063421 Chr.16: 2177594-2177594 on Build GRCh38
rs928124017 Chr.16: 2177648-2177648 on Build GRCh38
rs901338127 Chr.16: 2177776-2177776 on Build GRCh38
rs1035061371 Chr.16: 2178041-2178041 on Build GRCh38
rs577518738 Chr.16: 2177817-2177817 on Build GRCh38
rs528630482 Chr.16: 2177825-2177825 on Build GRCh38
rs918653161 Chr.16: 2177978-2177978 on Build GRCh38

View Full Product Details