Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 5486273-5486762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIS12
Entrez Gene ID
79003
Gene Name
MIS12, kinetochore complex component
Gene Aliases
2510025F08Rik, KNTC2AP, MTW1, hMis12
UniGene
Hs.267194
Gene Chromosome Location
Chr.17: 5486374-5490814 on Build GRCh38
Transcripts
NM_001258217.1 NM_001258218.1 NM_001258219.1 NM_001258220.1 XM_017025035.1

SNP Detail

SNP ID Chromosome Location
rs960703176 Chr.17: 5486616-5486616 on Build GRCh38
rs929315662 Chr.17: 5486318-5486318 on Build GRCh38
rs551748180 Chr.17: 5486362-5486362 on Build GRCh38
rs574201427 Chr.17: 5486306-5486306 on Build GRCh38
rs974819909 Chr.17: 5486312-5486312 on Build GRCh38
rs753629697 Chr.17: 5486705-5486705 on Build GRCh38
rs964869164 Chr.17: 5486557-5486557 on Build GRCh38
rs571373192 Chr.17: 5486384-5486384 on Build GRCh38
rs1031879453 Chr.17: 5486298-5486298 on Build GRCh38
rs1013459266 Chr.17: 5486544-5486544 on Build GRCh38
rs1009213880 Chr.17: 5486362-5486363 on Build GRCh38
rs983554073 Chr.17: 5486597-5486597 on Build GRCh38
rs890917366 Chr.17: 5486406-5486406 on Build GRCh38
rs373213443 Chr.17: 5486413-5486413 on Build GRCh38
rs945161015 Chr.17: 5486740-5486740 on Build GRCh38
rs547137930 Chr.17: 5486364-5486364 on Build GRCh38
rs1029259180 Chr.17: 5486361-5486362 on Build GRCh38
rs956316591 Chr.17: 5486299-5486299 on Build GRCh38
rs547173399 Chr.17: 5486356-5486357 on Build GRCh38
rs1001075134 Chr.17: 5486363-5486364 on Build GRCh38
rs758720341 Chr.17: 5486711-5486711 on Build GRCh38
rs528515753 Chr.17: 5486326-5486326 on Build GRCh38
rs909226713 Chr.17: 5486631-5486631 on Build GRCh38
rs935731202 Chr.17: 5486669-5486669 on Build GRCh38
rs908441605 Chr.17: 5486342-5486342 on Build GRCh38
rs1805434 Chr.17: 5486742-5486742 on Build GRCh38
rs190942528 Chr.17: 5486692-5486692 on Build GRCh38
rs965257938 Chr.17: 5486311-5486311 on Build GRCh38
rs1009150005 Chr.17: 5486418-5486418 on Build GRCh38
rs996564079 Chr.17: 5486360-5486361 on Build GRCh38
rs138511255 Chr.17: 5486438-5486438 on Build GRCh38
rs567473883 Chr.17: 5486385-5486385 on Build GRCh38
rs370948635 Chr.17: 5486357-5486358 on Build GRCh38
rs150601778 Chr.17: 5486357-5486357 on Build GRCh38
rs763802682 Chr.17: 5486663-5486663 on Build GRCh38
rs115036059 Chr.17: 5486636-5486636 on Build GRCh38
rs987579922 Chr.17: 5486308-5486308 on Build GRCh38
rs917943218 Chr.17: 5486315-5486315 on Build GRCh38
rs1019087369 Chr.17: 5486310-5486310 on Build GRCh38
rs975003163 Chr.17: 5486694-5486694 on Build GRCh38
rs1042064585 Chr.17: 5486419-5486419 on Build GRCh38
rs930627970 Chr.17: 5486324-5486324 on Build GRCh38
rs529749471 Chr.17: 5486405-5486405 on Build GRCh38
rs912281203 Chr.17: 5486719-5486719 on Build GRCh38
rs3026156 Chr.17: 5486322-5486322 on Build GRCh38
rs907845954 Chr.17: 5486600-5486600 on Build GRCh38
rs933481007 Chr.17: 5486709-5486709 on Build GRCh38
rs925689131 Chr.17: 5486664-5486664 on Build GRCh38
rs948571295 Chr.17: 5486650-5486650 on Build GRCh38
rs992718001 Chr.17: 5486624-5486624 on Build GRCh38
rs374573350 Chr.17: 5486654-5486654 on Build GRCh38
rs554804954 Chr.17: 5486661-5486661 on Build GRCh38
rs996788721 Chr.17: 5486561-5486561 on Build GRCh38
rs1034037529 Chr.17: 5486472-5486472 on Build GRCh38
rs909414588 Chr.17: 5486364-5486365 on Build GRCh38
rs952136211 Chr.17: 5486582-5486582 on Build GRCh38
rs1805433 Chr.17: 5486568-5486568 on Build GRCh38
rs186918169 Chr.17: 5486294-5486294 on Build GRCh38
rs565466833 Chr.17: 5486363-5486363 on Build GRCh38
rs1009076713 Chr.17: 5486469-5486469 on Build GRCh38
rs939895848 Chr.17: 5486352-5486352 on Build GRCh38
rs891483018 Chr.17: 5486459-5486459 on Build GRCh38
rs999807495 Chr.17: 5486423-5486423 on Build GRCh38
rs377287718 Chr.17: 5486511-5486511 on Build GRCh38
rs538286215 Chr.17: 5486500-5486500 on Build GRCh38

View Full Product Details