Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 31185165-31185671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKAIN1
Entrez Gene ID
79570
Gene Name
Na+/K+ transporting ATPase interacting 1
Gene Aliases
FAM77C
UniGene
Hs.470259
Gene Chromosome Location
Chr.1: 31179270-31239887 on Build GRCh38
Transcripts
NM_024522.2 XM_017002320.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3400661 c.193-2A>G p.? Chr.1: 31185329-31185329 on Build GRCh38
2077084 c.114C>G p.F38L Chr.1: 31185274-31185274 on Build GRCh38
7379652 c.85G>A p.V29I Chr.1: 31185303-31185303 on Build GRCh38
3400660 c.61-2A>G p.? Chr.1: 31185329-31185329 on Build GRCh38
2077085 c.246C>G p.F82L Chr.1: 31185274-31185274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759605510 Chr.1: 31185348-31185348 on Build GRCh38
rs998085577 Chr.1: 31185423-31185423 on Build GRCh38
rs1045686153 Chr.1: 31185290-31185290 on Build GRCh38
rs772549946 Chr.1: 31185218-31185218 on Build GRCh38
rs767560302 Chr.1: 31185361-31185361 on Build GRCh38
rs761905513 Chr.1: 31185265-31185265 on Build GRCh38
rs1051249457 Chr.1: 31185513-31185513 on Build GRCh38
rs923721987 Chr.1: 31185425-31185425 on Build GRCh38
rs994328968 Chr.1: 31185586-31185586 on Build GRCh38
rs986347164 Chr.1: 31185443-31185443 on Build GRCh38
rs953613209 Chr.1: 31185439-31185439 on Build GRCh38
rs150786621 Chr.1: 31185303-31185303 on Build GRCh38
rs117650046 Chr.1: 31185206-31185206 on Build GRCh38
rs1055732379 Chr.1: 31185308-31185308 on Build GRCh38
rs141322561 Chr.1: 31185363-31185363 on Build GRCh38
rs746386319 Chr.1: 31185216-31185216 on Build GRCh38
rs192482380 Chr.1: 31185551-31185551 on Build GRCh38
rs1047234091 Chr.1: 31185643-31185643 on Build GRCh38
rs973871910 Chr.1: 31185511-31185511 on Build GRCh38
rs1017118237 Chr.1: 31185191-31185191 on Build GRCh38
rs756820425 Chr.1: 31185204-31185204 on Build GRCh38
rs375212830 Chr.1: 31185462-31185462 on Build GRCh38
rs746903355 Chr.1: 31185339-31185339 on Build GRCh38
rs756800892 Chr.1: 31185379-31185379 on Build GRCh38
rs977919043 Chr.1: 31185424-31185424 on Build GRCh38
rs1020769906 Chr.1: 31185396-31185396 on Build GRCh38
rs775896900 Chr.1: 31185222-31185222 on Build GRCh38
rs753532553 Chr.1: 31185372-31185372 on Build GRCh38
rs898734477 Chr.1: 31185585-31185585 on Build GRCh38
rs186916373 Chr.1: 31185411-31185411 on Build GRCh38
rs760539771 Chr.1: 31185368-31185368 on Build GRCh38
rs553775280 Chr.1: 31185456-31185456 on Build GRCh38
rs1038094969 Chr.1: 31185217-31185217 on Build GRCh38
rs929868463 Chr.1: 31185591-31185591 on Build GRCh38
rs760360055 Chr.1: 31185435-31185435 on Build GRCh38
rs559224904 Chr.1: 31185257-31185257 on Build GRCh38
rs776936737 Chr.1: 31185259-31185259 on Build GRCh38
rs763755273 Chr.1: 31185370-31185370 on Build GRCh38
rs768532110 Chr.1: 31185453-31185453 on Build GRCh38
rs747365079 Chr.1: 31185255-31185255 on Build GRCh38
rs372026148 Chr.1: 31185299-31185299 on Build GRCh38
rs540670823 Chr.1: 31185423-31185423 on Build GRCh38
rs569789158 Chr.1: 31185237-31185238 on Build GRCh38
rs910908187 Chr.1: 31185517-31185517 on Build GRCh38
rs898682297 Chr.1: 31185200-31185200 on Build GRCh38
rs990745863 Chr.1: 31185388-31185388 on Build GRCh38
rs1038619734 Chr.1: 31185570-31185570 on Build GRCh38

View Full Product Details