Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43217176-43217678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM106A
Entrez Gene ID
113277
Gene Name
transmembrane protein 106A
Gene Aliases
-
UniGene
Hs.536474
Gene Chromosome Location
Chr.17: 43211827-43220041 on Build GRCh38
Transcripts
NM_001291586.1 NM_001291587.1 NM_001291588.1 NM_145041.3 XM_006721658.2 XM_006721659.3 XM_011524260.1 XM_017024113.1 XM_017024114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5770249 c.645C>T p.V215V Chr.17: 43217289-43217289 on Build GRCh38
3378207 c.668+1G>C p.? Chr.17: 43217313-43217313 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs576596116 Chr.17: 43217198-43217198 on Build GRCh38
rs995142784 Chr.17: 43217636-43217636 on Build GRCh38
rs954166159 Chr.17: 43217227-43217227 on Build GRCh38
rs762673168 Chr.17: 43217305-43217305 on Build GRCh38
rs897214821 Chr.17: 43217598-43217598 on Build GRCh38
rs942896157 Chr.17: 43217521-43217521 on Build GRCh38
rs374993878 Chr.17: 43217254-43217254 on Build GRCh38
rs756630239 Chr.17: 43217208-43217208 on Build GRCh38
rs201717997 Chr.17: 43217226-43217226 on Build GRCh38
rs774989880 Chr.17: 43217294-43217294 on Build GRCh38
rs375439542 Chr.17: 43217597-43217597 on Build GRCh38
rs1016315863 Chr.17: 43217605-43217605 on Build GRCh38
rs767716966 Chr.17: 43217308-43217308 on Build GRCh38
rs915741873 Chr.17: 43217372-43217372 on Build GRCh38
rs754394627 Chr.17: 43217337-43217337 on Build GRCh38
rs1051806426 Chr.17: 43217475-43217475 on Build GRCh38
rs761135875 Chr.17: 43217328-43217328 on Build GRCh38
rs371558701 Chr.17: 43217230-43217230 on Build GRCh38
rs565231211 Chr.17: 43217250-43217250 on Build GRCh38
rs1035432093 Chr.17: 43217626-43217626 on Build GRCh38
rs530449086 Chr.17: 43217343-43217343 on Build GRCh38
rs777626761 Chr.17: 43217237-43217237 on Build GRCh38
rs992716204 Chr.17: 43217360-43217360 on Build GRCh38
rs35371523 Chr.17: 43217615-43217615 on Build GRCh38
rs765422377 Chr.17: 43217358-43217358 on Build GRCh38
rs760850406 Chr.17: 43217299-43217300 on Build GRCh38
rs766844488 Chr.17: 43217332-43217332 on Build GRCh38
rs187524967 Chr.17: 43217270-43217270 on Build GRCh38
rs758655941 Chr.17: 43217632-43217632 on Build GRCh38
rs759963121 Chr.17: 43217250-43217250 on Build GRCh38
rs961339152 Chr.17: 43217252-43217252 on Build GRCh38
rs369021981 Chr.17: 43217649-43217649 on Build GRCh38
rs372931504 Chr.17: 43217210-43217210 on Build GRCh38
rs771597742 Chr.17: 43217219-43217219 on Build GRCh38
rs998372533 Chr.17: 43217211-43217211 on Build GRCh38
rs927423727 Chr.17: 43217421-43217421 on Build GRCh38
rs752792127 Chr.17: 43217361-43217361 on Build GRCh38
rs747211557 Chr.17: 43217382-43217382 on Build GRCh38
rs764133300 Chr.17: 43217646-43217646 on Build GRCh38
rs946336446 Chr.17: 43217448-43217448 on Build GRCh38
rs536256137 Chr.17: 43217309-43217309 on Build GRCh38
rs776689307 Chr.17: 43217435-43217435 on Build GRCh38
rs767831866 Chr.17: 43217266-43217267 on Build GRCh38
rs752962121 Chr.17: 43217284-43217284 on Build GRCh38
rs759467744 Chr.17: 43217546-43217546 on Build GRCh38
rs375149576 Chr.17: 43217219-43217219 on Build GRCh38
rs192979255 Chr.17: 43217280-43217280 on Build GRCh38
rs948481893 Chr.17: 43217376-43217376 on Build GRCh38
rs748981220 Chr.17: 43217249-43217250 on Build GRCh38
rs746862874 Chr.17: 43217251-43217251 on Build GRCh38
rs34209148 Chr.17: 43217379-43217379 on Build GRCh38
rs934328952 Chr.17: 43217405-43217405 on Build GRCh38
rs913456477 Chr.17: 43217436-43217436 on Build GRCh38
rs552976853 Chr.17: 43217485-43217485 on Build GRCh38
rs559816440 Chr.17: 43217311-43217311 on Build GRCh38
rs184835376 Chr.17: 43217429-43217429 on Build GRCh38
rs1005613838 Chr.17: 43217583-43217583 on Build GRCh38
rs530516482 Chr.17: 43217503-43217503 on Build GRCh38
rs748462372 Chr.17: 43217234-43217234 on Build GRCh38
rs769475458 Chr.17: 43217289-43217289 on Build GRCh38
rs980150348 Chr.17: 43217388-43217389 on Build GRCh38

View Full Product Details