Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 101268931-101269457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT11
Entrez Gene ID
55571
Gene Name
CCR4-NOT transcription complex subunit 11
Gene Aliases
C2orf29, C40
UniGene
Hs.744080
Gene Chromosome Location
Chr.2: 101252883-101270316 on Build GRCh38
Transcripts
NM_017546.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
85587 c.1325_1335+12del23 p.? Chr.2: 101269126-101269148 on Build GRCh38
1398446 c.1348C>T p.R450C Chr.2: 101269228-101269228 on Build GRCh38
7348000 c.1381C>T p.R461C Chr.2: 101269261-101269261 on Build GRCh38
475665 c.1448G>C p.R483T Chr.2: 101269328-101269328 on Build GRCh38
1613233 c.1283A>T p.Y428F Chr.2: 101269084-101269084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760037019 Chr.2: 101269233-101269233 on Build GRCh38
rs777239365 Chr.2: 101269363-101269374 on Build GRCh38
rs137932829 Chr.2: 101269335-101269335 on Build GRCh38
rs749281273 Chr.2: 101269405-101269405 on Build GRCh38
rs749007189 Chr.2: 101269033-101269034 on Build GRCh38
rs779850238 Chr.2: 101269398-101269398 on Build GRCh38
rs997530825 Chr.2: 101269417-101269417 on Build GRCh38
rs753326454 Chr.2: 101269276-101269276 on Build GRCh38
rs1039498456 Chr.2: 101269202-101269202 on Build GRCh38
rs763713361 Chr.2: 101268995-101268995 on Build GRCh38
rs368313168 Chr.2: 101269106-101269106 on Build GRCh38
rs771080537 Chr.2: 101269145-101269145 on Build GRCh38
rs745915110 Chr.2: 101269381-101269381 on Build GRCh38
rs553388441 Chr.2: 101269194-101269194 on Build GRCh38
rs747296765 Chr.2: 101269136-101269136 on Build GRCh38
rs751103262 Chr.2: 101269341-101269341 on Build GRCh38
rs376617049 Chr.2: 101269435-101269435 on Build GRCh38
rs768504754 Chr.2: 101269182-101269182 on Build GRCh38
rs759153224 Chr.2: 101269194-101269195 on Build GRCh38
rs372136762 Chr.2: 101269403-101269403 on Build GRCh38
rs757586441 Chr.2: 101269359-101269359 on Build GRCh38
rs775547902 Chr.2: 101269180-101269180 on Build GRCh38
rs1046203325 Chr.2: 101269050-101269050 on Build GRCh38
rs762245036 Chr.2: 101269376-101269377 on Build GRCh38
rs370388156 Chr.2: 101269033-101269033 on Build GRCh38
rs535204256 Chr.2: 101269147-101269147 on Build GRCh38
rs781137934 Chr.2: 101269152-101269152 on Build GRCh38
rs911036972 Chr.2: 101269225-101269225 on Build GRCh38
rs764474081 Chr.2: 101269233-101269236 on Build GRCh38
rs75139593 Chr.2: 101269355-101269355 on Build GRCh38
rs771721006 Chr.2: 101269195-101269195 on Build GRCh38
rs374094778 Chr.2: 101269055-101269055 on Build GRCh38
rs751250890 Chr.2: 101269000-101269000 on Build GRCh38
rs774925592 Chr.2: 101269195-101269196 on Build GRCh38
rs756259209 Chr.2: 101269397-101269397 on Build GRCh38
rs765655965 Chr.2: 101269426-101269427 on Build GRCh38
rs770808491 Chr.2: 101269147-101269147 on Build GRCh38
rs762705112 Chr.2: 101269181-101269181 on Build GRCh38
rs769643196 Chr.2: 101269161-101269161 on Build GRCh38
rs17855903 Chr.2: 101269399-101269399 on Build GRCh38
rs768381843 Chr.2: 101269425-101269425 on Build GRCh38
rs745778448 Chr.2: 101269160-101269160 on Build GRCh38
rs781284403 Chr.2: 101269368-101269368 on Build GRCh38
rs761307846 Chr.2: 101269005-101269005 on Build GRCh38
rs766982348 Chr.2: 101269195-101269195 on Build GRCh38
rs765636631 Chr.2: 101269271-101269271 on Build GRCh38
rs371453238 Chr.2: 101268967-101268967 on Build GRCh38
rs754309313 Chr.2: 101269024-101269024 on Build GRCh38
rs199683232 Chr.2: 101268993-101268993 on Build GRCh38
rs761468511 Chr.2: 101269190-101269190 on Build GRCh38
rs1030384840 Chr.2: 101269426-101269426 on Build GRCh38
rs749928791 Chr.2: 101269207-101269207 on Build GRCh38
rs760294301 Chr.2: 101269233-101269234 on Build GRCh38
rs753022927 Chr.2: 101269061-101269061 on Build GRCh38
rs751768106 Chr.2: 101269339-101269339 on Build GRCh38
rs367945274 Chr.2: 101269032-101269032 on Build GRCh38
rs773979969 Chr.2: 101269185-101269185 on Build GRCh38
rs766993170 Chr.2: 101269020-101269020 on Build GRCh38
rs778139241 Chr.2: 101269109-101269109 on Build GRCh38
rs961391443 Chr.2: 101268974-101268974 on Build GRCh38
rs935027073 Chr.2: 101269150-101269150 on Build GRCh38
rs973182090 Chr.2: 101269012-101269012 on Build GRCh38
rs946557625 Chr.2: 101269278-101269278 on Build GRCh38
rs552111367 Chr.2: 101269320-101269320 on Build GRCh38

View Full Product Details