Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 135839961-135840509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCM6
Entrez Gene ID
4175
Gene Name
minichromosome maintenance complex component 6
Gene Aliases
MCG40308, Mis5, P105MCM
UniGene
Hs.444118
Gene Chromosome Location
Chr.2: 135839626-135876477 on Build GRCh38
Transcripts
NM_005915.5

SNP Detail

SNP ID Chromosome Location
rs756532715 Chr.2: 135840293-135840293 on Build GRCh38
rs533605224 Chr.2: 135840178-135840178 on Build GRCh38
rs943880729 Chr.2: 135840234-135840235 on Build GRCh38
rs895529093 Chr.2: 135840314-135840314 on Build GRCh38
rs973395410 Chr.2: 135840129-135840129 on Build GRCh38
rs894835337 Chr.2: 135840300-135840300 on Build GRCh38
rs891803819 Chr.2: 135840194-135840194 on Build GRCh38
rs573646069 Chr.2: 135840236-135840236 on Build GRCh38
rs1012877517 Chr.2: 135840046-135840046 on Build GRCh38
rs921889179 Chr.2: 135840140-135840140 on Build GRCh38
rs549576909 Chr.2: 135840044-135840044 on Build GRCh38
rs866005440 Chr.2: 135840076-135840076 on Build GRCh38
rs1050146410 Chr.2: 135840191-135840191 on Build GRCh38
rs398060567 Chr.2: 135840240-135840242 on Build GRCh38
rs75795578 Chr.2: 135840237-135840239 on Build GRCh38
rs1000576561 Chr.2: 135840238-135840238 on Build GRCh38
rs953431380 Chr.2: 135840425-135840425 on Build GRCh38
rs904804093 Chr.2: 135840255-135840255 on Build GRCh38
rs944136487 Chr.2: 135840103-135840103 on Build GRCh38
rs373134803 Chr.2: 135840355-135840355 on Build GRCh38
rs566551850 Chr.2: 135840102-135840102 on Build GRCh38
rs911715052 Chr.2: 135840437-135840437 on Build GRCh38
rs3739021 Chr.2: 135840342-135840342 on Build GRCh38
rs963402709 Chr.2: 135840106-135840106 on Build GRCh38
rs796452980 Chr.2: 135840181-135840181 on Build GRCh38
rs764778791 Chr.2: 135839989-135839989 on Build GRCh38
rs4988285 Chr.2: 135839996-135839996 on Build GRCh38
rs912462935 Chr.2: 135840203-135840203 on Build GRCh38
rs539223869 Chr.2: 135840011-135840011 on Build GRCh38
rs11656 Chr.2: 135840481-135840481 on Build GRCh38
rs34870773 Chr.2: 135840241-135840241 on Build GRCh38
rs867817359 Chr.2: 135840325-135840325 on Build GRCh38
rs4988284 Chr.2: 135840301-135840301 on Build GRCh38
rs1001684806 Chr.2: 135840263-135840263 on Build GRCh38
rs1040983038 Chr.2: 135840128-135840128 on Build GRCh38
rs931814706 Chr.2: 135840027-135840027 on Build GRCh38
rs575946874 Chr.2: 135840376-135840376 on Build GRCh38
rs1027560039 Chr.2: 135840423-135840423 on Build GRCh38
rs552320009 Chr.2: 135840286-135840287 on Build GRCh38
rs557600805 Chr.2: 135840090-135840090 on Build GRCh38
rs1005023045 Chr.2: 135840413-135840413 on Build GRCh38
rs3087347 Chr.2: 135840430-135840430 on Build GRCh38
rs535528450 Chr.2: 135840290-135840290 on Build GRCh38
rs750757121 Chr.2: 135840073-135840073 on Build GRCh38
rs55710286 Chr.2: 135840224-135840224 on Build GRCh38
rs555352610 Chr.2: 135840205-135840205 on Build GRCh38
rs983566906 Chr.2: 135840100-135840100 on Build GRCh38
rs76855121 Chr.2: 135840235-135840235 on Build GRCh38
rs1029074832 Chr.2: 135840440-135840440 on Build GRCh38
rs973089007 Chr.2: 135840411-135840411 on Build GRCh38
rs796273982 Chr.2: 135840238-135840239 on Build GRCh38
rs200023890 Chr.2: 135840239-135840239 on Build GRCh38
rs182222696 Chr.2: 135840142-135840142 on Build GRCh38
rs774639395 Chr.2: 135840045-135840045 on Build GRCh38
rs1000545277 Chr.2: 135840013-135840013 on Build GRCh38

View Full Product Details