Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 187343536-187344075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCRL
Entrez Gene ID
10203
Gene Name
calcitonin receptor like receptor
Gene Aliases
CGRPR, CRLR
UniGene
Hs.470882
Gene Chromosome Location
Chr.2: 187341963-187448307 on Build GRCh38
Transcripts
NM_001271751.1 NM_005795.5 XM_005246231.3 XM_005246232.3 XM_005246234.3 XM_017003151.1

SNP Detail

SNP ID Chromosome Location
rs1014687742 Chr.2: 187343890-187343890 on Build GRCh38
rs545405303 Chr.2: 187343922-187343922 on Build GRCh38
rs1050171399 Chr.2: 187343611-187343611 on Build GRCh38
rs931692623 Chr.2: 187343600-187343600 on Build GRCh38
rs977724337 Chr.2: 187343735-187343735 on Build GRCh38
rs892687960 Chr.2: 187343748-187343748 on Build GRCh38
rs908188920 Chr.2: 187343886-187343886 on Build GRCh38
rs573603218 Chr.2: 187343617-187343617 on Build GRCh38
rs907712201 Chr.2: 187343940-187343940 on Build GRCh38
rs1028403070 Chr.2: 187343651-187343651 on Build GRCh38
rs1022274536 Chr.2: 187343783-187343783 on Build GRCh38
rs186958976 Chr.2: 187343700-187343700 on Build GRCh38
rs139096882 Chr.2: 187343879-187343879 on Build GRCh38
rs28458997 Chr.2: 187343774-187343774 on Build GRCh38
rs560066897 Chr.2: 187343942-187343942 on Build GRCh38
rs1038629480 Chr.2: 187343975-187343975 on Build GRCh38
rs1031235058 Chr.2: 187343657-187343657 on Build GRCh38
rs1021346952 Chr.2: 187343733-187343734 on Build GRCh38
rs183040183 Chr.2: 187343841-187343841 on Build GRCh38
rs891181562 Chr.2: 187343662-187343662 on Build GRCh38
rs940568492 Chr.2: 187343965-187343965 on Build GRCh38
rs898842693 Chr.2: 187343578-187343578 on Build GRCh38
rs890622094 Chr.2: 187343615-187343615 on Build GRCh38
rs112839817 Chr.2: 187344010-187344010 on Build GRCh38
rs1008248247 Chr.2: 187343695-187343695 on Build GRCh38
rs56035809 Chr.2: 187343563-187343563 on Build GRCh38
rs186536556 Chr.2: 187343861-187343861 on Build GRCh38
rs1009510135 Chr.2: 187343637-187343637 on Build GRCh38
rs915665720 Chr.2: 187343851-187343851 on Build GRCh38
rs553637267 Chr.2: 187343734-187343734 on Build GRCh38
rs940307276 Chr.2: 187343954-187343954 on Build GRCh38
rs16828899 Chr.2: 187343989-187343989 on Build GRCh38
rs959881174 Chr.2: 187343869-187343869 on Build GRCh38
rs769670458 Chr.2: 187344023-187344023 on Build GRCh38
rs546529027 Chr.2: 187344013-187344013 on Build GRCh38
rs932813521 Chr.2: 187344001-187344001 on Build GRCh38
rs144133982 Chr.2: 187344028-187344028 on Build GRCh38
rs191233497 Chr.2: 187343836-187343836 on Build GRCh38
rs779201264 Chr.2: 187343934-187343934 on Build GRCh38

View Full Product Details