Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 187345832-187346341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCRL
Entrez Gene ID
10203
Gene Name
calcitonin receptor like receptor
Gene Aliases
CGRPR, CRLR
UniGene
Hs.470882
Gene Chromosome Location
Chr.2: 187341963-187448307 on Build GRCh38
Transcripts
NM_001271751.1 NM_005795.5 XM_005246231.3 XM_005246232.3 XM_005246234.3 XM_017003151.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3575272 c.1348G>A p.E450K Chr.2: 187346222-187346222 on Build GRCh38
1660959 c.1324A>T p.N442Y Chr.2: 187346246-187346246 on Build GRCh38
6156322 c.1312A>G p.S438G Chr.2: 187346258-187346258 on Build GRCh38
3575273 c.1328G>A p.G443E Chr.2: 187346242-187346242 on Build GRCh38
5590040 c.1327G>A p.G443R Chr.2: 187346243-187346243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985396310 Chr.2: 187346056-187346056 on Build GRCh38
rs1004087889 Chr.2: 187346192-187346192 on Build GRCh38
rs371222442 Chr.2: 187346284-187346284 on Build GRCh38
rs192848670 Chr.2: 187346169-187346169 on Build GRCh38
rs190030563 Chr.2: 187345946-187345946 on Build GRCh38
rs557836271 Chr.2: 187346015-187346016 on Build GRCh38
rs1042361107 Chr.2: 187345864-187345864 on Build GRCh38
rs577853176 Chr.2: 187346217-187346217 on Build GRCh38
rs999201640 Chr.2: 187346086-187346086 on Build GRCh38
rs146718835 Chr.2: 187346313-187346313 on Build GRCh38
rs761424831 Chr.2: 187346314-187346314 on Build GRCh38
rs34127561 Chr.2: 187346007-187346007 on Build GRCh38
rs780308265 Chr.2: 187346252-187346253 on Build GRCh38
rs763791299 Chr.2: 187346038-187346040 on Build GRCh38
rs911219681 Chr.2: 187346089-187346089 on Build GRCh38
rs962078264 Chr.2: 187345914-187345914 on Build GRCh38
rs376050022 Chr.2: 187345859-187345859 on Build GRCh38
rs1025102272 Chr.2: 187345944-187345944 on Build GRCh38
rs144506821 Chr.2: 187345999-187346001 on Build GRCh38
rs367741433 Chr.2: 187346257-187346257 on Build GRCh38
rs556716014 Chr.2: 187345947-187345947 on Build GRCh38
rs370295233 Chr.2: 187345901-187345901 on Build GRCh38
rs755016433 Chr.2: 187346283-187346283 on Build GRCh38
rs751203938 Chr.2: 187346137-187346137 on Build GRCh38
rs761240404 Chr.2: 187346191-187346191 on Build GRCh38
rs376392998 Chr.2: 187346172-187346172 on Build GRCh38
rs553910398 Chr.2: 187345927-187345927 on Build GRCh38
rs763161195 Chr.2: 187346101-187346101 on Build GRCh38
rs770364445 Chr.2: 187346174-187346174 on Build GRCh38
rs773846824 Chr.2: 187346189-187346189 on Build GRCh38
rs748059138 Chr.2: 187346309-187346309 on Build GRCh38
rs199902996 Chr.2: 187346141-187346141 on Build GRCh38
rs945220025 Chr.2: 187346205-187346205 on Build GRCh38
rs1035114393 Chr.2: 187346218-187346218 on Build GRCh38
rs764560081 Chr.2: 187346195-187346195 on Build GRCh38
rs754239021 Chr.2: 187346203-187346203 on Build GRCh38
rs770616041 Chr.2: 187346318-187346318 on Build GRCh38
rs765584434 Chr.2: 187346224-187346224 on Build GRCh38
rs959124617 Chr.2: 187345922-187345922 on Build GRCh38
rs775829315 Chr.2: 187345989-187345989 on Build GRCh38
rs369317777 Chr.2: 187346233-187346233 on Build GRCh38
rs568397597 Chr.2: 187345916-187345916 on Build GRCh38
rs920209143 Chr.2: 187346043-187346043 on Build GRCh38
rs927989855 Chr.2: 187345897-187345897 on Build GRCh38
rs952698865 Chr.2: 187346050-187346050 on Build GRCh38
rs201875819 Chr.2: 187346150-187346150 on Build GRCh38
rs200051216 Chr.2: 187346228-187346228 on Build GRCh38
rs572232395 Chr.2: 187345931-187345931 on Build GRCh38
rs780062941 Chr.2: 187346258-187346258 on Build GRCh38
rs911041913 Chr.2: 187346001-187346002 on Build GRCh38
rs754097092 Chr.2: 187346226-187346228 on Build GRCh38
rs772771162 Chr.2: 187346171-187346171 on Build GRCh38
rs760016081 Chr.2: 187345884-187345884 on Build GRCh38
rs748900537 Chr.2: 187346317-187346317 on Build GRCh38
rs747936357 Chr.2: 187346167-187346167 on Build GRCh38
rs903506812 Chr.2: 187345883-187345883 on Build GRCh38
rs918528011 Chr.2: 187345929-187345929 on Build GRCh38
rs148965095 Chr.2: 187346278-187346278 on Build GRCh38
rs112380150 Chr.2: 187345943-187345943 on Build GRCh38

View Full Product Details