Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 66926060-66926572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIER1
Entrez Gene ID
57708
Gene Name
MIER1 transcriptional regulator
Gene Aliases
ER1, MI-ER1
UniGene
Hs.605432
Gene Chromosome Location
Chr.1: 66924895-66988619 on Build GRCh38
Transcripts
NM_001077700.2 NM_001077702.2 NM_001077703.2 NM_001146110.1 NM_001146111.1 NM_001146112.1 NM_001146113.1 NM_001278215.1 NM_020948.3 XM_005271076.3 XM_011541864.2 XM_011541865.2 XM_017001919.1 XM_017001920.1 XM_017001921.1 XM_017001922.1 XM_017001923.1 XM_017001924.1 XM_017001925.1 XM_017001926.1 XM_017001927.1 XM_017001928.1 XM_017001929.1 XM_017001931.1 XM_017001932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1344031 c.75G>T p.V25V Chr.1: 66926149-66926149 on Build GRCh38
5154452 c.135G>A p.L45L Chr.1: 66926209-66926209 on Build GRCh38
266930 c.117G>A p.W39* Chr.1: 66926191-66926191 on Build GRCh38
6006569 c.128A>T p.N43I Chr.1: 66926202-66926202 on Build GRCh38
6006570 c.128A>T p.N43I Chr.1: 66926202-66926202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942489202 Chr.1: 66926243-66926243 on Build GRCh38
rs111746408 Chr.1: 66926216-66926216 on Build GRCh38
rs374529842 Chr.1: 66926213-66926213 on Build GRCh38
rs116538367 Chr.1: 66926488-66926488 on Build GRCh38
rs965650082 Chr.1: 66926217-66926217 on Build GRCh38
rs896421702 Chr.1: 66926306-66926306 on Build GRCh38
rs1016721108 Chr.1: 66926458-66926459 on Build GRCh38
rs916049140 Chr.1: 66926321-66926321 on Build GRCh38
rs986007883 Chr.1: 66926205-66926205 on Build GRCh38
rs750873137 Chr.1: 66926124-66926124 on Build GRCh38
rs755248591 Chr.1: 66926145-66926145 on Build GRCh38
rs1053541925 Chr.1: 66926398-66926398 on Build GRCh38
rs373259192 Chr.1: 66926115-66926115 on Build GRCh38
rs762267873 Chr.1: 66926112-66926112 on Build GRCh38
rs764598729 Chr.1: 66926095-66926095 on Build GRCh38
rs1007355593 Chr.1: 66926513-66926513 on Build GRCh38
rs377695956 Chr.1: 66926229-66926229 on Build GRCh38
rs112502115 Chr.1: 66926453-66926453 on Build GRCh38
rs545940574 Chr.1: 66926514-66926514 on Build GRCh38
rs1016299107 Chr.1: 66926097-66926097 on Build GRCh38
rs907454544 Chr.1: 66926377-66926377 on Build GRCh38
rs545537020 Chr.1: 66926264-66926264 on Build GRCh38
rs918991395 Chr.1: 66926253-66926253 on Build GRCh38
rs754102601 Chr.1: 66926138-66926138 on Build GRCh38
rs202108897 Chr.1: 66926305-66926305 on Build GRCh38
rs536685746 Chr.1: 66926105-66926105 on Build GRCh38
rs953262304 Chr.1: 66926196-66926196 on Build GRCh38
rs779102352 Chr.1: 66926152-66926152 on Build GRCh38
rs771512718 Chr.1: 66926117-66926117 on Build GRCh38
rs965953751 Chr.1: 66926528-66926528 on Build GRCh38
rs370311228 Chr.1: 66926281-66926281 on Build GRCh38
rs866612524 Chr.1: 66926515-66926515 on Build GRCh38
rs559500017 Chr.1: 66926277-66926277 on Build GRCh38
rs556519918 Chr.1: 66926135-66926135 on Build GRCh38
rs530810935 Chr.1: 66926531-66926531 on Build GRCh38
rs924974996 Chr.1: 66926297-66926297 on Build GRCh38
rs770686765 Chr.1: 66926161-66926161 on Build GRCh38
rs762541860 Chr.1: 66926231-66926231 on Build GRCh38
rs369976632 Chr.1: 66926194-66926194 on Build GRCh38
rs984587390 Chr.1: 66926296-66926296 on Build GRCh38
rs767725108 Chr.1: 66926276-66926276 on Build GRCh38
rs948897698 Chr.1: 66926324-66926324 on Build GRCh38
rs986287533 Chr.1: 66926215-66926215 on Build GRCh38
rs748660929 Chr.1: 66926468-66926468 on Build GRCh38
rs1045979037 Chr.1: 66926372-66926372 on Build GRCh38
rs762318844 Chr.1: 66926272-66926272 on Build GRCh38
rs200891260 Chr.1: 66926098-66926098 on Build GRCh38
rs768494786 Chr.1: 66926260-66926260 on Build GRCh38
rs1044958486 Chr.1: 66926359-66926359 on Build GRCh38
rs905018580 Chr.1: 66926452-66926452 on Build GRCh38
rs746845950 Chr.1: 66926157-66926157 on Build GRCh38
rs751400856 Chr.1: 66926289-66926289 on Build GRCh38
rs893164006 Chr.1: 66926402-66926402 on Build GRCh38
rs781206860 Chr.1: 66926171-66926171 on Build GRCh38
rs1010710842 Chr.1: 66926153-66926153 on Build GRCh38
rs774929647 Chr.1: 66926383-66926383 on Build GRCh38
rs769347989 Chr.1: 66926202-66926202 on Build GRCh38
rs201496882 Chr.1: 66926542-66926542 on Build GRCh38
rs35543659 Chr.1: 66926331-66926332 on Build GRCh38
rs768180074 Chr.1: 66926118-66926118 on Build GRCh38
rs191540051 Chr.1: 66926188-66926188 on Build GRCh38

View Full Product Details