Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44196530-44197030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSER1
Entrez Gene ID
51526
Gene Name
oxidative stress responsive serine rich 1
Gene Aliases
C20orf111, HSPC207, Osr1, Perit1, dJ1183I21.1
UniGene
Hs.75798
Gene Chromosome Location
Chr.20: 44195939-44217229 on Build GRCh38
Transcripts
NM_016470.7 XM_011528851.1 XM_011528853.2 XM_011528854.2 XM_017027873.1

SNP Detail

SNP ID Chromosome Location
rs964488368 Chr.20: 44196880-44196880 on Build GRCh38
rs886549583 Chr.20: 44196851-44196851 on Build GRCh38
rs974600395 Chr.20: 44196633-44196633 on Build GRCh38
rs533823505 Chr.20: 44196629-44196629 on Build GRCh38
rs923718919 Chr.20: 44196701-44196701 on Build GRCh38
rs537894783 Chr.20: 44196593-44196593 on Build GRCh38
rs377481938 Chr.20: 44196887-44196887 on Build GRCh38
rs1018763959 Chr.20: 44196624-44196624 on Build GRCh38
rs899173208 Chr.20: 44196814-44196814 on Build GRCh38
rs1026784151 Chr.20: 44196596-44196596 on Build GRCh38
rs1044864074 Chr.20: 44196865-44196865 on Build GRCh38
rs1016242483 Chr.20: 44196606-44196606 on Build GRCh38
rs542042475 Chr.20: 44196674-44196674 on Build GRCh38
rs953731361 Chr.20: 44196661-44196661 on Build GRCh38
rs139098229 Chr.20: 44196860-44196860 on Build GRCh38
rs767100617 Chr.20: 44197005-44197005 on Build GRCh38
rs1019205232 Chr.20: 44196872-44196872 on Build GRCh38
rs1029250947 Chr.20: 44196635-44196635 on Build GRCh38
rs1008972998 Chr.20: 44196870-44196870 on Build GRCh38
rs148618772 Chr.20: 44196608-44196608 on Build GRCh38
rs1013155749 Chr.20: 44196558-44196558 on Build GRCh38
rs1044430878 Chr.20: 44196576-44196576 on Build GRCh38
rs9875 Chr.20: 44196847-44196847 on Build GRCh38
rs145184351 Chr.20: 44197006-44197006 on Build GRCh38
rs977817181 Chr.20: 44196685-44196685 on Build GRCh38
rs370270936 Chr.20: 44196832-44196832 on Build GRCh38
rs998369067 Chr.20: 44196947-44196950 on Build GRCh38
rs191815961 Chr.20: 44196802-44196802 on Build GRCh38
rs968267208 Chr.20: 44196704-44196704 on Build GRCh38
rs1029882226 Chr.20: 44196949-44196949 on Build GRCh38
rs923843697 Chr.20: 44196790-44196790 on Build GRCh38
rs753897471 Chr.20: 44196919-44196920 on Build GRCh38
rs1022132662 Chr.20: 44196930-44196930 on Build GRCh38
rs889435907 Chr.20: 44196966-44196966 on Build GRCh38
rs756643518 Chr.20: 44196861-44196861 on Build GRCh38
rs150735901 Chr.20: 44196671-44196671 on Build GRCh38
rs774585929 Chr.20: 44196609-44196609 on Build GRCh38
rs977989609 Chr.20: 44196753-44196753 on Build GRCh38
rs767504272 Chr.20: 44196755-44196755 on Build GRCh38
rs113580409 Chr.20: 44196972-44196972 on Build GRCh38
rs996051132 Chr.20: 44196924-44196924 on Build GRCh38
rs879002742 Chr.20: 44196719-44196722 on Build GRCh38
rs895826458 Chr.20: 44196557-44196557 on Build GRCh38
rs985147587 Chr.20: 44196676-44196676 on Build GRCh38
rs939111778 Chr.20: 44196736-44196736 on Build GRCh38
rs948715024 Chr.20: 44196848-44196848 on Build GRCh38
rs41282014 Chr.20: 44196590-44196590 on Build GRCh38
rs370458663 Chr.20: 44196952-44196952 on Build GRCh38
rs142072453 Chr.20: 44196655-44196655 on Build GRCh38
rs1022458817 Chr.20: 44197004-44197005 on Build GRCh38
rs1057489502 Chr.20: 44196743-44196743 on Build GRCh38
rs187322793 Chr.20: 44196625-44196625 on Build GRCh38
rs899141765 Chr.20: 44196586-44196586 on Build GRCh38
rs948968661 Chr.20: 44196780-44196780 on Build GRCh38
rs1005750172 Chr.20: 44196600-44196600 on Build GRCh38

View Full Product Details