Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44960065-44960585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOMM34
Entrez Gene ID
10953
Gene Name
translocase of outer mitochondrial membrane 34
Gene Aliases
HTOM34P, TOM34, URCC3
UniGene
Hs.517066
Gene Chromosome Location
Chr.20: 44942130-44960520 on Build GRCh38
Transcripts
NM_006809.4 XM_005260254.2 XM_017027600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5796526 c.72G>C p.Q24H Chr.20: 44960262-44960262 on Build GRCh38
5818387 c.6C>A p.A2A Chr.20: 44960328-44960328 on Build GRCh38
6806075 c.43G>A p.A15T Chr.20: 44960291-44960291 on Build GRCh38
1027141 c.22T>G p.S8A Chr.20: 44960312-44960312 on Build GRCh38
6248510 c.77C>A p.A26D Chr.20: 44960257-44960257 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs937005483 Chr.20: 44960116-44960116 on Build GRCh38
rs2234199 Chr.20: 44960192-44960192 on Build GRCh38
rs1042497454 Chr.20: 44960449-44960449 on Build GRCh38
rs528637350 Chr.20: 44960327-44960327 on Build GRCh38
rs549225406 Chr.20: 44960180-44960180 on Build GRCh38
rs751267785 Chr.20: 44960199-44960199 on Build GRCh38
rs147512772 Chr.20: 44960378-44960379 on Build GRCh38
rs777186463 Chr.20: 44960289-44960289 on Build GRCh38
rs547044009 Chr.20: 44960473-44960473 on Build GRCh38
rs531020626 Chr.20: 44960361-44960361 on Build GRCh38
rs773121896 Chr.20: 44960373-44960374 on Build GRCh38
rs757145523 Chr.20: 44960247-44960247 on Build GRCh38
rs368500690 Chr.20: 44960350-44960350 on Build GRCh38
rs772241307 Chr.20: 44960179-44960179 on Build GRCh38
rs765513123 Chr.20: 44960197-44960197 on Build GRCh38
rs768705567 Chr.20: 44960288-44960288 on Build GRCh38
rs890822210 Chr.20: 44960484-44960484 on Build GRCh38
rs899610764 Chr.20: 44960508-44960508 on Build GRCh38
rs375970042 Chr.20: 44960238-44960238 on Build GRCh38
rs957632947 Chr.20: 44960410-44960410 on Build GRCh38
rs1009035198 Chr.20: 44960221-44960221 on Build GRCh38
rs751825883 Chr.20: 44960384-44960384 on Build GRCh38
rs767011488 Chr.20: 44960318-44960318 on Build GRCh38
rs1009537215 Chr.20: 44960421-44960421 on Build GRCh38
rs919805576 Chr.20: 44960477-44960477 on Build GRCh38
rs971745351 Chr.20: 44960517-44960517 on Build GRCh38
rs1033150268 Chr.20: 44960526-44960526 on Build GRCh38
rs34668024 Chr.20: 44960090-44960091 on Build GRCh38
rs529247322 Chr.20: 44960489-44960489 on Build GRCh38
rs908873051 Chr.20: 44960138-44960138 on Build GRCh38
rs766129966 Chr.20: 44960382-44960382 on Build GRCh38
rs910404800 Chr.20: 44960561-44960561 on Build GRCh38
rs933137244 Chr.20: 44960167-44960167 on Build GRCh38
rs753873648 Chr.20: 44960337-44960337 on Build GRCh38
rs769079759 Chr.20: 44960191-44960191 on Build GRCh38
rs1045981278 Chr.20: 44960152-44960152 on Build GRCh38
rs761201459 Chr.20: 44960187-44960187 on Build GRCh38
rs772245329 Chr.20: 44960209-44960210 on Build GRCh38
rs959784941 Chr.20: 44960482-44960482 on Build GRCh38
rs770246135 Chr.20: 44960311-44960311 on Build GRCh38
rs369783328 Chr.20: 44960378-44960378 on Build GRCh38
rs748715499 Chr.20: 44960357-44960357 on Build GRCh38
rs566837004 Chr.20: 44960174-44960174 on Build GRCh38
rs971312272 Chr.20: 44960305-44960305 on Build GRCh38
rs2234197 Chr.20: 44960400-44960400 on Build GRCh38
rs2234194 Chr.20: 44960466-44960466 on Build GRCh38
rs1053679885 Chr.20: 44960403-44960403 on Build GRCh38
rs2234196 Chr.20: 44960438-44960438 on Build GRCh38
rs913040059 Chr.20: 44960115-44960115 on Build GRCh38
rs560301411 Chr.20: 44960481-44960481 on Build GRCh38
rs375679371 Chr.20: 44960541-44960541 on Build GRCh38
rs757426666 Chr.20: 44960450-44960450 on Build GRCh38
rs181732423 Chr.20: 44960178-44960178 on Build GRCh38
rs779255539 Chr.20: 44960168-44960168 on Build GRCh38
rs893793986 Chr.20: 44960203-44960203 on Build GRCh38
rs971797674 Chr.20: 44960376-44960376 on Build GRCh38
rs756125775 Chr.20: 44960223-44960223 on Build GRCh38
rs954515403 Chr.20: 44960550-44960550 on Build GRCh38
rs577351004 Chr.20: 44960354-44960354 on Build GRCh38
rs957391756 Chr.20: 44960287-44960287 on Build GRCh38
rs778548835 Chr.20: 44960262-44960262 on Build GRCh38
rs369673094 Chr.20: 44960377-44960377 on Build GRCh38
rs918912073 Chr.20: 44960545-44960545 on Build GRCh38
rs571007503 Chr.20: 44960114-44960114 on Build GRCh38
rs1052121054 Chr.20: 44960175-44960175 on Build GRCh38
rs1005775444 Chr.20: 44960485-44960485 on Build GRCh38
rs988787440 Chr.20: 44960414-44960414 on Build GRCh38
rs768368648 Chr.20: 44960408-44960408 on Build GRCh38
rs371576618 Chr.20: 44960339-44960339 on Build GRCh38
rs189874196 Chr.20: 44960478-44960478 on Build GRCh38
rs1010938170 Chr.20: 44960553-44960553 on Build GRCh38
rs749025823 Chr.20: 44960157-44960157 on Build GRCh38
rs374740712 Chr.20: 44960279-44960279 on Build GRCh38
rs748620288 Chr.20: 44960295-44960295 on Build GRCh38
rs764066256 Chr.20: 44960224-44960224 on Build GRCh38
rs762198125 Chr.20: 44960190-44960190 on Build GRCh38
rs765110323 Chr.20: 44960344-44960344 on Build GRCh38
rs568714701 Chr.20: 44960211-44960211 on Build GRCh38
rs2234200 Chr.20: 44960161-44960161 on Build GRCh38
rs8113914 Chr.20: 44960336-44960336 on Build GRCh38
rs1009674166 Chr.20: 44960232-44960232 on Build GRCh38
rs112033922 Chr.20: 44960149-44960149 on Build GRCh38
rs746123677 Chr.20: 44960265-44960265 on Build GRCh38
rs750118449 Chr.20: 44960347-44960347 on Build GRCh38
rs544862824 Chr.20: 44960402-44960402 on Build GRCh38
rs542165757 Chr.20: 44960368-44960368 on Build GRCh38
rs562325240 Chr.20: 44960371-44960371 on Build GRCh38
rs913262177 Chr.20: 44960416-44960416 on Build GRCh38
rs971497102 Chr.20: 44960427-44960427 on Build GRCh38
rs752219115 Chr.20: 44960222-44960222 on Build GRCh38
rs2234198 Chr.20: 44960367-44960367 on Build GRCh38
rs941811627 Chr.20: 44960148-44960148 on Build GRCh38
rs770375757 Chr.20: 44960107-44960107 on Build GRCh38
rs955311904 Chr.20: 44960193-44960193 on Build GRCh38
rs533862044 Chr.20: 44960356-44960356 on Build GRCh38
rs774283876 Chr.20: 44960117-44960117 on Build GRCh38
rs973913263 Chr.20: 44960476-44960476 on Build GRCh38
rs747225443 Chr.20: 44960353-44960353 on Build GRCh38
rs773402878 Chr.20: 44960313-44960313 on Build GRCh38
rs780112071 Chr.20: 44960282-44960282 on Build GRCh38
rs970897001 Chr.20: 44960091-44960091 on Build GRCh38
rs749557889 Chr.20: 44960365-44960365 on Build GRCh38
rs982785197 Chr.20: 44960097-44960097 on Build GRCh38
rs368407728 Chr.20: 44960558-44960558 on Build GRCh38
rs1039452460 Chr.20: 44960506-44960506 on Build GRCh38
rs910365993 Chr.20: 44960389-44960389 on Build GRCh38
rs2234195 Chr.20: 44960454-44960454 on Build GRCh38
rs759268746 Chr.20: 44960316-44960316 on Build GRCh38
rs1019535570 Chr.20: 44960234-44960234 on Build GRCh38
rs747029403 Chr.20: 44960284-44960284 on Build GRCh38
rs774907011 Chr.20: 44960165-44960165 on Build GRCh38
rs963021041 Chr.20: 44960502-44960502 on Build GRCh38
rs780309180 Chr.20: 44960351-44960351 on Build GRCh38
rs201330529 Chr.20: 44960323-44960323 on Build GRCh38

View Full Product Details