Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 57169660-57170159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP7
Entrez Gene ID
655
Gene Name
bone morphogenetic protein 7
Gene Aliases
OP-1
UniGene
Hs.473163
Gene Chromosome Location
Chr.20: 57168753-57266651 on Build GRCh38
Transcripts
NM_001719.2

SNP Detail

SNP ID Chromosome Location
rs181499788 Chr.20: 57170057-57170057 on Build GRCh38
rs749875399 Chr.20: 57169733-57169733 on Build GRCh38
rs74182711 Chr.20: 57169848-57169849 on Build GRCh38
rs1023545871 Chr.20: 57169903-57169903 on Build GRCh38
rs1014754134 Chr.20: 57170010-57170010 on Build GRCh38
rs371643626 Chr.20: 57169760-57169760 on Build GRCh38
rs994676419 Chr.20: 57169835-57169835 on Build GRCh38
rs1019399846 Chr.20: 57169850-57169850 on Build GRCh38
rs542624240 Chr.20: 57169760-57169760 on Build GRCh38
rs538342951 Chr.20: 57170098-57170098 on Build GRCh38
rs1027755235 Chr.20: 57169956-57169956 on Build GRCh38
rs930125443 Chr.20: 57170095-57170095 on Build GRCh38
rs775358430 Chr.20: 57169820-57169820 on Build GRCh38
rs767463809 Chr.20: 57169692-57169692 on Build GRCh38
rs900573462 Chr.20: 57169864-57169864 on Build GRCh38
rs6025418 Chr.20: 57170017-57170017 on Build GRCh38
rs996687060 Chr.20: 57169879-57169879 on Build GRCh38
rs991789265 Chr.20: 57169734-57169734 on Build GRCh38
rs573978388 Chr.20: 57169726-57169726 on Build GRCh38
rs6025417 Chr.20: 57169849-57169849 on Build GRCh38
rs1052133020 Chr.20: 57169689-57169689 on Build GRCh38
rs969270323 Chr.20: 57169915-57169915 on Build GRCh38
rs145397954 Chr.20: 57169788-57169788 on Build GRCh38
rs935065081 Chr.20: 57169684-57169684 on Build GRCh38
rs923014021 Chr.20: 57169999-57169999 on Build GRCh38
rs951941420 Chr.20: 57169959-57169959 on Build GRCh38
rs565066120 Chr.20: 57169877-57169877 on Build GRCh38
rs985792678 Chr.20: 57170012-57170012 on Build GRCh38
rs983305363 Chr.20: 57169704-57169704 on Build GRCh38
rs898673149 Chr.20: 57170079-57170079 on Build GRCh38
rs370587136 Chr.20: 57169998-57169998 on Build GRCh38
rs896565622 Chr.20: 57170101-57170101 on Build GRCh38
rs956543146 Chr.20: 57169976-57169976 on Build GRCh38
rs1025699099 Chr.20: 57169838-57169838 on Build GRCh38
rs35613217 Chr.20: 57169700-57169701 on Build GRCh38
rs765973657 Chr.20: 57169836-57169836 on Build GRCh38
rs529708449 Chr.20: 57170052-57170052 on Build GRCh38
rs550856258 Chr.20: 57170009-57170009 on Build GRCh38
rs557327761 Chr.20: 57169710-57169710 on Build GRCh38
rs1032117675 Chr.20: 57169943-57169943 on Build GRCh38
rs546262592 Chr.20: 57170053-57170053 on Build GRCh38
rs1013575979 Chr.20: 57170125-57170125 on Build GRCh38
rs1044556655 Chr.20: 57169778-57169778 on Build GRCh38
rs75929478 Chr.20: 57169769-57169769 on Build GRCh38
rs79814067 Chr.20: 57169701-57169701 on Build GRCh38
rs936167271 Chr.20: 57169819-57169819 on Build GRCh38
rs1051354359 Chr.20: 57169707-57169707 on Build GRCh38
rs6025416 Chr.20: 57169848-57169848 on Build GRCh38
rs979263943 Chr.20: 57169918-57169918 on Build GRCh38
rs947830407 Chr.20: 57169773-57169773 on Build GRCh38
rs879573029 Chr.20: 57169848-57169849 on Build GRCh38
rs932973540 Chr.20: 57169702-57169702 on Build GRCh38

View Full Product Details