Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62881656-62882156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIDO1
Entrez Gene ID
11083
Gene Name
death inducer-obliterator 1
Gene Aliases
BYE1, C20orf158, DATF-1, DATF1, DIDO2, DIDO3, DIO-1, DIO1, dJ885L7.8
UniGene
Hs.517172
Gene Chromosome Location
Chr.20: 62877738-62937952 on Build GRCh38
Transcripts
NM_001193369.1 NM_033081.2 XM_011528504.1 XM_011528505.1 XM_011528506.1 XM_011528507.1 XM_011528508.1 XM_011528509.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6439623 c.3902C>T p.S1301F Chr.20: 62882054-62882054 on Build GRCh38
6643997 c.3909C>T p.T1303T Chr.20: 62882047-62882047 on Build GRCh38
1190307 c.4061T>G p.V1354G Chr.20: 62881895-62881895 on Build GRCh38
3841463 c.4152A>G p.P1384P Chr.20: 62881804-62881804 on Build GRCh38
1578837 c.4066G>T p.A1356S Chr.20: 62881890-62881890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757294711 Chr.20: 62881698-62881698 on Build GRCh38
rs143268485 Chr.20: 62881716-62881716 on Build GRCh38
rs564972806 Chr.20: 62881881-62881881 on Build GRCh38
rs960059308 Chr.20: 62881724-62881724 on Build GRCh38
rs759533170 Chr.20: 62882026-62882026 on Build GRCh38
rs142794112 Chr.20: 62881968-62881968 on Build GRCh38
rs747268602 Chr.20: 62882091-62882091 on Build GRCh38
rs755315237 Chr.20: 62881787-62881787 on Build GRCh38
rs768778660 Chr.20: 62881927-62881927 on Build GRCh38
rs757777470 Chr.20: 62882047-62882047 on Build GRCh38
rs764398815 Chr.20: 62882041-62882041 on Build GRCh38
rs929959168 Chr.20: 62882100-62882100 on Build GRCh38
rs574536133 Chr.20: 62881709-62881709 on Build GRCh38
rs374946091 Chr.20: 62881783-62881783 on Build GRCh38
rs745976405 Chr.20: 62882079-62882079 on Build GRCh38
rs769832742 Chr.20: 62882008-62882008 on Build GRCh38
rs773924861 Chr.20: 62881830-62881830 on Build GRCh38
rs940328539 Chr.20: 62881877-62881877 on Build GRCh38
rs762160593 Chr.20: 62882105-62882105 on Build GRCh38
rs766723373 Chr.20: 62881884-62881884 on Build GRCh38
rs139155956 Chr.20: 62882036-62882036 on Build GRCh38
rs777037587 Chr.20: 62882104-62882104 on Build GRCh38
rs746766559 Chr.20: 62881926-62881926 on Build GRCh38
rs770549757 Chr.20: 62881731-62881731 on Build GRCh38
rs756705280 Chr.20: 62881894-62881894 on Build GRCh38
rs755493984 Chr.20: 62881714-62881714 on Build GRCh38
rs145621116 Chr.20: 62881948-62881948 on Build GRCh38
rs558826220 Chr.20: 62882063-62882063 on Build GRCh38
rs772882940 Chr.20: 62881718-62881718 on Build GRCh38
rs199834139 Chr.20: 62882107-62882107 on Build GRCh38
rs1048447964 Chr.20: 62881892-62881892 on Build GRCh38
rs775439423 Chr.20: 62881752-62881752 on Build GRCh38
rs759369005 Chr.20: 62881736-62881736 on Build GRCh38
rs759855227 Chr.20: 62881947-62881947 on Build GRCh38
rs752986067 Chr.20: 62881893-62881893 on Build GRCh38
rs6062735 Chr.20: 62882016-62882016 on Build GRCh38
rs746822914 Chr.20: 62881993-62881993 on Build GRCh38
rs775809565 Chr.20: 62882087-62882087 on Build GRCh38
rs756695576 Chr.20: 62881903-62881903 on Build GRCh38
rs781676976 Chr.20: 62881715-62881715 on Build GRCh38
rs771850719 Chr.20: 62881839-62881839 on Build GRCh38
rs372016333 Chr.20: 62881772-62881772 on Build GRCh38
rs748136945 Chr.20: 62882007-62882007 on Build GRCh38
rs150301288 Chr.20: 62881882-62881882 on Build GRCh38
rs748980038 Chr.20: 62881729-62881729 on Build GRCh38
rs758739203 Chr.20: 62881775-62881775 on Build GRCh38
rs369491823 Chr.20: 62881768-62881768 on Build GRCh38
rs748652457 Chr.20: 62881804-62881804 on Build GRCh38
rs138200370 Chr.20: 62881967-62881967 on Build GRCh38
rs762099681 Chr.20: 62881940-62881940 on Build GRCh38
rs536712014 Chr.20: 62881953-62881953 on Build GRCh38
rs918526271 Chr.20: 62882096-62882096 on Build GRCh38
rs751784204 Chr.20: 62881887-62881887 on Build GRCh38
rs775321335 Chr.20: 62881854-62881854 on Build GRCh38
rs1031709938 Chr.20: 62881950-62881950 on Build GRCh38
rs750015505 Chr.20: 62882046-62882046 on Build GRCh38
rs779567709 Chr.20: 62881985-62881985 on Build GRCh38
rs760703565 Chr.20: 62881866-62881866 on Build GRCh38
rs138473301 Chr.20: 62881963-62881963 on Build GRCh38
rs768524936 Chr.20: 62881996-62881996 on Build GRCh38
rs536977764 Chr.20: 62882054-62882054 on Build GRCh38
rs141398076 Chr.20: 62882078-62882078 on Build GRCh38
rs774265030 Chr.20: 62881945-62881945 on Build GRCh38
rs878996913 Chr.20: 62881859-62881859 on Build GRCh38
rs1020358744 Chr.20: 62881759-62881759 on Build GRCh38
rs761047955 Chr.20: 62882039-62882039 on Build GRCh38
rs781576393 Chr.20: 62881800-62881800 on Build GRCh38
rs769302795 Chr.20: 62881717-62881717 on Build GRCh38
rs771410043 Chr.20: 62881974-62881976 on Build GRCh38
rs984064280 Chr.20: 62882114-62882114 on Build GRCh38
rs146696480 Chr.20: 62881732-62881732 on Build GRCh38
rs148894471 Chr.20: 62881870-62881870 on Build GRCh38
rs762356761 Chr.20: 62882042-62882042 on Build GRCh38
rs781631041 Chr.20: 62881992-62881993 on Build GRCh38
rs374530132 Chr.20: 62882111-62882111 on Build GRCh38
rs762558487 Chr.20: 62882110-62882110 on Build GRCh38
rs766080364 Chr.20: 62882060-62882060 on Build GRCh38
rs1013566695 Chr.20: 62882102-62882102 on Build GRCh38
rs867592340 Chr.20: 62882106-62882106 on Build GRCh38
rs766692767 Chr.20: 62881777-62881777 on Build GRCh38
rs146468791 Chr.20: 62881975-62881975 on Build GRCh38
rs547360425 Chr.20: 62881896-62881896 on Build GRCh38
rs751092911 Chr.20: 62882131-62882131 on Build GRCh38
rs978620782 Chr.20: 62881819-62881819 on Build GRCh38
rs745490114 Chr.20: 62881920-62881920 on Build GRCh38
rs758127215 Chr.20: 62881983-62881983 on Build GRCh38
rs781182899 Chr.20: 62882064-62882064 on Build GRCh38
rs369592108 Chr.20: 62881812-62881812 on Build GRCh38
rs767964314 Chr.20: 62881891-62881891 on Build GRCh38
rs375864108 Chr.20: 62881921-62881921 on Build GRCh38
rs769795711 Chr.20: 62882083-62882091 on Build GRCh38
rs987399397 Chr.20: 62881862-62881862 on Build GRCh38
rs754735447 Chr.20: 62882132-62882132 on Build GRCh38
rs369813858 Chr.20: 62881938-62881938 on Build GRCh38
rs770639541 Chr.20: 62881829-62881829 on Build GRCh38
rs778429998 Chr.20: 62881971-62881971 on Build GRCh38
rs569832850 Chr.20: 62882038-62882038 on Build GRCh38
rs781311308 Chr.20: 62881999-62881999 on Build GRCh38
rs778554789 Chr.20: 62881908-62881908 on Build GRCh38
rs146732987 Chr.20: 62881816-62881816 on Build GRCh38
rs374696200 Chr.20: 62882075-62882086 on Build GRCh38
rs747559296 Chr.20: 62881780-62881809 on Build GRCh38
rs372485756 Chr.20: 62881834-62881834 on Build GRCh38
rs779016748 Chr.20: 62881699-62881699 on Build GRCh38
rs774713444 Chr.20: 62881850-62881850 on Build GRCh38
rs112034091 Chr.20: 62881705-62881705 on Build GRCh38
rs1029278124 Chr.20: 62881998-62881999 on Build GRCh38
rs767479028 Chr.20: 62881694-62881694 on Build GRCh38
rs753349222 Chr.20: 62881898-62881898 on Build GRCh38
rs750624326 Chr.20: 62881704-62881704 on Build GRCh38
rs767359068 Chr.20: 62881743-62881743 on Build GRCh38
rs75790071 Chr.20: 62881941-62881941 on Build GRCh38
rs143994206 Chr.20: 62881969-62881969 on Build GRCh38
rs201393387 Chr.20: 62881735-62881735 on Build GRCh38
rs747352064 Chr.20: 62881713-62881713 on Build GRCh38
rs777996414 Chr.20: 62881675-62881676 on Build GRCh38
rs116887394 Chr.20: 62881962-62881962 on Build GRCh38
rs8123160 Chr.20: 62882094-62882094 on Build GRCh38
rs1009935680 Chr.20: 62881966-62881966 on Build GRCh38
rs776654184 Chr.20: 62881933-62881933 on Build GRCh38
rs540990053 Chr.20: 62881687-62881687 on Build GRCh38
rs761820036 Chr.20: 62881754-62881754 on Build GRCh38
rs747946873 Chr.20: 62882068-62882068 on Build GRCh38
rs201165565 Chr.20: 62881815-62881815 on Build GRCh38
rs1002227008 Chr.20: 62881899-62881899 on Build GRCh38
rs199561380 Chr.20: 62882083-62882083 on Build GRCh38
rs751133319 Chr.20: 62882061-62882061 on Build GRCh38
rs374108825 Chr.20: 62881695-62881695 on Build GRCh38
rs756191111 Chr.20: 62882074-62882074 on Build GRCh38
rs914358587 Chr.20: 62881765-62881765 on Build GRCh38
rs1020283850 Chr.20: 62881956-62881956 on Build GRCh38

View Full Product Details