Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 18313027-18313570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS15
Entrez Gene ID
5651
Gene Name
transmembrane protease, serine 15
Gene Aliases
ENTK, PRSS7
UniGene
Hs.149473
Gene Chromosome Location
Chr.21: 18269107-18403806 on Build GRCh38
Transcripts
NM_002772.2 XM_011529654.1 XM_011529655.1 XM_011529656.2 XM_011529657.1 XM_011529658.1 XM_011529659.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
323893 c.2036G>A p.R679H Chr.21: 18313074-18313074 on Build GRCh38
4734725 c.2075G>A p.R692Q Chr.21: 18313035-18313035 on Build GRCh38
3991889 c.2080A>C p.R694R Chr.21: 18313030-18313030 on Build GRCh38
1268177 c.2051C>A p.T684K Chr.21: 18313059-18313059 on Build GRCh38
1413448 c.2055G>A p.T685T Chr.21: 18313055-18313055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554702800 Chr.21: 18313207-18313207 on Build GRCh38
rs750285967 Chr.21: 18313056-18313056 on Build GRCh38
rs757525993 Chr.21: 18313078-18313078 on Build GRCh38
rs888282609 Chr.21: 18313436-18313436 on Build GRCh38
rs151065817 Chr.21: 18313386-18313386 on Build GRCh38
rs59702784 Chr.21: 18313350-18313351 on Build GRCh38
rs958908011 Chr.21: 18313501-18313501 on Build GRCh38
rs768905318 Chr.21: 18313124-18313124 on Build GRCh38
rs538350727 Chr.21: 18313492-18313492 on Build GRCh38
rs1028522989 Chr.21: 18313496-18313496 on Build GRCh38
rs1057052092 Chr.21: 18313387-18313387 on Build GRCh38
rs988191671 Chr.21: 18313336-18313339 on Build GRCh38
rs889405237 Chr.21: 18313384-18313384 on Build GRCh38
rs143954180 Chr.21: 18313349-18313349 on Build GRCh38
rs1052009355 Chr.21: 18313383-18313383 on Build GRCh38
rs1016107116 Chr.21: 18313480-18313480 on Build GRCh38
rs114994530 Chr.21: 18313241-18313241 on Build GRCh38
rs1012778436 Chr.21: 18313244-18313244 on Build GRCh38
rs997729500 Chr.21: 18313302-18313302 on Build GRCh38
rs1012749426 Chr.21: 18313428-18313428 on Build GRCh38
rs575644491 Chr.21: 18313351-18313352 on Build GRCh38
rs965777828 Chr.21: 18313284-18313284 on Build GRCh38
rs958148530 Chr.21: 18313336-18313337 on Build GRCh38
rs965394369 Chr.21: 18313491-18313491 on Build GRCh38
rs892926426 Chr.21: 18313138-18313138 on Build GRCh38
rs780771531 Chr.21: 18313091-18313091 on Build GRCh38
rs1046061722 Chr.21: 18313219-18313219 on Build GRCh38
rs6517809 Chr.21: 18313243-18313243 on Build GRCh38
rs747069529 Chr.21: 18313108-18313108 on Build GRCh38
rs545764123 Chr.21: 18313482-18313482 on Build GRCh38
rs945932230 Chr.21: 18313201-18313202 on Build GRCh38
rs550314806 Chr.21: 18313074-18313074 on Build GRCh38
rs921409488 Chr.21: 18313103-18313103 on Build GRCh38
rs765740901 Chr.21: 18313069-18313069 on Build GRCh38
rs947032700 Chr.21: 18313144-18313144 on Build GRCh38
rs145916814 Chr.21: 18313349-18313350 on Build GRCh38
rs981707659 Chr.21: 18313445-18313447 on Build GRCh38
rs765120268 Chr.21: 18313055-18313055 on Build GRCh38
rs932818830 Chr.21: 18313347-18313347 on Build GRCh38
rs550879831 Chr.21: 18313405-18313406 on Build GRCh38
rs894437702 Chr.21: 18313391-18313391 on Build GRCh38
rs941728303 Chr.21: 18313360-18313360 on Build GRCh38
rs1030299321 Chr.21: 18313064-18313064 on Build GRCh38
rs780801754 Chr.21: 18313075-18313075 on Build GRCh38
rs952863114 Chr.21: 18313263-18313263 on Build GRCh38
rs913025962 Chr.21: 18313345-18313345 on Build GRCh38
rs981350233 Chr.21: 18313348-18313351 on Build GRCh38
rs897973097 Chr.21: 18313371-18313371 on Build GRCh38
rs112884868 Chr.21: 18313171-18313171 on Build GRCh38
rs954260771 Chr.21: 18313498-18313498 on Build GRCh38
rs560196876 Chr.21: 18313429-18313429 on Build GRCh38
rs568570679 Chr.21: 18313082-18313082 on Build GRCh38
rs921548312 Chr.21: 18313305-18313305 on Build GRCh38
rs79179727 Chr.21: 18313228-18313228 on Build GRCh38
rs574541242 Chr.21: 18313364-18313364 on Build GRCh38
rs908956135 Chr.21: 18313355-18313355 on Build GRCh38
rs937013876 Chr.21: 18313240-18313240 on Build GRCh38
rs1050122228 Chr.21: 18313135-18313135 on Build GRCh38
rs367616414 Chr.21: 18313125-18313125 on Build GRCh38
rs902850378 Chr.21: 18313217-18313217 on Build GRCh38
rs865918330 Chr.21: 18313132-18313132 on Build GRCh38
rs1015557846 Chr.21: 18313276-18313276 on Build GRCh38
rs371037903 Chr.21: 18313126-18313126 on Build GRCh38
rs989692690 Chr.21: 18313520-18313520 on Build GRCh38
rs944271447 Chr.21: 18313080-18313080 on Build GRCh38
rs557198662 Chr.21: 18313335-18313336 on Build GRCh38
rs879319021 Chr.21: 18313533-18313533 on Build GRCh38
rs752390425 Chr.21: 18313077-18313077 on Build GRCh38
rs188127749 Chr.21: 18313303-18313303 on Build GRCh38
rs991430959 Chr.21: 18313531-18313531 on Build GRCh38
rs1007995773 Chr.21: 18313311-18313311 on Build GRCh38
rs751066756 Chr.21: 18313072-18313072 on Build GRCh38
rs937006157 Chr.21: 18313350-18313351 on Build GRCh38
rs915388029 Chr.21: 18313541-18313541 on Build GRCh38
rs924270043 Chr.21: 18313381-18313381 on Build GRCh38
rs774761117 Chr.21: 18313271-18313273 on Build GRCh38
rs8133003 Chr.21: 18313351-18313351 on Build GRCh38
rs1032425134 Chr.21: 18313335-18313335 on Build GRCh38
rs762968217 Chr.21: 18313066-18313066 on Build GRCh38
rs114931165 Chr.21: 18313438-18313438 on Build GRCh38
rs187735282 Chr.21: 18313353-18313353 on Build GRCh38
rs7281534 Chr.21: 18313088-18313088 on Build GRCh38
rs182961016 Chr.21: 18313254-18313254 on Build GRCh38
rs772560060 Chr.21: 18313088-18313091 on Build GRCh38
rs983281780 Chr.21: 18313272-18313272 on Build GRCh38

View Full Product Details