Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30932460-30932968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MORC2
Entrez Gene ID
22880
Gene Name
MORC family CW-type zinc finger 2
Gene Aliases
CMT2Z, ZCW3, ZCWCC1
UniGene
Hs.555918
Gene Chromosome Location
Chr.22: 30925130-30968497 on Build GRCh38
Transcripts
NM_001303256.2 NM_001303257.2 NM_014941.3 XM_011530004.1 XM_017028667.1 XM_017028668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3553424 c.2474C>T p.S825F Chr.22: 30932632-30932632 on Build GRCh38
2937508 c.2618T>A p.V873E Chr.22: 30932674-30932674 on Build GRCh38
6714316 c.2534G>A p.G845D Chr.22: 30932758-30932758 on Build GRCh38
7377431 c.2704A>G p.S902G Chr.22: 30932588-30932588 on Build GRCh38
6161960 c.2745C>G p.L915L Chr.22: 30932547-30932547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041799887 Chr.22: 30932857-30932857 on Build GRCh38
rs146999378 Chr.22: 30932783-30932783 on Build GRCh38
rs776286972 Chr.22: 30932638-30932638 on Build GRCh38
rs565441094 Chr.22: 30932625-30932625 on Build GRCh38
rs375446261 Chr.22: 30932809-30932809 on Build GRCh38
rs774384535 Chr.22: 30932878-30932878 on Build GRCh38
rs974685925 Chr.22: 30932545-30932545 on Build GRCh38
rs200687252 Chr.22: 30932854-30932854 on Build GRCh38
rs369960820 Chr.22: 30932839-30932839 on Build GRCh38
rs775277249 Chr.22: 30932749-30932749 on Build GRCh38
rs764410932 Chr.22: 30932844-30932844 on Build GRCh38
rs199686987 Chr.22: 30932872-30932872 on Build GRCh38
rs751197811 Chr.22: 30932553-30932553 on Build GRCh38
rs547761745 Chr.22: 30932673-30932673 on Build GRCh38
rs751164695 Chr.22: 30932590-30932590 on Build GRCh38
rs11549247 Chr.22: 30932562-30932562 on Build GRCh38
rs973620342 Chr.22: 30932817-30932817 on Build GRCh38
rs773717593 Chr.22: 30932795-30932795 on Build GRCh38
rs375217252 Chr.22: 30932682-30932682 on Build GRCh38
rs757399034 Chr.22: 30932842-30932842 on Build GRCh38
rs765643613 Chr.22: 30932818-30932818 on Build GRCh38
rs753682452 Chr.22: 30932489-30932489 on Build GRCh38
rs772878146 Chr.22: 30932806-30932806 on Build GRCh38
rs954359222 Chr.22: 30932768-30932768 on Build GRCh38
rs779496791 Chr.22: 30932587-30932588 on Build GRCh38
rs753118579 Chr.22: 30932838-30932838 on Build GRCh38
rs766203288 Chr.22: 30932936-30932936 on Build GRCh38
rs369585779 Chr.22: 30932729-30932729 on Build GRCh38
rs368417146 Chr.22: 30932539-30932539 on Build GRCh38
rs147997704 Chr.22: 30932568-30932568 on Build GRCh38
rs141374674 Chr.22: 30932583-30932583 on Build GRCh38
rs754127740 Chr.22: 30932669-30932669 on Build GRCh38
rs140142437 Chr.22: 30932709-30932709 on Build GRCh38
rs766313848 Chr.22: 30932487-30932487 on Build GRCh38
rs372297066 Chr.22: 30932642-30932642 on Build GRCh38
rs747135816 Chr.22: 30932805-30932805 on Build GRCh38
rs764479914 Chr.22: 30932943-30932943 on Build GRCh38
rs373048769 Chr.22: 30932744-30932744 on Build GRCh38
rs761366027 Chr.22: 30932650-30932650 on Build GRCh38
rs762499940 Chr.22: 30932584-30932584 on Build GRCh38
rs773604531 Chr.22: 30932556-30932556 on Build GRCh38
rs772615451 Chr.22: 30932906-30932906 on Build GRCh38
rs202243108 Chr.22: 30932614-30932614 on Build GRCh38
rs568585811 Chr.22: 30932825-30932825 on Build GRCh38
rs201090445 Chr.22: 30932624-30932624 on Build GRCh38
rs773690053 Chr.22: 30932733-30932733 on Build GRCh38
rs767254485 Chr.22: 30932659-30932659 on Build GRCh38
rs927128734 Chr.22: 30932828-30932828 on Build GRCh38
rs747144945 Chr.22: 30932626-30932626 on Build GRCh38
rs772397976 Chr.22: 30932551-30932551 on Build GRCh38
rs781161903 Chr.22: 30932853-30932853 on Build GRCh38
rs746210102 Chr.22: 30932505-30932505 on Build GRCh38
rs939369167 Chr.22: 30932821-30932821 on Build GRCh38
rs978049954 Chr.22: 30932774-30932774 on Build GRCh38
rs777461768 Chr.22: 30932500-30932500 on Build GRCh38
rs907589141 Chr.22: 30932807-30932807 on Build GRCh38
rs754967937 Chr.22: 30932491-30932491 on Build GRCh38
rs764596581 Chr.22: 30932496-30932496 on Build GRCh38
rs749698880 Chr.22: 30932518-30932518 on Build GRCh38
rs373417061 Chr.22: 30932841-30932841 on Build GRCh38
rs771192309 Chr.22: 30932632-30932632 on Build GRCh38
rs748062673 Chr.22: 30932891-30932891 on Build GRCh38
rs775447036 Chr.22: 30932645-30932645 on Build GRCh38
rs143046823 Chr.22: 30932687-30932687 on Build GRCh38
rs67357185 Chr.22: 30932539-30932540 on Build GRCh38
rs548088557 Chr.22: 30932757-30932757 on Build GRCh38
rs756946772 Chr.22: 30932605-30932605 on Build GRCh38
rs780488751 Chr.22: 30932507-30932507 on Build GRCh38
rs904594126 Chr.22: 30932861-30932861 on Build GRCh38
rs372199572 Chr.22: 30932648-30932648 on Build GRCh38
rs371685129 Chr.22: 30932607-30932607 on Build GRCh38
rs150496681 Chr.22: 30932615-30932615 on Build GRCh38
rs557895027 Chr.22: 30932867-30932867 on Build GRCh38
rs770586964 Chr.22: 30932567-30932567 on Build GRCh38
rs769208894 Chr.22: 30932522-30932522 on Build GRCh38
rs41279971 Chr.22: 30932942-30932942 on Build GRCh38
rs759328437 Chr.22: 30932576-30932576 on Build GRCh38
rs762899928 Chr.22: 30932647-30932647 on Build GRCh38
rs141633101 Chr.22: 30932587-30932587 on Build GRCh38
rs138350528 Chr.22: 30932675-30932675 on Build GRCh38
rs1042426757 Chr.22: 30932581-30932581 on Build GRCh38
rs376197429 Chr.22: 30932529-30932529 on Build GRCh38
rs765227719 Chr.22: 30932577-30932577 on Build GRCh38
rs146132479 Chr.22: 30932688-30932688 on Build GRCh38
rs374410153 Chr.22: 30932564-30932564 on Build GRCh38
rs763289871 Chr.22: 30932530-30932531 on Build GRCh38
rs753210391 Chr.22: 30932678-30932678 on Build GRCh38
rs768646218 Chr.22: 30932793-30932793 on Build GRCh38
rs189421529 Chr.22: 30932643-30932643 on Build GRCh38
rs771558349 Chr.22: 30932893-30932893 on Build GRCh38
rs748203997 Chr.22: 30932618-30932618 on Build GRCh38
rs200411634 Chr.22: 30932498-30932498 on Build GRCh38
rs771046855 Chr.22: 30932721-30932721 on Build GRCh38
rs755312428 Chr.22: 30932488-30932488 on Build GRCh38
rs768556129 Chr.22: 30932877-30932877 on Build GRCh38
rs760341830 Chr.22: 30932907-30932907 on Build GRCh38
rs761432963 Chr.22: 30932796-30932796 on Build GRCh38

View Full Product Details