Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 126451250-126451755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZXDC
Entrez Gene ID
79364
Gene Name
ZXD family zinc finger C
Gene Aliases
ZXDL
UniGene
Hs.440049
Gene Chromosome Location
Chr.3: 126437348-126475938 on Build GRCh38
Transcripts
NM_025112.4 NR_104249.1 XM_005247757.3 XM_006713741.2 XM_011513119.2

SNP Detail

SNP ID Chromosome Location
rs918577383 Chr.3: 126451345-126451345 on Build GRCh38
rs748124678 Chr.3: 126451433-126451433 on Build GRCh38
rs746974352 Chr.3: 126451631-126451631 on Build GRCh38
rs879303546 Chr.3: 126451410-126451410 on Build GRCh38
rs971665810 Chr.3: 126451660-126451660 on Build GRCh38
rs917070380 Chr.3: 126451300-126451300 on Build GRCh38
rs909412097 Chr.3: 126451586-126451586 on Build GRCh38
rs575461302 Chr.3: 126451282-126451282 on Build GRCh38
rs1032646392 Chr.3: 126451605-126451605 on Build GRCh38
rs867766009 Chr.3: 126451314-126451314 on Build GRCh38
rs1007209318 Chr.3: 126451702-126451702 on Build GRCh38
rs187900671 Chr.3: 126451619-126451619 on Build GRCh38
rs1018153795 Chr.3: 126451703-126451703 on Build GRCh38
rs867206702 Chr.3: 126451606-126451606 on Build GRCh38
rs183549537 Chr.3: 126451449-126451449 on Build GRCh38
rs982199485 Chr.3: 126451672-126451672 on Build GRCh38
rs775371292 Chr.3: 126451551-126451551 on Build GRCh38
rs1039834344 Chr.3: 126451413-126451413 on Build GRCh38
rs942954561 Chr.3: 126451398-126451398 on Build GRCh38
rs559778542 Chr.3: 126451534-126451534 on Build GRCh38
rs552114832 Chr.3: 126451710-126451710 on Build GRCh38
rs982617258 Chr.3: 126451341-126451341 on Build GRCh38
rs148217197 Chr.3: 126451495-126451495 on Build GRCh38
rs111858888 Chr.3: 126451482-126451482 on Build GRCh38
rs991303036 Chr.3: 126451280-126451280 on Build GRCh38
rs1055420134 Chr.3: 126451623-126451623 on Build GRCh38
rs544113870 Chr.3: 126451397-126451397 on Build GRCh38
rs532223557 Chr.3: 126451687-126451687 on Build GRCh38
rs965073246 Chr.3: 126451431-126451431 on Build GRCh38
rs777921468 Chr.3: 126451436-126451436 on Build GRCh38
rs1024111401 Chr.3: 126451643-126451643 on Build GRCh38
rs1013500898 Chr.3: 126451632-126451632 on Build GRCh38
rs999593396 Chr.3: 126451681-126451681 on Build GRCh38
rs895143137 Chr.3: 126451625-126451625 on Build GRCh38
rs772896223 Chr.3: 126451610-126451610 on Build GRCh38
rs901356946 Chr.3: 126451422-126451422 on Build GRCh38
rs570169253 Chr.3: 126451475-126451475 on Build GRCh38
rs929983412 Chr.3: 126451367-126451367 on Build GRCh38
rs769809936 Chr.3: 126451699-126451699 on Build GRCh38
rs1013266275 Chr.3: 126451629-126451629 on Build GRCh38
rs1046399138 Chr.3: 126451640-126451640 on Build GRCh38
rs745659734 Chr.3: 126451304-126451304 on Build GRCh38
rs976421144 Chr.3: 126451469-126451469 on Build GRCh38
rs951353380 Chr.3: 126451706-126451706 on Build GRCh38
rs942942699 Chr.3: 126451731-126451731 on Build GRCh38
rs902576481 Chr.3: 126451653-126451653 on Build GRCh38
rs1048673716 Chr.3: 126451396-126451396 on Build GRCh38
rs569518633 Chr.3: 126451579-126451579 on Build GRCh38
rs999859714 Chr.3: 126451507-126451507 on Build GRCh38
rs1048334169 Chr.3: 126451564-126451564 on Build GRCh38
rs1032187756 Chr.3: 126451324-126451324 on Build GRCh38
rs56021247 Chr.3: 126451722-126451722 on Build GRCh38
rs999291007 Chr.3: 126451305-126451305 on Build GRCh38

View Full Product Details