Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 115029652-115030154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSHB
Entrez Gene ID
7252
Gene Name
thyroid stimulating hormone beta
Gene Aliases
TSH-B, TSH-BETA
UniGene
Hs.406687
Gene Chromosome Location
Chr.1: 115029824-115035935 on Build GRCh38
Transcripts
NM_000549.4

SNP Detail

SNP ID Chromosome Location
rs983415841 Chr.1: 115029763-115029763 on Build GRCh38
rs949442808 Chr.1: 115029772-115029772 on Build GRCh38
rs945334027 Chr.1: 115029891-115029891 on Build GRCh38
rs900258745 Chr.1: 115029991-115029991 on Build GRCh38
rs553866876 Chr.1: 115029787-115029787 on Build GRCh38
rs749959881 Chr.1: 115030059-115030059 on Build GRCh38
rs757821387 Chr.1: 115029705-115029705 on Build GRCh38
rs185178055 Chr.1: 115029878-115029878 on Build GRCh38
rs893105499 Chr.1: 115029948-115029948 on Build GRCh38
rs1048832767 Chr.1: 115029865-115029865 on Build GRCh38
rs530241569 Chr.1: 115030043-115030043 on Build GRCh38
rs577131274 Chr.1: 115029960-115029960 on Build GRCh38
rs56137624 Chr.1: 115030096-115030096 on Build GRCh38
rs948446819 Chr.1: 115030060-115030061 on Build GRCh38
rs548415245 Chr.1: 115030070-115030070 on Build GRCh38
rs540959873 Chr.1: 115029986-115029986 on Build GRCh38
rs573351184 Chr.1: 115029983-115029983 on Build GRCh38
rs1053051755 Chr.1: 115029911-115029911 on Build GRCh38
rs908553516 Chr.1: 115029766-115029766 on Build GRCh38
rs549795828 Chr.1: 115029730-115029730 on Build GRCh38
rs989949785 Chr.1: 115030072-115030072 on Build GRCh38
rs77331078 Chr.1: 115029849-115029849 on Build GRCh38
rs945233604 Chr.1: 115030112-115030112 on Build GRCh38
rs778511248 Chr.1: 115029998-115029998 on Build GRCh38
rs1321108 Chr.1: 115029744-115029744 on Build GRCh38
rs1042671312 Chr.1: 115029896-115029896 on Build GRCh38
rs911798942 Chr.1: 115029774-115029774 on Build GRCh38
rs554574053 Chr.1: 115029868-115029868 on Build GRCh38
rs181432291 Chr.1: 115030032-115030032 on Build GRCh38
rs868604790 Chr.1: 115029714-115029714 on Build GRCh38
rs971940994 Chr.1: 115030028-115030028 on Build GRCh38
rs982957872 Chr.1: 115030019-115030019 on Build GRCh38
rs866065701 Chr.1: 115029715-115029715 on Build GRCh38
rs1027172140 Chr.1: 115030004-115030004 on Build GRCh38
rs952992283 Chr.1: 115029753-115029753 on Build GRCh38
rs952962107 Chr.1: 115030018-115030018 on Build GRCh38
rs982112126 Chr.1: 115030055-115030055 on Build GRCh38
rs970951450 Chr.1: 115030036-115030036 on Build GRCh38
rs189829460 Chr.1: 115029887-115029887 on Build GRCh38
rs971741551 Chr.1: 115029793-115029793 on Build GRCh38
rs149406514 Chr.1: 115030126-115030126 on Build GRCh38
rs76566517 Chr.1: 115029882-115029882 on Build GRCh38
rs999492486 Chr.1: 115029910-115029911 on Build GRCh38
rs916101749 Chr.1: 115029867-115029867 on Build GRCh38
rs538771827 Chr.1: 115029786-115029786 on Build GRCh38
rs187984951 Chr.1: 115029719-115029719 on Build GRCh38
rs748392493 Chr.1: 115029898-115029898 on Build GRCh38
rs536404403 Chr.1: 115029857-115029857 on Build GRCh38
rs1022093365 Chr.1: 115029754-115029754 on Build GRCh38
rs191988426 Chr.1: 115029721-115029721 on Build GRCh38

View Full Product Details