Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8212622-8213129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3TC1
Entrez Gene ID
54436
Gene Name
SH3 domain and tetratricopeptide repeats 1
Gene Aliases
-
UniGene
Hs.479116
Gene Chromosome Location
Chr.4: 8182072-8241103 on Build GRCh38
Transcripts
NM_001318480.1 NM_018986.4 NR_134639.1 XM_011513488.1 XM_011513489.2 XM_017008300.1 XM_017008301.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1694133 c.278G>A p.R93K Chr.4: 8212731-8212731 on Build GRCh38
6397632 c.314C>A p.T105N Chr.4: 8212767-8212767 on Build GRCh38
1265506 c.289C>T p.L97L Chr.4: 8212742-8212742 on Build GRCh38
3918253 c.249C>T p.D83D Chr.4: 8212702-8212702 on Build GRCh38
1310302 c.252G>C p.L84L Chr.4: 8212705-8212705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982395684 Chr.4: 8212935-8212935 on Build GRCh38
rs987886084 Chr.4: 8213036-8213036 on Build GRCh38
rs1000426295 Chr.4: 8212997-8212997 on Build GRCh38
rs755220961 Chr.4: 8212761-8212761 on Build GRCh38
rs1009335928 Chr.4: 8213091-8213091 on Build GRCh38
rs547880929 Chr.4: 8212727-8212727 on Build GRCh38
rs766610685 Chr.4: 8212819-8212819 on Build GRCh38
rs930562077 Chr.4: 8212938-8212938 on Build GRCh38
rs748977972 Chr.4: 8212785-8212785 on Build GRCh38
rs930491266 Chr.4: 8213105-8213105 on Build GRCh38
rs11734194 Chr.4: 8212717-8212717 on Build GRCh38
rs566154689 Chr.4: 8212762-8212762 on Build GRCh38
rs751533591 Chr.4: 8212674-8212674 on Build GRCh38
rs567397185 Chr.4: 8212806-8212806 on Build GRCh38
rs747473671 Chr.4: 8212778-8212778 on Build GRCh38
rs145155067 Chr.4: 8212967-8212967 on Build GRCh38
rs1003622025 Chr.4: 8212960-8212960 on Build GRCh38
rs530209470 Chr.4: 8213029-8213029 on Build GRCh38
rs755416883 Chr.4: 8212700-8212700 on Build GRCh38
rs1712287 Chr.4: 8212909-8212909 on Build GRCh38
rs140999243 Chr.4: 8212807-8212807 on Build GRCh38
rs145592188 Chr.4: 8212760-8212760 on Build GRCh38
rs752108493 Chr.4: 8212841-8212841 on Build GRCh38
rs149980258 Chr.4: 8212966-8212966 on Build GRCh38
rs567413991 Chr.4: 8213041-8213041 on Build GRCh38
rs538409960 Chr.4: 8212911-8212911 on Build GRCh38
rs368647715 Chr.4: 8212837-8212837 on Build GRCh38
rs768187416 Chr.4: 8212805-8212805 on Build GRCh38
rs146325907 Chr.4: 8212942-8212942 on Build GRCh38
rs750110227 Chr.4: 8213033-8213033 on Build GRCh38
rs776802403 Chr.4: 8212975-8212975 on Build GRCh38
rs911933886 Chr.4: 8213037-8213037 on Build GRCh38
rs372665200 Chr.4: 8212686-8212686 on Build GRCh38
rs747292766 Chr.4: 8213051-8213051 on Build GRCh38
rs995340450 Chr.4: 8212793-8212793 on Build GRCh38
rs200305611 Chr.4: 8212818-8212818 on Build GRCh38
rs542403040 Chr.4: 8212893-8212893 on Build GRCh38
rs370117010 Chr.4: 8212663-8212663 on Build GRCh38
rs760873262 Chr.4: 8212754-8212754 on Build GRCh38
rs780811426 Chr.4: 8212720-8212720 on Build GRCh38
rs372342036 Chr.4: 8212746-8212746 on Build GRCh38
rs1025521665 Chr.4: 8212642-8212642 on Build GRCh38
rs924647987 Chr.4: 8212658-8212658 on Build GRCh38
rs762540603 Chr.4: 8212728-8212728 on Build GRCh38
rs370874852 Chr.4: 8212724-8212724 on Build GRCh38
rs886132686 Chr.4: 8212751-8212751 on Build GRCh38
rs758717410 Chr.4: 8212839-8212839 on Build GRCh38
rs1038191351 Chr.4: 8212952-8212952 on Build GRCh38
rs141307214 Chr.4: 8212687-8212687 on Build GRCh38
rs1056182418 Chr.4: 8212961-8212961 on Build GRCh38
rs377293008 Chr.4: 8212817-8212817 on Build GRCh38
rs752988270 Chr.4: 8212772-8212772 on Build GRCh38
rs754077279 Chr.4: 8212699-8212699 on Build GRCh38
rs769312435 Chr.4: 8212779-8212779 on Build GRCh38
rs184763432 Chr.4: 8212773-8212773 on Build GRCh38
rs142579744 Chr.4: 8212820-8212820 on Build GRCh38
rs1052310477 Chr.4: 8212683-8212683 on Build GRCh38
rs773983242 Chr.4: 8212745-8212745 on Build GRCh38
rs374037428 Chr.4: 8212769-8212769 on Build GRCh38
rs957795768 Chr.4: 8213081-8213081 on Build GRCh38
rs748731885 Chr.4: 8212713-8212713 on Build GRCh38
rs938031270 Chr.4: 8212946-8212946 on Build GRCh38
rs148270559 Chr.4: 8212654-8212654 on Build GRCh38
rs572093415 Chr.4: 8212875-8212875 on Build GRCh38
rs757117502 Chr.4: 8212678-8212678 on Build GRCh38
rs139648827 Chr.4: 8212954-8212954 on Build GRCh38
rs1032360286 Chr.4: 8213002-8213002 on Build GRCh38
rs1032080484 Chr.4: 8213075-8213075 on Build GRCh38
rs779125439 Chr.4: 8212702-8212702 on Build GRCh38
rs988336589 Chr.4: 8212842-8212842 on Build GRCh38
rs571899759 Chr.4: 8213017-8213017 on Build GRCh38
rs766518746 Chr.4: 8212758-8212758 on Build GRCh38
rs1023071125 Chr.4: 8212974-8212974 on Build GRCh38
rs920395335 Chr.4: 8213089-8213089 on Build GRCh38
rs756207650 Chr.4: 8212694-8212694 on Build GRCh38
rs765038398 Chr.4: 8212825-8212826 on Build GRCh38
rs745381976 Chr.4: 8212721-8212721 on Build GRCh38
rs1712288 Chr.4: 8213040-8213040 on Build GRCh38
rs202092857 Chr.4: 8212826-8212826 on Build GRCh38
rs531422086 Chr.4: 8213042-8213042 on Build GRCh38
rs1008533920 Chr.4: 8213031-8213031 on Build GRCh38
rs780432334 Chr.4: 8212697-8212697 on Build GRCh38
rs139277479 Chr.4: 8212784-8212784 on Build GRCh38
rs1038881873 Chr.4: 8212910-8212910 on Build GRCh38
rs559825284 Chr.4: 8212979-8212979 on Build GRCh38
rs756260540 Chr.4: 8212753-8212753 on Build GRCh38
rs768486125 Chr.4: 8212737-8212737 on Build GRCh38

View Full Product Details