Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 46909081-46909591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX7B2
Entrez Gene ID
170712
Gene Name
cytochrome c oxidase subunit 7B2
Gene Aliases
-
UniGene
Hs.479656
Gene Chromosome Location
Chr.4: 46734827-46909266 on Build GRCh38
Transcripts
NM_130902.2 XM_011513630.2 XM_011513631.2 XM_011513632.2 XM_011513633.2 XM_011513634.2 XM_011513635.2 XM_011513637.2 XM_011513638.2

SNP Detail

SNP ID Chromosome Location
rs887391660 Chr.4: 46909444-46909444 on Build GRCh38
rs961626954 Chr.4: 46909338-46909338 on Build GRCh38
rs114153598 Chr.4: 46909566-46909566 on Build GRCh38
rs34129155 Chr.4: 46909218-46909219 on Build GRCh38
rs532506308 Chr.4: 46909553-46909553 on Build GRCh38
rs975465612 Chr.4: 46909209-46909209 on Build GRCh38
rs67592528 Chr.4: 46909283-46909283 on Build GRCh38
rs1027970463 Chr.4: 46909306-46909306 on Build GRCh38
rs574714331 Chr.4: 46909496-46909496 on Build GRCh38
rs184146276 Chr.4: 46909563-46909563 on Build GRCh38
rs572248491 Chr.4: 46909522-46909522 on Build GRCh38
rs576731692 Chr.4: 46909340-46909340 on Build GRCh38
rs955636364 Chr.4: 46909144-46909144 on Build GRCh38
rs931021689 Chr.4: 46909429-46909429 on Build GRCh38
rs890823805 Chr.4: 46909310-46909310 on Build GRCh38
rs906974745 Chr.4: 46909259-46909259 on Build GRCh38
rs1008270153 Chr.4: 46909529-46909529 on Build GRCh38
rs1001705206 Chr.4: 46909271-46909271 on Build GRCh38
rs145452041 Chr.4: 46909533-46909533 on Build GRCh38
rs990057371 Chr.4: 46909332-46909332 on Build GRCh38
rs189810790 Chr.4: 46909167-46909167 on Build GRCh38
rs550622637 Chr.4: 46909195-46909195 on Build GRCh38
rs537644098 Chr.4: 46909448-46909448 on Build GRCh38
rs938096252 Chr.4: 46909196-46909196 on Build GRCh38
rs567041559 Chr.4: 46909284-46909284 on Build GRCh38
rs925267170 Chr.4: 46909314-46909314 on Build GRCh38
rs961571192 Chr.4: 46909190-46909190 on Build GRCh38
rs781049524 Chr.4: 46909393-46909393 on Build GRCh38
rs995682094 Chr.4: 46909280-46909280 on Build GRCh38
rs959433171 Chr.4: 46909368-46909368 on Build GRCh38
rs17641171 Chr.4: 46909192-46909192 on Build GRCh38
rs903114274 Chr.4: 46909234-46909234 on Build GRCh38
rs981348440 Chr.4: 46909296-46909296 on Build GRCh38
rs139945777 Chr.4: 46909502-46909502 on Build GRCh38
rs191949334 Chr.4: 46909503-46909503 on Build GRCh38
rs779389535 Chr.4: 46909358-46909358 on Build GRCh38
rs376155950 Chr.4: 46909173-46909173 on Build GRCh38
rs1054374647 Chr.4: 46909214-46909214 on Build GRCh38
rs771476483 Chr.4: 46909539-46909539 on Build GRCh38
rs975919053 Chr.4: 46909312-46909312 on Build GRCh38
rs991338331 Chr.4: 46909372-46909375 on Build GRCh38
rs890887081 Chr.4: 46909526-46909526 on Build GRCh38
rs913034583 Chr.4: 46909215-46909215 on Build GRCh38
rs1035002338 Chr.4: 46909109-46909109 on Build GRCh38
rs370890528 Chr.4: 46909468-46909468 on Build GRCh38
rs1028417562 Chr.4: 46909127-46909127 on Build GRCh38
rs949980416 Chr.4: 46909219-46909219 on Build GRCh38
rs937068208 Chr.4: 46909407-46909407 on Build GRCh38
rs1027138614 Chr.4: 46909552-46909552 on Build GRCh38
rs945895689 Chr.4: 46909239-46909239 on Build GRCh38
rs913191548 Chr.4: 46909335-46909335 on Build GRCh38
rs376324957 Chr.4: 46909400-46909401 on Build GRCh38
rs1055960726 Chr.4: 46909417-46909417 on Build GRCh38
rs912688686 Chr.4: 46909193-46909193 on Build GRCh38
rs533829781 Chr.4: 46909289-46909289 on Build GRCh38
rs185326815 Chr.4: 46909143-46909143 on Build GRCh38
rs1015288497 Chr.4: 46909534-46909534 on Build GRCh38
rs898155313 Chr.4: 46909426-46909426 on Build GRCh38
rs964130377 Chr.4: 46909297-46909297 on Build GRCh38
rs987482830 Chr.4: 46909153-46909153 on Build GRCh38
rs917923373 Chr.4: 46909372-46909372 on Build GRCh38
rs558269717 Chr.4: 46909317-46909317 on Build GRCh38
rs900000724 Chr.4: 46909486-46909486 on Build GRCh38
rs748492514 Chr.4: 46909432-46909432 on Build GRCh38
rs949451427 Chr.4: 46909387-46909387 on Build GRCh38
rs951558936 Chr.4: 46909183-46909183 on Build GRCh38
rs1046101532 Chr.4: 46909233-46909233 on Build GRCh38
rs996180156 Chr.4: 46909164-46909164 on Build GRCh38
rs9992220 Chr.4: 46909362-46909362 on Build GRCh38
rs773735402 Chr.4: 46909509-46909509 on Build GRCh38
rs1045505782 Chr.4: 46909241-46909241 on Build GRCh38
rs935908839 Chr.4: 46909211-46909211 on Build GRCh38
rs892994802 Chr.4: 46909494-46909494 on Build GRCh38
rs536423711 Chr.4: 46909274-46909274 on Build GRCh38
rs898158493 Chr.4: 46909275-46909275 on Build GRCh38
rs546034479 Chr.4: 46909524-46909524 on Build GRCh38
rs768364288 Chr.4: 46909379-46909379 on Build GRCh38
rs147619112 Chr.4: 46909210-46909210 on Build GRCh38
rs1019754361 Chr.4: 46909451-46909451 on Build GRCh38

View Full Product Details