Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 64248878-64249376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF679
Entrez Gene ID
168417
Gene Name
zinc finger protein 679
Gene Aliases
-
UniGene
Hs.520884
Gene Chromosome Location
Chr.7: 64228474-64266931 on Build GRCh38
Transcripts
NM_153363.2 XM_017011797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3085027 c.23C>G p.P8R Chr.7: 64249140-64249140 on Build GRCh38
371989 c.15G>C p.P5P Chr.7: 64249132-64249132 on Build GRCh38
3950693 c.24T>G p.P8P Chr.7: 64249141-64249141 on Build GRCh38
3412225 c.31C>A p.R11R Chr.7: 64249148-64249148 on Build GRCh38
4449955 c.39+1G>A p.? Chr.7: 64249157-64249157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186800690 Chr.7: 64249071-64249071 on Build GRCh38
rs762935077 Chr.7: 64249161-64249161 on Build GRCh38
rs760270593 Chr.7: 64249228-64249228 on Build GRCh38
rs1009946193 Chr.7: 64248960-64248960 on Build GRCh38
rs897833543 Chr.7: 64249188-64249188 on Build GRCh38
rs1034069868 Chr.7: 64248995-64248995 on Build GRCh38
rs775231255 Chr.7: 64249208-64249208 on Build GRCh38
rs758807040 Chr.7: 64249195-64249196 on Build GRCh38
rs757775142 Chr.7: 64249117-64249117 on Build GRCh38
rs966799452 Chr.7: 64249276-64249276 on Build GRCh38
rs546628495 Chr.7: 64248948-64248948 on Build GRCh38
rs535620208 Chr.7: 64248937-64248937 on Build GRCh38
rs1028157302 Chr.7: 64249211-64249211 on Build GRCh38
rs1007806529 Chr.7: 64249287-64249287 on Build GRCh38
rs570579532 Chr.7: 64249194-64249194 on Build GRCh38
rs148745340 Chr.7: 64249177-64249177 on Build GRCh38
rs774275736 Chr.7: 64249166-64249166 on Build GRCh38
rs142547963 Chr.7: 64249344-64249344 on Build GRCh38
rs752275976 Chr.7: 64249190-64249191 on Build GRCh38
rs753928240 Chr.7: 64249176-64249176 on Build GRCh38
rs748250233 Chr.7: 64248912-64248912 on Build GRCh38
rs751414091 Chr.7: 64249093-64249093 on Build GRCh38
rs527692929 Chr.7: 64249323-64249323 on Build GRCh38
rs982831901 Chr.7: 64249042-64249042 on Build GRCh38
rs759288329 Chr.7: 64249167-64249167 on Build GRCh38
rs768482949 Chr.7: 64249070-64249070 on Build GRCh38
rs144272859 Chr.7: 64249089-64249089 on Build GRCh38
rs779224016 Chr.7: 64249190-64249190 on Build GRCh38
rs766168914 Chr.7: 64249163-64249163 on Build GRCh38
rs746179983 Chr.7: 64249132-64249132 on Build GRCh38
rs1037374798 Chr.7: 64249308-64249308 on Build GRCh38
rs1055458270 Chr.7: 64249291-64249291 on Build GRCh38
rs995708689 Chr.7: 64249159-64249159 on Build GRCh38
rs897785782 Chr.7: 64249321-64249321 on Build GRCh38
rs535422178 Chr.7: 64249336-64249336 on Build GRCh38
rs746818260 Chr.7: 64249030-64249030 on Build GRCh38
rs1027276452 Chr.7: 64249036-64249036 on Build GRCh38
rs565328516 Chr.7: 64249235-64249235 on Build GRCh38
rs764234104 Chr.7: 64249174-64249174 on Build GRCh38
rs191648017 Chr.7: 64249148-64249148 on Build GRCh38
rs779781972 Chr.7: 64248999-64248999 on Build GRCh38
rs962176067 Chr.7: 64248964-64248964 on Build GRCh38
rs780601882 Chr.7: 64249199-64249199 on Build GRCh38
rs758115949 Chr.7: 64248954-64248954 on Build GRCh38
rs374951221 Chr.7: 64249045-64249045 on Build GRCh38
rs958109320 Chr.7: 64249196-64249196 on Build GRCh38
rs1054987197 Chr.7: 64249289-64249289 on Build GRCh38
rs921314447 Chr.7: 64249065-64249065 on Build GRCh38
rs779327113 Chr.7: 64249130-64249130 on Build GRCh38
rs966004001 Chr.7: 64249307-64249307 on Build GRCh38
rs754779231 Chr.7: 64249097-64249097 on Build GRCh38
rs977748789 Chr.7: 64249255-64249255 on Build GRCh38
rs906982738 Chr.7: 64248976-64248976 on Build GRCh38
rs931032342 Chr.7: 64249304-64249304 on Build GRCh38
rs376632500 Chr.7: 64249219-64249219 on Build GRCh38
rs760403459 Chr.7: 64249069-64249069 on Build GRCh38
rs1002093582 Chr.7: 64248918-64248918 on Build GRCh38
rs117470203 Chr.7: 64249109-64249109 on Build GRCh38
rs952232042 Chr.7: 64249018-64249018 on Build GRCh38
rs539414514 Chr.7: 64249245-64249245 on Build GRCh38
rs925085434 Chr.7: 64249140-64249140 on Build GRCh38
rs761364448 Chr.7: 64249075-64249075 on Build GRCh38
rs767657990 Chr.7: 64249100-64249100 on Build GRCh38
rs995632514 Chr.7: 64249028-64249028 on Build GRCh38
rs549832967 Chr.7: 64249279-64249279 on Build GRCh38
rs762835450 Chr.7: 64249088-64249088 on Build GRCh38
rs772573145 Chr.7: 64249136-64249136 on Build GRCh38
rs767324781 Chr.7: 64249170-64249170 on Build GRCh38
rs937823182 Chr.7: 64249288-64249288 on Build GRCh38
rs958866985 Chr.7: 64249001-64249001 on Build GRCh38
rs925055726 Chr.7: 64249282-64249282 on Build GRCh38
rs1019647046 Chr.7: 64249301-64249301 on Build GRCh38
rs975917384 Chr.7: 64249057-64249057 on Build GRCh38
rs965768070 Chr.7: 64249189-64249189 on Build GRCh38
rs1004352145 Chr.7: 64249329-64249329 on Build GRCh38
rs993572765 Chr.7: 64249192-64249192 on Build GRCh38
rs773175625 Chr.7: 64249085-64249085 on Build GRCh38
rs752894994 Chr.7: 64249172-64249172 on Build GRCh38
rs541171498 Chr.7: 64249067-64249067 on Build GRCh38
rs867078648 Chr.7: 64249110-64249110 on Build GRCh38
rs1024806618 Chr.7: 64248963-64248963 on Build GRCh38
rs756139591 Chr.7: 64249106-64249106 on Build GRCh38
rs182240624 Chr.7: 64248984-64248984 on Build GRCh38
rs1004889226 Chr.7: 64249187-64249187 on Build GRCh38
rs772664590 Chr.7: 64249157-64249157 on Build GRCh38
rs569919852 Chr.7: 64249297-64249297 on Build GRCh38
rs563979238 Chr.7: 64249104-64249104 on Build GRCh38
rs887286204 Chr.7: 64249316-64249316 on Build GRCh38
rs576281472 Chr.7: 64248919-64248919 on Build GRCh38
rs780403377 Chr.7: 64249285-64249285 on Build GRCh38
rs368170297 Chr.7: 64249052-64249052 on Build GRCh38
rs951601810 Chr.7: 64249039-64249039 on Build GRCh38
rs913542340 Chr.7: 64249247-64249247 on Build GRCh38
rs879473587 Chr.7: 64249154-64249154 on Build GRCh38
rs769312900 Chr.7: 64249155-64249155 on Build GRCh38
rs769389374 Chr.7: 64249206-64249206 on Build GRCh38
rs373495804 Chr.7: 64249223-64249223 on Build GRCh38
rs564242090 Chr.7: 64249068-64249068 on Build GRCh38
rs746397099 Chr.7: 64249191-64249191 on Build GRCh38
rs1033596738 Chr.7: 64248926-64248926 on Build GRCh38
rs75059739 Chr.7: 64248947-64248947 on Build GRCh38
rs994059081 Chr.7: 64248977-64248977 on Build GRCh38
rs917826929 Chr.7: 64249293-64249293 on Build GRCh38
rs903760569 Chr.7: 64248904-64248904 on Build GRCh38
rs778150581 Chr.7: 64249021-64249021 on Build GRCh38
rs747794749 Chr.7: 64249203-64249203 on Build GRCh38
rs745552363 Chr.7: 64249149-64249149 on Build GRCh38
rs559743213 Chr.7: 64249131-64249131 on Build GRCh38
rs764673619 Chr.7: 64249076-64249076 on Build GRCh38

View Full Product Details