Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100105148-100105650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4M1
Entrez Gene ID
9179
Gene Name
adaptor related protein complex 4 mu 1 subunit
Gene Aliases
CPSQ3, MU-4, MU-ARP2, SPG50
UniGene
Hs.632317
Gene Chromosome Location
Chr.7: 100101413-100107180 on Build GRCh38
Transcripts
NM_004722.3 XM_005250689.4 XM_005250690.4 XM_006716175.3 XM_011516685.2 XM_017012790.1 XM_017012791.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6565794 c.826C>T p.Q276* Chr.7: 100105338-100105338 on Build GRCh38
3715865 c.799C>G p.H267D Chr.7: 100105311-100105311 on Build GRCh38
297846 c.733_736delGGGC p.G245fs*14 Chr.7: 100105245-100105248 on Build GRCh38
3762938 c.812G>A p.R271H Chr.7: 100105324-100105324 on Build GRCh38
266017 c.861C>T p.S287S Chr.7: 100105471-100105471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200552467 Chr.7: 100105545-100105545 on Build GRCh38
rs930269909 Chr.7: 100105587-100105587 on Build GRCh38
rs907295366 Chr.7: 100105509-100105509 on Build GRCh38
rs759324120 Chr.7: 100105223-100105223 on Build GRCh38
rs979414211 Chr.7: 100105485-100105485 on Build GRCh38
rs542768500 Chr.7: 100105530-100105530 on Build GRCh38
rs895548740 Chr.7: 100105321-100105321 on Build GRCh38
rs369306233 Chr.7: 100105384-100105384 on Build GRCh38
rs201580004 Chr.7: 100105490-100105490 on Build GRCh38
rs376551171 Chr.7: 100105228-100105228 on Build GRCh38
rs551847933 Chr.7: 100105577-100105577 on Build GRCh38
rs553061919 Chr.7: 100105273-100105273 on Build GRCh38
rs759651013 Chr.7: 100105498-100105498 on Build GRCh38
rs768778642 Chr.7: 100105500-100105500 on Build GRCh38
rs370260221 Chr.7: 100105239-100105239 on Build GRCh38
rs561103710 Chr.7: 100105375-100105375 on Build GRCh38
rs1026112326 Chr.7: 100105507-100105507 on Build GRCh38
rs545459309 Chr.7: 100105283-100105283 on Build GRCh38
rs927923217 Chr.7: 100105175-100105175 on Build GRCh38
rs766717023 Chr.7: 100105429-100105429 on Build GRCh38
rs937805871 Chr.7: 100105176-100105176 on Build GRCh38
rs565167866 Chr.7: 100105403-100105403 on Build GRCh38
rs780030221 Chr.7: 100105314-100105314 on Build GRCh38
rs575096086 Chr.7: 100105457-100105457 on Build GRCh38
rs769502788 Chr.7: 100105263-100105263 on Build GRCh38
rs369874184 Chr.7: 100105406-100105406 on Build GRCh38
rs777352543 Chr.7: 100105343-100105343 on Build GRCh38
rs759428856 Chr.7: 100105278-100105278 on Build GRCh38
rs748639289 Chr.7: 100105218-100105219 on Build GRCh38
rs759838658 Chr.7: 100105510-100105510 on Build GRCh38
rs776788025 Chr.7: 100105450-100105451 on Build GRCh38
rs768541738 Chr.7: 100105440-100105440 on Build GRCh38
rs769652536 Chr.7: 100105505-100105505 on Build GRCh38
rs781616312 Chr.7: 100105323-100105323 on Build GRCh38
rs565296810 Chr.7: 100105315-100105315 on Build GRCh38
rs142325884 Chr.7: 100105471-100105471 on Build GRCh38
rs373696169 Chr.7: 100105254-100105254 on Build GRCh38
rs772113588 Chr.7: 100105512-100105512 on Build GRCh38
rs750601796 Chr.7: 100105246-100105246 on Build GRCh38
rs773785064 Chr.7: 100105354-100105354 on Build GRCh38
rs754225835 Chr.7: 100105483-100105483 on Build GRCh38
rs767611212 Chr.7: 100105472-100105472 on Build GRCh38
rs779704427 Chr.7: 100105563-100105563 on Build GRCh38
rs745310821 Chr.7: 100105412-100105412 on Build GRCh38
rs757649099 Chr.7: 100105488-100105488 on Build GRCh38
rs745807118 Chr.7: 100105355-100105355 on Build GRCh38
rs774897281 Chr.7: 100105219-100105220 on Build GRCh38
rs753222064 Chr.7: 100105393-100105393 on Build GRCh38
rs767953033 Chr.7: 100105392-100105392 on Build GRCh38
rs767244572 Chr.7: 100105224-100105224 on Build GRCh38
rs759950294 Chr.7: 100105177-100105177 on Build GRCh38
rs897456893 Chr.7: 100105332-100105332 on Build GRCh38
rs750012862 Chr.7: 100105539-100105539 on Build GRCh38
rs373162905 Chr.7: 100105538-100105538 on Build GRCh38
rs774646136 Chr.7: 100105470-100105470 on Build GRCh38
rs144253083 Chr.7: 100105289-100105289 on Build GRCh38
rs534701383 Chr.7: 100105610-100105610 on Build GRCh38
rs765352573 Chr.7: 100105178-100105178 on Build GRCh38
rs147670505 Chr.7: 100105325-100105325 on Build GRCh38
rs771629792 Chr.7: 100105418-100105418 on Build GRCh38
rs757605059 Chr.7: 100105232-100105232 on Build GRCh38
rs368830741 Chr.7: 100105258-100105258 on Build GRCh38
rs971782025 Chr.7: 100105299-100105299 on Build GRCh38
rs1042146797 Chr.7: 100105226-100105226 on Build GRCh38
rs200726009 Chr.7: 100105267-100105267 on Build GRCh38
rs200900793 Chr.7: 100105405-100105405 on Build GRCh38
rs377672447 Chr.7: 100105207-100105207 on Build GRCh38
rs570256454 Chr.7: 100105495-100105495 on Build GRCh38
rs772522035 Chr.7: 100105219-100105224 on Build GRCh38
rs775577262 Chr.7: 100105367-100105367 on Build GRCh38
rs747839303 Chr.7: 100105504-100105504 on Build GRCh38
rs953631200 Chr.7: 100105362-100105362 on Build GRCh38
rs775520329 Chr.7: 100105421-100105421 on Build GRCh38
rs781303911 Chr.7: 100105257-100105257 on Build GRCh38
rs368267562 Chr.7: 100105352-100105352 on Build GRCh38
rs751045786 Chr.7: 100105577-100105577 on Build GRCh38
rs1027881207 Chr.7: 100105351-100105351 on Build GRCh38
rs990666465 Chr.7: 100105622-100105622 on Build GRCh38
rs766443912 Chr.7: 100105467-100105467 on Build GRCh38
rs1051745199 Chr.7: 100105253-100105253 on Build GRCh38
rs771974744 Chr.7: 100105360-100105360 on Build GRCh38
rs139122007 Chr.7: 100105515-100105515 on Build GRCh38
rs752745814 Chr.7: 100105262-100105262 on Build GRCh38
rs145887263 Chr.7: 100105252-100105252 on Build GRCh38
rs766269157 Chr.7: 100105208-100105208 on Build GRCh38
rs748967782 Chr.7: 100105344-100105344 on Build GRCh38
rs750687666 Chr.7: 100105298-100105298 on Build GRCh38
rs982409621 Chr.7: 100105172-100105172 on Build GRCh38
rs371795483 Chr.7: 100105394-100105394 on Build GRCh38
rs755494077 Chr.7: 100105319-100105319 on Build GRCh38
rs376000948 Chr.7: 100105378-100105378 on Build GRCh38
rs375129284 Chr.7: 100105216-100105216 on Build GRCh38
rs199982051 Chr.7: 100105499-100105499 on Build GRCh38
rs374565045 Chr.7: 100105422-100105422 on Build GRCh38
rs762387733 Chr.7: 100105266-100105266 on Build GRCh38
rs962896326 Chr.7: 100105265-100105265 on Build GRCh38
rs902147115 Chr.7: 100105230-100105230 on Build GRCh38
rs371973205 Chr.7: 100105376-100105376 on Build GRCh38
rs774391984 Chr.7: 100105508-100105508 on Build GRCh38
rs772766548 Chr.7: 100105202-100105202 on Build GRCh38
rs775602399 Chr.7: 100105516-100105516 on Build GRCh38
rs1048780951 Chr.7: 100105623-100105631 on Build GRCh38
rs781278962 Chr.7: 100105501-100105501 on Build GRCh38
rs766467478 Chr.7: 100105380-100105380 on Build GRCh38
rs41280968 Chr.7: 100105324-100105324 on Build GRCh38
rs192270933 Chr.7: 100105489-100105489 on Build GRCh38
rs971452596 Chr.7: 100105171-100105171 on Build GRCh38
rs946448880 Chr.7: 100105218-100105218 on Build GRCh38
rs994513548 Chr.7: 100105338-100105338 on Build GRCh38
rs758175403 Chr.7: 100105549-100105549 on Build GRCh38
rs377269608 Chr.7: 100105458-100105458 on Build GRCh38
rs754386660 Chr.7: 100105527-100105527 on Build GRCh38
rs369459721 Chr.7: 100105526-100105526 on Build GRCh38
rs773729417 Chr.7: 100105219-100105222 on Build GRCh38
rs756660680 Chr.7: 100105477-100105477 on Build GRCh38
rs774302896 Chr.7: 100105274-100105274 on Build GRCh38
rs753168955 Chr.7: 100105474-100105474 on Build GRCh38
rs761168776 Chr.7: 100105525-100105525 on Build GRCh38
rs760489210 Chr.7: 100105371-100105371 on Build GRCh38
rs117820338 Chr.7: 100105390-100105390 on Build GRCh38
rs764431556 Chr.7: 100105482-100105482 on Build GRCh38

View Full Product Details