Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159857629-159858152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VSIG8
Entrez Gene ID
391123
Gene Name
V-set and immunoglobulin domain containing 8
Gene Aliases
-
UniGene
Hs.647718
Gene Chromosome Location
Chr.1: 159854316-159862657 on Build GRCh38
Transcripts
NM_001013661.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4647371 c.624G>A p.Q208Q Chr.1: 159857773-159857773 on Build GRCh38
1335397 c.511G>A p.G171R Chr.1: 159857886-159857886 on Build GRCh38
4740620 c.522G>A p.Q174Q Chr.1: 159857875-159857875 on Build GRCh38
4617152 c.567C>A p.P189P Chr.1: 159857830-159857830 on Build GRCh38
1270307 c.385A>G p.K129E Chr.1: 159858135-159858135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376402680 Chr.1: 159857932-159857932 on Build GRCh38
rs776077431 Chr.1: 159857839-159857839 on Build GRCh38
rs766010179 Chr.1: 159857868-159857869 on Build GRCh38
rs368238440 Chr.1: 159858041-159858041 on Build GRCh38
rs764354396 Chr.1: 159857725-159857725 on Build GRCh38
rs1034991015 Chr.1: 159857661-159857661 on Build GRCh38
rs191695804 Chr.1: 159857709-159857709 on Build GRCh38
rs753341544 Chr.1: 159858109-159858109 on Build GRCh38
rs780703384 Chr.1: 159858061-159858062 on Build GRCh38
rs757081885 Chr.1: 159857736-159857736 on Build GRCh38
rs370695926 Chr.1: 159857987-159857987 on Build GRCh38
rs146657303 Chr.1: 159857836-159857836 on Build GRCh38
rs750153493 Chr.1: 159857881-159857881 on Build GRCh38
rs367756324 Chr.1: 159858100-159858100 on Build GRCh38
rs770014534 Chr.1: 159858075-159858075 on Build GRCh38
rs962409888 Chr.1: 159858130-159858130 on Build GRCh38
rs766556889 Chr.1: 159858086-159858086 on Build GRCh38
rs747828713 Chr.1: 159857810-159857810 on Build GRCh38
rs780102611 Chr.1: 159857780-159857780 on Build GRCh38
rs749002103 Chr.1: 159858010-159858010 on Build GRCh38
rs746162076 Chr.1: 159857933-159857933 on Build GRCh38
rs112455697 Chr.1: 159858114-159858114 on Build GRCh38
rs764305063 Chr.1: 159857844-159857844 on Build GRCh38
rs747439868 Chr.1: 159858056-159858056 on Build GRCh38
rs755634394 Chr.1: 159858000-159858000 on Build GRCh38
rs975377048 Chr.1: 159857804-159857804 on Build GRCh38
rs751349846 Chr.1: 159857992-159857992 on Build GRCh38
rs748859745 Chr.1: 159858087-159858087 on Build GRCh38
rs750177640 Chr.1: 159857883-159857883 on Build GRCh38
rs375901772 Chr.1: 159858083-159858083 on Build GRCh38
rs746435603 Chr.1: 159857796-159857796 on Build GRCh38
rs754015695 Chr.1: 159857867-159857867 on Build GRCh38
rs752716387 Chr.1: 159857719-159857719 on Build GRCh38
rs747314848 Chr.1: 159858124-159858124 on Build GRCh38
rs777244351 Chr.1: 159858003-159858003 on Build GRCh38
rs756224539 Chr.1: 159857720-159857720 on Build GRCh38
rs752198145 Chr.1: 159857997-159857997 on Build GRCh38
rs377592150 Chr.1: 159857888-159857888 on Build GRCh38
rs372070298 Chr.1: 159857774-159857774 on Build GRCh38
rs377276625 Chr.1: 159857718-159857718 on Build GRCh38
rs368321634 Chr.1: 159857841-159857841 on Build GRCh38
rs769422803 Chr.1: 159857929-159857929 on Build GRCh38
rs745836177 Chr.1: 159857694-159857694 on Build GRCh38
rs772510529 Chr.1: 159857934-159857934 on Build GRCh38
rs1031776544 Chr.1: 159858036-159858036 on Build GRCh38
rs41264827 Chr.1: 159857957-159857957 on Build GRCh38
rs775822199 Chr.1: 159857940-159857940 on Build GRCh38
rs779730402 Chr.1: 159858030-159858030 on Build GRCh38
rs761216329 Chr.1: 159857952-159857952 on Build GRCh38
rs376771902 Chr.1: 159857825-159857825 on Build GRCh38
rs777306859 Chr.1: 159858108-159858108 on Build GRCh38
rs754136409 Chr.1: 159857734-159857734 on Build GRCh38
rs755583040 Chr.1: 159858104-159858104 on Build GRCh38
rs140295916 Chr.1: 159857914-159857914 on Build GRCh38
rs150177506 Chr.1: 159858113-159858113 on Build GRCh38
rs778823958 Chr.1: 159857771-159857771 on Build GRCh38
rs372910764 Chr.1: 159857985-159857985 on Build GRCh38
rs757533419 Chr.1: 159857880-159857880 on Build GRCh38
rs768221659 Chr.1: 159857798-159857798 on Build GRCh38
rs754443619 Chr.1: 159857913-159857913 on Build GRCh38
rs200943268 Chr.1: 159857853-159857853 on Build GRCh38
rs575224629 Chr.1: 159857651-159857651 on Build GRCh38
rs11584200 Chr.1: 159857685-159857685 on Build GRCh38
rs145862475 Chr.1: 159857816-159857816 on Build GRCh38
rs774842804 Chr.1: 159857700-159857700 on Build GRCh38
rs541229576 Chr.1: 159857768-159857768 on Build GRCh38
rs757602362 Chr.1: 159857738-159857739 on Build GRCh38
rs759392553 Chr.1: 159858099-159858099 on Build GRCh38
rs760014309 Chr.1: 159857830-159857830 on Build GRCh38
rs780447637 Chr.1: 159858044-159858044 on Build GRCh38
rs2494503 Chr.1: 159857666-159857666 on Build GRCh38
rs765485143 Chr.1: 159857971-159857971 on Build GRCh38
rs773263677 Chr.1: 159858082-159858082 on Build GRCh38
rs1014396647 Chr.1: 159857674-159857674 on Build GRCh38
rs756922614 Chr.1: 159858110-159858110 on Build GRCh38
rs919709190 Chr.1: 159857889-159857889 on Build GRCh38
rs746985048 Chr.1: 159857894-159857894 on Build GRCh38
rs772089749 Chr.1: 159857698-159857698 on Build GRCh38
rs752653947 Chr.1: 159858102-159858102 on Build GRCh38
rs747705889 Chr.1: 159857922-159857922 on Build GRCh38
rs768187218 Chr.1: 159857701-159857701 on Build GRCh38
rs79128166 Chr.1: 159857963-159857963 on Build GRCh38
rs751355665 Chr.1: 159858092-159858092 on Build GRCh38
rs754495567 Chr.1: 159857881-159857882 on Build GRCh38
rs776214466 Chr.1: 159857805-159857805 on Build GRCh38
rs745727406 Chr.1: 159857773-159857773 on Build GRCh38
rs750856815 Chr.1: 159857797-159857797 on Build GRCh38
rs561162388 Chr.1: 159857717-159857717 on Build GRCh38
rs201522497 Chr.1: 159857948-159857948 on Build GRCh38
rs368817560 Chr.1: 159857961-159857961 on Build GRCh38
rs777279375 Chr.1: 159858061-159858061 on Build GRCh38
rs371126436 Chr.1: 159858068-159858068 on Build GRCh38
rs968896835 Chr.1: 159857785-159857785 on Build GRCh38
rs769285512 Chr.1: 159858060-159858060 on Build GRCh38
rs751188187 Chr.1: 159857877-159857877 on Build GRCh38
rs750666450 Chr.1: 159857984-159857984 on Build GRCh38
rs754885556 Chr.1: 159857996-159857996 on Build GRCh38
rs764729595 Chr.1: 159857772-159857772 on Build GRCh38
rs766750688 Chr.1: 159857712-159857712 on Build GRCh38
rs1016628954 Chr.1: 159857754-159857754 on Build GRCh38
rs1002800604 Chr.1: 159857665-159857665 on Build GRCh38

View Full Product Details