Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32987987-32988483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APTX
Entrez Gene ID
54840
Gene Name
aprataxin
Gene Aliases
AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT
UniGene
Hs.20158
Gene Chromosome Location
Chr.9: 32883872-33025129 on Build GRCh38
Transcripts
NM_001195248.1 NM_001195249.1 NM_001195250.1 NM_001195251.1 NM_001195252.1 NM_001195254.1 NM_175069.2 NM_175073.2 NR_036576.1 NR_036577.1 NR_036578.1 NR_036579.1 XM_006716791.3 XM_006716792.2 XM_011517938.1 XM_011517939.2 XM_017014831.1 XM_017014832.1 XM_017014833.1 XM_017014834.1 XM_017014835.1 XM_017014836.1 XM_017014837.1 XM_017014838.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3996514 c.211A>T p.K71* Chr.9: 32988094-32988094 on Build GRCh38
2772394 c.212A>G p.K71R Chr.9: 32988093-32988093 on Build GRCh38
1462040 c.176A>C p.K59T Chr.9: 32988087-32988087 on Build GRCh38
2772393 c.170A>G p.K57R Chr.9: 32988093-32988093 on Build GRCh38
1245250 c.168C>T p.V56V Chr.9: 32988095-32988095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057519198 Chr.9: 32988099-32988099 on Build GRCh38
rs755083054 Chr.9: 32988113-32988114 on Build GRCh38
rs765752653 Chr.9: 32988062-32988062 on Build GRCh38
rs141053938 Chr.9: 32988352-32988352 on Build GRCh38
rs79744073 Chr.9: 32988048-32988048 on Build GRCh38
rs556048747 Chr.9: 32988049-32988049 on Build GRCh38
rs570123260 Chr.9: 32988429-32988429 on Build GRCh38
rs17226291 Chr.9: 32988187-32988187 on Build GRCh38
rs192017512 Chr.9: 32988401-32988401 on Build GRCh38
rs368922494 Chr.9: 32988157-32988157 on Build GRCh38
rs1008736950 Chr.9: 32988024-32988024 on Build GRCh38
rs192342726 Chr.9: 32988175-32988175 on Build GRCh38
rs1888867 Chr.9: 32988405-32988405 on Build GRCh38
rs748761458 Chr.9: 32988288-32988288 on Build GRCh38
rs377603034 Chr.9: 32988050-32988050 on Build GRCh38
rs995713662 Chr.9: 32988404-32988404 on Build GRCh38
rs764458907 Chr.9: 32988201-32988201 on Build GRCh38
rs917279247 Chr.9: 32988430-32988430 on Build GRCh38
rs200939883 Chr.9: 32988163-32988163 on Build GRCh38
rs763391606 Chr.9: 32988179-32988179 on Build GRCh38
rs369431727 Chr.9: 32988115-32988115 on Build GRCh38
rs188796889 Chr.9: 32988303-32988303 on Build GRCh38
rs890777738 Chr.9: 32988346-32988346 on Build GRCh38
rs1049976978 Chr.9: 32988276-32988276 on Build GRCh38
rs983523238 Chr.9: 32988427-32988427 on Build GRCh38
rs1000682699 Chr.9: 32988059-32988059 on Build GRCh38
rs10971271 Chr.9: 32988457-32988457 on Build GRCh38
rs1009936127 Chr.9: 32988301-32988301 on Build GRCh38
rs758718457 Chr.9: 32988164-32988165 on Build GRCh38
rs747872887 Chr.9: 32988438-32988438 on Build GRCh38
rs778102109 Chr.9: 32988179-32988180 on Build GRCh38
rs780105237 Chr.9: 32988131-32988131 on Build GRCh38
rs940223750 Chr.9: 32988198-32988198 on Build GRCh38
rs577830048 Chr.9: 32988415-32988415 on Build GRCh38
rs749913379 Chr.9: 32988051-32988051 on Build GRCh38
rs113391831 Chr.9: 32988141-32988141 on Build GRCh38
rs74922775 Chr.9: 32988343-32988343 on Build GRCh38
rs184407340 Chr.9: 32988188-32988188 on Build GRCh38
rs572063713 Chr.9: 32988093-32988093 on Build GRCh38
rs1038496817 Chr.9: 32988218-32988218 on Build GRCh38
rs779023417 Chr.9: 32988162-32988163 on Build GRCh38
rs955329812 Chr.9: 32988067-32988067 on Build GRCh38
rs375978921 Chr.9: 32988140-32988140 on Build GRCh38
rs752912612 Chr.9: 32988164-32988164 on Build GRCh38
rs757431868 Chr.9: 32988103-32988103 on Build GRCh38
rs200146043 Chr.9: 32988162-32988162 on Build GRCh38
rs927505570 Chr.9: 32988458-32988458 on Build GRCh38
rs772793361 Chr.9: 32988145-32988145 on Build GRCh38
rs751955477 Chr.9: 32988236-32988236 on Build GRCh38
rs1000733170 Chr.9: 32988420-32988420 on Build GRCh38
rs769809499 Chr.9: 32988121-32988121 on Build GRCh38
rs950812117 Chr.9: 32988422-32988422 on Build GRCh38
rs932899788 Chr.9: 32988239-32988239 on Build GRCh38
rs4324990 Chr.9: 32988310-32988310 on Build GRCh38
rs548155770 Chr.9: 32988190-32988190 on Build GRCh38
rs745911325 Chr.9: 32988118-32988118 on Build GRCh38
rs533248843 Chr.9: 32988164-32988164 on Build GRCh38
rs771709792 Chr.9: 32988143-32988143 on Build GRCh38
rs985334100 Chr.9: 32988069-32988069 on Build GRCh38
rs767459701 Chr.9: 32988044-32988046 on Build GRCh38
rs760129710 Chr.9: 32988058-32988058 on Build GRCh38
rs959194649 Chr.9: 32988443-32988443 on Build GRCh38
rs891505742 Chr.9: 32988250-32988250 on Build GRCh38
rs750499052 Chr.9: 32988043-32988044 on Build GRCh38
rs758759074 Chr.9: 32988085-32988085 on Build GRCh38
rs751933830 Chr.9: 32988094-32988094 on Build GRCh38
rs761547802 Chr.9: 32988142-32988142 on Build GRCh38
rs753242838 Chr.9: 32988070-32988070 on Build GRCh38
rs568365108 Chr.9: 32988222-32988222 on Build GRCh38
rs1018584224 Chr.9: 32988368-32988368 on Build GRCh38

View Full Product Details